Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 30026602-30027096 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FKBP14
Entrez Gene ID
55033
Gene Name
FK506 binding protein 14
Gene Aliases
EDSKMH, FKBP22, IPBP12
UniGene
Hs.390838
Gene Chromosome Location
Chr.7: 30010583-30026801 on Build GRCh38
Transcripts
NM_017946.3 NR_046478.1 NR_046479.1

SNP Detail

SNP ID Chromosome Location
rs764128464 Chr.7: 30026790-30026790 on Build GRCh38
rs1052015183 Chr.7: 30026830-30026830 on Build GRCh38
rs956046464 Chr.7: 30026850-30026850 on Build GRCh38
rs954600164 Chr.7: 30026657-30026657 on Build GRCh38
rs867687062 Chr.7: 30026633-30026633 on Build GRCh38
rs899919740 Chr.7: 30026783-30026783 on Build GRCh38
rs1021750419 Chr.7: 30026866-30026866 on Build GRCh38
rs966406365 Chr.7: 30026627-30026627 on Build GRCh38
rs1035614186 Chr.7: 30026956-30026956 on Build GRCh38
rs990259838 Chr.7: 30026648-30026648 on Build GRCh38
rs980620718 Chr.7: 30026884-30026884 on Build GRCh38
rs902110798 Chr.7: 30026824-30026824 on Build GRCh38
rs956162576 Chr.7: 30026634-30026634 on Build GRCh38
rs1018234072 Chr.7: 30026791-30026791 on Build GRCh38
rs926506054 Chr.7: 30026901-30026901 on Build GRCh38
rs542225973 Chr.7: 30027014-30027014 on Build GRCh38
rs77872461 Chr.7: 30027016-30027016 on Build GRCh38
rs536669735 Chr.7: 30026658-30026658 on Build GRCh38
rs887529620 Chr.7: 30026965-30026965 on Build GRCh38
rs553249096 Chr.7: 30026829-30026829 on Build GRCh38
rs577643727 Chr.7: 30026888-30026888 on Build GRCh38
rs929009839 Chr.7: 30026944-30026944 on Build GRCh38
rs1032376761 Chr.7: 30027063-30027063 on Build GRCh38
rs915254446 Chr.7: 30026757-30026757 on Build GRCh38
rs1014649528 Chr.7: 30026878-30026878 on Build GRCh38
rs138252952 Chr.7: 30026907-30026907 on Build GRCh38
rs71766273 Chr.7: 30026971-30026972 on Build GRCh38
rs146327266 Chr.7: 30026767-30026767 on Build GRCh38
rs980674886 Chr.7: 30026668-30026668 on Build GRCh38
rs1007690993 Chr.7: 30026787-30026787 on Build GRCh38
rs1056148261 Chr.7: 30026704-30026704 on Build GRCh38
rs549250128 Chr.7: 30026665-30026665 on Build GRCh38
rs998318592 Chr.7: 30026793-30026793 on Build GRCh38
rs1030120928 Chr.7: 30026640-30026640 on Build GRCh38
rs150804085 Chr.7: 30027051-30027051 on Build GRCh38
rs1052505338 Chr.7: 30026739-30026739 on Build GRCh38
rs898836429 Chr.7: 30026881-30026881 on Build GRCh38
rs943516319 Chr.7: 30026981-30026981 on Build GRCh38
rs779480921 Chr.7: 30026713-30026713 on Build GRCh38
rs976111327 Chr.7: 30026632-30026632 on Build GRCh38
rs1031658443 Chr.7: 30026839-30026839 on Build GRCh38
rs963011175 Chr.7: 30026674-30026674 on Build GRCh38
rs142730779 Chr.7: 30027026-30027026 on Build GRCh38
rs946757130 Chr.7: 30026980-30026980 on Build GRCh38
rs967838291 Chr.7: 30026883-30026883 on Build GRCh38
rs142091438 Chr.7: 30026673-30026673 on Build GRCh38
rs918927760 Chr.7: 30026922-30026922 on Build GRCh38
rs975376763 Chr.7: 30027010-30027010 on Build GRCh38
rs990228733 Chr.7: 30026853-30026853 on Build GRCh38
rs1048836846 Chr.7: 30026776-30026776 on Build GRCh38
rs528261003 Chr.7: 30026745-30026745 on Build GRCh38
rs1048766424 Chr.7: 30026948-30026948 on Build GRCh38
rs538321316 Chr.7: 30026906-30026906 on Build GRCh38
rs938993942 Chr.7: 30026699-30026699 on Build GRCh38
rs969624986 Chr.7: 30026700-30026700 on Build GRCh38
rs928096805 Chr.7: 30026712-30026712 on Build GRCh38
rs935393573 Chr.7: 30026731-30026731 on Build GRCh38
rs753549287 Chr.7: 30026684-30026684 on Build GRCh38
rs565090278 Chr.7: 30026957-30026957 on Build GRCh38
rs897515049 Chr.7: 30026711-30026711 on Build GRCh38
rs982323934 Chr.7: 30026941-30026941 on Build GRCh38
rs191552304 Chr.7: 30026923-30026923 on Build GRCh38
rs1042956822 Chr.7: 30026771-30026771 on Build GRCh38
rs946569362 Chr.7: 30026663-30026663 on Build GRCh38
rs900351344 Chr.7: 30026628-30026628 on Build GRCh38
rs756969683 Chr.7: 30026686-30026686 on Build GRCh38
rs973660270 Chr.7: 30026681-30026681 on Build GRCh38

View Full Product Details