Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 36852821-36853317 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELMO1
Entrez Gene ID
9844
Gene Name
engulfment and cell motility 1
Gene Aliases
CED-12, CED12, ELMO-1
UniGene
Hs.434989
Gene Chromosome Location
Chr.7: 36852906-37449409 on Build GRCh38
Transcripts
NM_001039459.2 NM_001206480.2 NM_001206482.1 NM_014800.10 NM_130442.3 NR_038120.1

SNP Detail

SNP ID Chromosome Location
rs567074287 Chr.7: 36852864-36852864 on Build GRCh38
rs904820864 Chr.7: 36853205-36853205 on Build GRCh38
rs1013262417 Chr.7: 36853138-36853138 on Build GRCh38
rs932005138 Chr.7: 36852970-36852970 on Build GRCh38
rs759005796 Chr.7: 36853193-36853193 on Build GRCh38
rs962934871 Chr.7: 36853226-36853226 on Build GRCh38
rs559338577 Chr.7: 36853115-36853115 on Build GRCh38
rs995179144 Chr.7: 36853218-36853218 on Build GRCh38
rs116349636 Chr.7: 36852945-36852945 on Build GRCh38
rs190653914 Chr.7: 36853280-36853280 on Build GRCh38
rs912531774 Chr.7: 36853281-36853281 on Build GRCh38
rs776978088 Chr.7: 36852874-36852879 on Build GRCh38
rs370779724 Chr.7: 36852924-36852924 on Build GRCh38
rs137932493 Chr.7: 36852899-36852899 on Build GRCh38
rs982668929 Chr.7: 36852900-36852900 on Build GRCh38
rs1004434027 Chr.7: 36853261-36853264 on Build GRCh38
rs1028420520 Chr.7: 36853249-36853249 on Build GRCh38
rs912497621 Chr.7: 36853052-36853052 on Build GRCh38
rs774231856 Chr.7: 36853038-36853038 on Build GRCh38
rs569059545 Chr.7: 36852923-36852923 on Build GRCh38
rs369734026 Chr.7: 36852993-36852993 on Build GRCh38
rs374468745 Chr.7: 36853120-36853120 on Build GRCh38
rs925548532 Chr.7: 36853004-36853004 on Build GRCh38
rs542536624 Chr.7: 36853097-36853097 on Build GRCh38
rs142417319 Chr.7: 36852969-36852969 on Build GRCh38
rs978019855 Chr.7: 36853293-36853293 on Build GRCh38
rs907456027 Chr.7: 36853163-36853163 on Build GRCh38
rs974635866 Chr.7: 36853227-36853227 on Build GRCh38
rs544295384 Chr.7: 36852980-36852980 on Build GRCh38
rs908671223 Chr.7: 36852919-36852919 on Build GRCh38
rs932698468 Chr.7: 36853023-36853023 on Build GRCh38
rs554570412 Chr.7: 36853042-36853042 on Build GRCh38
rs202063905 Chr.7: 36853255-36853255 on Build GRCh38
rs865792820 Chr.7: 36852949-36852949 on Build GRCh38
rs1024588974 Chr.7: 36852875-36852875 on Build GRCh38
rs954247312 Chr.7: 36853259-36853259 on Build GRCh38
rs1026906100 Chr.7: 36852877-36852877 on Build GRCh38
rs936500220 Chr.7: 36853104-36853104 on Build GRCh38
rs903732188 Chr.7: 36853092-36853092 on Build GRCh38
rs36078123 Chr.7: 36853208-36853209 on Build GRCh38
rs971608948 Chr.7: 36852880-36852880 on Build GRCh38
rs530842791 Chr.7: 36853157-36853157 on Build GRCh38
rs569630520 Chr.7: 36853068-36853068 on Build GRCh38
rs1019470693 Chr.7: 36853268-36853268 on Build GRCh38
rs1042212931 Chr.7: 36853079-36853079 on Build GRCh38
rs1013650241 Chr.7: 36852872-36852872 on Build GRCh38
rs1024621460 Chr.7: 36853144-36853144 on Build GRCh38
rs945372898 Chr.7: 36853286-36853286 on Build GRCh38
rs1004433765 Chr.7: 36853178-36853178 on Build GRCh38

View Full Product Details