Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 66089134-66089642 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASL
Entrez Gene ID
435
Gene Name
argininosuccinate lyase
Gene Aliases
ASAL
UniGene
Hs.632015
Gene Chromosome Location
Chr.7: 66075789-66093343 on Build GRCh38
Transcripts
NM_000048.3 NM_001024943.1 NM_001024944.1 NM_001024946.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5692213 c.897G>T p.K299N Chr.7: 66089154-66089154 on Build GRCh38
6340614 c.994G>T p.V332L Chr.7: 66089627-66089627 on Build GRCh38
7283280 c.882G>A p.E294E Chr.7: 66089139-66089139 on Build GRCh38
6110757 c.916C>T p.R306W Chr.7: 66089173-66089173 on Build GRCh38
1451835 c.956G>A p.S319N Chr.7: 66089313-66089313 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764356037 Chr.7: 66089335-66089335 on Build GRCh38
rs781331391 Chr.7: 66089180-66089180 on Build GRCh38
rs769506424 Chr.7: 66089282-66089282 on Build GRCh38
rs763844562 Chr.7: 66089573-66089573 on Build GRCh38
rs755245694 Chr.7: 66089174-66089174 on Build GRCh38
rs796051926 Chr.7: 66089612-66089612 on Build GRCh38
rs749306006 Chr.7: 66089249-66089249 on Build GRCh38
rs904475115 Chr.7: 66089474-66089474 on Build GRCh38
rs978132148 Chr.7: 66089242-66089242 on Build GRCh38
rs1002406129 Chr.7: 66089549-66089549 on Build GRCh38
rs755437561 Chr.7: 66089600-66089600 on Build GRCh38
rs560019979 Chr.7: 66089510-66089510 on Build GRCh38
rs375732584 Chr.7: 66089587-66089587 on Build GRCh38
rs111610234 Chr.7: 66089362-66089362 on Build GRCh38
rs774195710 Chr.7: 66089313-66089313 on Build GRCh38
rs370257293 Chr.7: 66089597-66089597 on Build GRCh38
rs752451547 Chr.7: 66089328-66089328 on Build GRCh38
rs748921821 Chr.7: 66089369-66089369 on Build GRCh38
rs748972501 Chr.7: 66089609-66089609 on Build GRCh38
rs151294789 Chr.7: 66089284-66089284 on Build GRCh38
rs1025332601 Chr.7: 66089372-66089372 on Build GRCh38
rs548786563 Chr.7: 66089352-66089352 on Build GRCh38
rs765260769 Chr.7: 66089584-66089584 on Build GRCh38
rs904413062 Chr.7: 66089340-66089340 on Build GRCh38
rs757313920 Chr.7: 66089253-66089253 on Build GRCh38
rs1051586035 Chr.7: 66089233-66089233 on Build GRCh38
rs781673231 Chr.7: 66089604-66089604 on Build GRCh38
rs746354859 Chr.7: 66089271-66089271 on Build GRCh38
rs770579327 Chr.7: 66089614-66089614 on Build GRCh38
rs758359461 Chr.7: 66089162-66089162 on Build GRCh38
rs2460432 Chr.7: 66089398-66089398 on Build GRCh38
rs776057556 Chr.7: 66089264-66089265 on Build GRCh38
rs756280770 Chr.7: 66089243-66089243 on Build GRCh38
rs770425849 Chr.7: 66089187-66089187 on Build GRCh38
rs765801478 Chr.7: 66089291-66089291 on Build GRCh38
rs911946423 Chr.7: 66089421-66089421 on Build GRCh38
rs746380396 Chr.7: 66089361-66089361 on Build GRCh38
rs160647 Chr.7: 66089365-66089365 on Build GRCh38
rs569859973 Chr.7: 66089336-66089336 on Build GRCh38
rs375562374 Chr.7: 66089164-66089164 on Build GRCh38
rs868834862 Chr.7: 66089173-66089173 on Build GRCh38
rs767240274 Chr.7: 66089322-66089322 on Build GRCh38
rs757448120 Chr.7: 66089580-66089580 on Build GRCh38
rs368624250 Chr.7: 66089346-66089346 on Build GRCh38
rs370250705 Chr.7: 66089434-66089434 on Build GRCh38
rs201002874 Chr.7: 66089198-66089198 on Build GRCh38
rs117651205 Chr.7: 66089381-66089381 on Build GRCh38
rs936795106 Chr.7: 66089260-66089260 on Build GRCh38
rs370949043 Chr.7: 66089230-66089230 on Build GRCh38
rs373861796 Chr.7: 66089220-66089220 on Build GRCh38
rs4281011 Chr.7: 66089373-66089373 on Build GRCh38
rs747996363 Chr.7: 66089513-66089513 on Build GRCh38
rs767424019 Chr.7: 66089562-66089562 on Build GRCh38
rs746531359 Chr.7: 66089162-66089163 on Build GRCh38
rs398123129 Chr.7: 66089333-66089333 on Build GRCh38
rs781009876 Chr.7: 66089232-66089232 on Build GRCh38
rs543218393 Chr.7: 66089441-66089441 on Build GRCh38
rs573359642 Chr.7: 66089538-66089538 on Build GRCh38
rs929269027 Chr.7: 66089406-66089406 on Build GRCh38
rs199585001 Chr.7: 66089266-66089266 on Build GRCh38
rs754800257 Chr.7: 66089231-66089231 on Build GRCh38
rs747831550 Chr.7: 66089281-66089281 on Build GRCh38
rs776183045 Chr.7: 66089217-66089217 on Build GRCh38
rs773070338 Chr.7: 66089367-66089367 on Build GRCh38
rs748398698 Chr.7: 66089183-66089183 on Build GRCh38
rs192494428 Chr.7: 66089578-66089578 on Build GRCh38
rs1039259950 Chr.7: 66089464-66089464 on Build GRCh38
rs772932166 Chr.7: 66089345-66089346 on Build GRCh38
rs752110932 Chr.7: 66089599-66089599 on Build GRCh38
rs758840140 Chr.7: 66089594-66089594 on Build GRCh38
rs775871020 Chr.7: 66089280-66089280 on Build GRCh38
rs759495870 Chr.7: 66089321-66089321 on Build GRCh38
rs557593161 Chr.7: 66089412-66089412 on Build GRCh38
rs763407938 Chr.7: 66089328-66089331 on Build GRCh38
rs919377349 Chr.7: 66089227-66089227 on Build GRCh38
rs769307500 Chr.7: 66089227-66089228 on Build GRCh38
rs776178127 Chr.7: 66089195-66089207 on Build GRCh38
rs777809575 Chr.7: 66089189-66089189 on Build GRCh38
rs890289218 Chr.7: 66089263-66089263 on Build GRCh38
rs756230523 Chr.7: 66089272-66089272 on Build GRCh38
rs781367717 Chr.7: 66089488-66089488 on Build GRCh38
rs759270998 Chr.7: 66089200-66089207 on Build GRCh38
rs372611875 Chr.7: 66089188-66089188 on Build GRCh38
rs765914227 Chr.7: 66089308-66089308 on Build GRCh38
rs535066653 Chr.7: 66089346-66089346 on Build GRCh38
rs368492142 Chr.7: 66089229-66089229 on Build GRCh38
rs1024029606 Chr.7: 66089357-66089357 on Build GRCh38
rs375625546 Chr.7: 66089234-66089234 on Build GRCh38
rs891477751 Chr.7: 66089512-66089512 on Build GRCh38
rs764859231 Chr.7: 66089224-66089224 on Build GRCh38
rs370805656 Chr.7: 66089211-66089211 on Build GRCh38
rs923996935 Chr.7: 66089539-66089539 on Build GRCh38
rs556672805 Chr.7: 66089537-66089538 on Build GRCh38
rs751249678 Chr.7: 66089169-66089169 on Build GRCh38
rs1046189066 Chr.7: 66089355-66089355 on Build GRCh38
rs550097103 Chr.7: 66089290-66089290 on Build GRCh38
rs188109158 Chr.7: 66089382-66089382 on Build GRCh38
rs923926957 Chr.7: 66089248-66089248 on Build GRCh38
rs749819433 Chr.7: 66089194-66089194 on Build GRCh38
rs756339957 Chr.7: 66089331-66089331 on Build GRCh38
rs534443154 Chr.7: 66089375-66089375 on Build GRCh38
rs535237668 Chr.7: 66089345-66089345 on Build GRCh38
rs1053004565 Chr.7: 66089505-66089505 on Build GRCh38
rs370109609 Chr.7: 66089571-66089571 on Build GRCh38

View Full Product Details