Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 100676509-100677005 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNB2
Entrez Gene ID
2783
Gene Name
G protein subunit beta 2
Gene Aliases
-
UniGene
Hs.185172
Gene Chromosome Location
Chr.7: 100673740-100679169 on Build GRCh38
Transcripts
NM_005273.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1082917 c.203_203+1insGT p.L69fs*33 Chr.7: 100676799-100676800 on Build GRCh38
1754977 c.83C>T p.S28L Chr.7: 100676560-100676560 on Build GRCh38
1208408 c.155G>A p.R52H Chr.7: 100676751-100676751 on Build GRCh38
1082918 c.203+1_203+2delGT p.? Chr.7: 100676800-100676801 on Build GRCh38
3745418 c.150C>T p.T50T Chr.7: 100676746-100676746 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779406494 Chr.7: 100676815-100676815 on Build GRCh38
rs980294142 Chr.7: 100676929-100676929 on Build GRCh38
rs746433531 Chr.7: 100676755-100676755 on Build GRCh38
rs769730613 Chr.7: 100676950-100676953 on Build GRCh38
rs916990705 Chr.7: 100676616-100676616 on Build GRCh38
rs572501424 Chr.7: 100676865-100676865 on Build GRCh38
rs759744244 Chr.7: 100676794-100676794 on Build GRCh38
rs745726593 Chr.7: 100676837-100676837 on Build GRCh38
rs112720424 Chr.7: 100676945-100676945 on Build GRCh38
rs767563612 Chr.7: 100676803-100676803 on Build GRCh38
rs766607385 Chr.7: 100676684-100676684 on Build GRCh38
rs752827335 Chr.7: 100676806-100676806 on Build GRCh38
rs762772879 Chr.7: 100676580-100676580 on Build GRCh38
rs112493346 Chr.7: 100676855-100676855 on Build GRCh38
rs748960110 Chr.7: 100676646-100676646 on Build GRCh38
rs374972485 Chr.7: 100676587-100676587 on Build GRCh38
rs190637876 Chr.7: 100676843-100676843 on Build GRCh38
rs550319791 Chr.7: 100676938-100676938 on Build GRCh38
rs566505927 Chr.7: 100676650-100676650 on Build GRCh38
rs531870258 Chr.7: 100676936-100676936 on Build GRCh38
rs753206801 Chr.7: 100676704-100676704 on Build GRCh38
rs558986512 Chr.7: 100676849-100676849 on Build GRCh38
rs754508989 Chr.7: 100676811-100676812 on Build GRCh38
rs774090438 Chr.7: 100676651-100676651 on Build GRCh38
rs758639208 Chr.7: 100676585-100676585 on Build GRCh38
rs766083132 Chr.7: 100676799-100676800 on Build GRCh38
rs1033651365 Chr.7: 100676872-100676872 on Build GRCh38
rs767566059 Chr.7: 100676702-100676702 on Build GRCh38
rs761536555 Chr.7: 100676681-100676681 on Build GRCh38
rs996007173 Chr.7: 100676854-100676854 on Build GRCh38
rs901246361 Chr.7: 100676823-100676823 on Build GRCh38
rs998347734 Chr.7: 100676825-100676825 on Build GRCh38
rs776379882 Chr.7: 100676764-100676764 on Build GRCh38
rs948934468 Chr.7: 100676751-100676751 on Build GRCh38
rs768442792 Chr.7: 100676978-100676978 on Build GRCh38
rs971775791 Chr.7: 100676912-100676912 on Build GRCh38
rs555170071 Chr.7: 100676810-100676810 on Build GRCh38
rs908621658 Chr.7: 100676668-100676668 on Build GRCh38
rs778746197 Chr.7: 100676734-100676734 on Build GRCh38
rs775337930 Chr.7: 100676661-100676661 on Build GRCh38
rs958428906 Chr.7: 100676873-100676873 on Build GRCh38
rs144815932 Chr.7: 100676850-100676850 on Build GRCh38
rs771675463 Chr.7: 100676658-100676658 on Build GRCh38
rs768648709 Chr.7: 100676761-100676761 on Build GRCh38
rs377291417 Chr.7: 100676737-100676737 on Build GRCh38
rs762850398 Chr.7: 100676976-100676976 on Build GRCh38
rs369463910 Chr.7: 100676791-100676791 on Build GRCh38
rs778076563 Chr.7: 100676710-100676710 on Build GRCh38
rs561208405 Chr.7: 100676906-100676906 on Build GRCh38
rs75270112 Chr.7: 100676811-100676811 on Build GRCh38
rs971890319 Chr.7: 100676742-100676742 on Build GRCh38
rs776485571 Chr.7: 100676679-100676679 on Build GRCh38
rs774608487 Chr.7: 100676792-100676792 on Build GRCh38
rs763926763 Chr.7: 100676581-100676581 on Build GRCh38
rs373422953 Chr.7: 100676671-100676671 on Build GRCh38
rs771765314 Chr.7: 100676740-100676740 on Build GRCh38
rs750668750 Chr.7: 100676583-100676583 on Build GRCh38
rs543246008 Chr.7: 100676901-100676901 on Build GRCh38
rs750166554 Chr.7: 100676816-100676816 on Build GRCh38
rs748660513 Chr.7: 100676606-100676606 on Build GRCh38
rs775451446 Chr.7: 100676881-100676881 on Build GRCh38
rs377481208 Chr.7: 100676813-100676813 on Build GRCh38
rs757649787 Chr.7: 100676725-100676725 on Build GRCh38
rs760426108 Chr.7: 100676567-100676567 on Build GRCh38
rs113456712 Chr.7: 100676705-100676705 on Build GRCh38
rs768009399 Chr.7: 100676847-100676847 on Build GRCh38
rs370524169 Chr.7: 100676826-100676826 on Build GRCh38
rs952714044 Chr.7: 100676652-100676652 on Build GRCh38
rs779612089 Chr.7: 100676745-100676745 on Build GRCh38
rs748126035 Chr.7: 100676667-100676667 on Build GRCh38
rs546364360 Chr.7: 100676594-100676594 on Build GRCh38
rs992918646 Chr.7: 100676902-100676902 on Build GRCh38
rs754999073 Chr.7: 100676597-100676597 on Build GRCh38
rs542959875 Chr.7: 100676854-100676855 on Build GRCh38
rs113752443 Chr.7: 100676598-100676598 on Build GRCh38
rs766517478 Chr.7: 100676590-100676590 on Build GRCh38
rs936255027 Chr.7: 100676937-100676937 on Build GRCh38
rs533992485 Chr.7: 100676656-100676656 on Build GRCh38
rs1027510902 Chr.7: 100676861-100676861 on Build GRCh38
rs759656094 Chr.7: 100676687-100676687 on Build GRCh38
rs763858229 Chr.7: 100676678-100676678 on Build GRCh38
rs964944096 Chr.7: 100676538-100676538 on Build GRCh38
rs751743759 Chr.7: 100676686-100676686 on Build GRCh38
rs1000302566 Chr.7: 100676586-100676586 on Build GRCh38
rs143353769 Chr.7: 100676713-100676713 on Build GRCh38
rs760614598 Chr.7: 100676809-100676809 on Build GRCh38
rs138290543 Chr.7: 100676800-100676801 on Build GRCh38
rs970231977 Chr.7: 100676918-100676918 on Build GRCh38
rs369726142 Chr.7: 100676970-100676970 on Build GRCh38
rs1005481104 Chr.7: 100676878-100676878 on Build GRCh38
rs756232526 Chr.7: 100676801-100676802 on Build GRCh38
rs369701849 Chr.7: 100676858-100676859 on Build GRCh38
rs756488674 Chr.7: 100676608-100676608 on Build GRCh38
rs984736232 Chr.7: 100676957-100676957 on Build GRCh38
rs186190125 Chr.7: 100676834-100676834 on Build GRCh38
rs753327051 Chr.7: 100676809-100676810 on Build GRCh38
rs755748525 Chr.7: 100676800-100676803 on Build GRCh38
rs917360528 Chr.7: 100676905-100676905 on Build GRCh38

View Full Product Details