Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 103074704-103075211 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARMC10
Entrez Gene ID
83787
Gene Name
armadillo repeat containing 10
Gene Aliases
PNAS-112, PNAS112, PSEC0198, SVH
UniGene
Hs.287412
Gene Chromosome Location
Chr.7: 103074727-103099763 on Build GRCh38
Transcripts
NM_001161009.2 NM_001161010.2 NM_001161011.2 NM_001161012.2 NM_001161013.2 NM_031905.4 XM_005250628.2 XM_011516601.2 XM_017012684.1 XM_017012685.1 XM_017012686.1 XM_017012688.1 XM_017012689.1 NM_001111038.1 NM_001287150.1 NR_105043.1 XM_005250205.3 XM_005250207.3 XM_005250208.3 XM_005250209.2 XM_011515928.2 XM_011515929.2 XM_011515932.2 XM_017011850.1 XM_017011851.1

Gene Symbol
FBXL13
Entrez Gene ID
222235
Gene Name
F-box and leucine rich repeat protein 13
Gene Aliases
DRC6, Fbl13
UniGene
Hs.660029
Gene Chromosome Location
Chr.7: 102699226-103074841 on Build GRCh38
Transcripts
NM_001161009.2 NM_001161010.2 NM_001161011.2 NM_001161012.2 NM_001161013.2 NM_031905.4 XM_005250628.2 XM_011516601.2 XM_017012684.1 XM_017012685.1 XM_017012686.1 XM_017012688.1 XM_017012689.1 NM_001111038.1 NM_001287150.1 NR_105043.1 XM_005250205.3 XM_005250207.3 XM_005250208.3 XM_005250209.2 XM_011515928.2 XM_011515929.2 XM_011515932.2 XM_017011850.1 XM_017011851.1

SNP Detail

SNP ID Chromosome Location
rs1048238448 Chr.7: 103075040-103075040 on Build GRCh38
rs1054848158 Chr.7: 103074931-103074931 on Build GRCh38
rs917357268 Chr.7: 103075192-103075192 on Build GRCh38
rs7797786 Chr.7: 103075010-103075010 on Build GRCh38
rs1014473072 Chr.7: 103074971-103074971 on Build GRCh38
rs895775553 Chr.7: 103075071-103075071 on Build GRCh38
rs778197095 Chr.7: 103074783-103074783 on Build GRCh38
rs956206326 Chr.7: 103075130-103075130 on Build GRCh38
rs564100581 Chr.7: 103075163-103075163 on Build GRCh38
rs899630893 Chr.7: 103074934-103074934 on Build GRCh38
rs937577006 Chr.7: 103075007-103075007 on Build GRCh38
rs528532506 Chr.7: 103074822-103074822 on Build GRCh38
rs993503560 Chr.7: 103074870-103074870 on Build GRCh38
rs754116898 Chr.7: 103074767-103074767 on Build GRCh38
rs996913379 Chr.7: 103074835-103074835 on Build GRCh38
rs921173124 Chr.7: 103075147-103075147 on Build GRCh38
rs533829468 Chr.7: 103075001-103075001 on Build GRCh38
rs1027718435 Chr.7: 103074959-103074959 on Build GRCh38
rs964961938 Chr.7: 103074953-103074953 on Build GRCh38
rs563879411 Chr.7: 103074911-103074912 on Build GRCh38
rs765388114 Chr.7: 103074828-103074828 on Build GRCh38
rs960342616 Chr.7: 103074851-103074851 on Build GRCh38
rs932515318 Chr.7: 103074935-103074935 on Build GRCh38
rs750432233 Chr.7: 103074735-103074735 on Build GRCh38
rs1014709284 Chr.7: 103075076-103075076 on Build GRCh38
rs943946015 Chr.7: 103074919-103074919 on Build GRCh38
rs888559164 Chr.7: 103074941-103074941 on Build GRCh38
rs936326894 Chr.7: 103074760-103074760 on Build GRCh38
rs1027035861 Chr.7: 103074836-103074836 on Build GRCh38
rs909737091 Chr.7: 103075041-103075041 on Build GRCh38
rs756670578 Chr.7: 103074754-103074754 on Build GRCh38
rs944110683 Chr.7: 103075190-103075190 on Build GRCh38
rs892424541 Chr.7: 103074797-103074797 on Build GRCh38
rs1018777428 Chr.7: 103074871-103074871 on Build GRCh38
rs893189315 Chr.7: 103074787-103074787 on Build GRCh38
rs778752982 Chr.7: 103075144-103075144 on Build GRCh38
rs116721018 Chr.7: 103075024-103075024 on Build GRCh38
rs536264677 Chr.7: 103075098-103075098 on Build GRCh38
rs1000349097 Chr.7: 103075119-103075119 on Build GRCh38
rs983226205 Chr.7: 103074901-103074901 on Build GRCh38
rs1004202315 Chr.7: 103074842-103074842 on Build GRCh38
rs958941204 Chr.7: 103075155-103075155 on Build GRCh38
rs1015522964 Chr.7: 103074954-103074954 on Build GRCh38
rs971119602 Chr.7: 103075123-103075123 on Build GRCh38
rs990726673 Chr.7: 103075158-103075158 on Build GRCh38
rs964665468 Chr.7: 103074881-103074881 on Build GRCh38
rs1054477168 Chr.7: 103074785-103074785 on Build GRCh38
rs539772233 Chr.7: 103074947-103074947 on Build GRCh38
rs1008767445 Chr.7: 103074865-103074866 on Build GRCh38
rs1040907567 Chr.7: 103074826-103074826 on Build GRCh38
rs899682388 Chr.7: 103074833-103074833 on Build GRCh38
rs111229327 Chr.7: 103075171-103075171 on Build GRCh38
rs780762205 Chr.7: 103074861-103074861 on Build GRCh38
rs888444843 Chr.7: 103074840-103074840 on Build GRCh38
rs569117550 Chr.7: 103074940-103074940 on Build GRCh38
rs1045227054 Chr.7: 103074745-103074745 on Build GRCh38
rs1048454337 Chr.7: 103074937-103074937 on Build GRCh38
rs566488462 Chr.7: 103074978-103074978 on Build GRCh38
rs944144905 Chr.7: 103075023-103075023 on Build GRCh38
rs1021126720 Chr.7: 103075139-103075139 on Build GRCh38
rs1037083657 Chr.7: 103075067-103075067 on Build GRCh38
rs924905212 Chr.7: 103074757-103074757 on Build GRCh38
rs1001908292 Chr.7: 103074779-103074779 on Build GRCh38
rs551367320 Chr.7: 103074967-103074967 on Build GRCh38
rs946012262 Chr.7: 103075016-103075016 on Build GRCh38
rs562202783 Chr.7: 103075179-103075179 on Build GRCh38
rs1037115269 Chr.7: 103075191-103075191 on Build GRCh38
rs547082580 Chr.7: 103074845-103074845 on Build GRCh38
rs1041825561 Chr.7: 103074933-103074933 on Build GRCh38
rs1052351126 Chr.7: 103074778-103074778 on Build GRCh38
rs547035863 Chr.7: 103074811-103074811 on Build GRCh38
rs1014618894 Chr.7: 103075048-103075048 on Build GRCh38
rs751148216 Chr.7: 103074743-103074743 on Build GRCh38
rs976851752 Chr.7: 103075145-103075145 on Build GRCh38
rs1014649682 Chr.7: 103074982-103074982 on Build GRCh38
rs550543139 Chr.7: 103074904-103074904 on Build GRCh38
rs11557627 Chr.7: 103075014-103075014 on Build GRCh38
rs976659201 Chr.7: 103075019-103075019 on Build GRCh38
rs936315592 Chr.7: 103074913-103074913 on Build GRCh38
rs913804578 Chr.7: 103074917-103074917 on Build GRCh38
rs1044645375 Chr.7: 103075083-103075083 on Build GRCh38
rs978960144 Chr.7: 103075000-103075000 on Build GRCh38
rs981585910 Chr.7: 103075006-103075007 on Build GRCh38
rs891857602 Chr.7: 103075036-103075036 on Build GRCh38
rs925065522 Chr.7: 103074912-103074912 on Build GRCh38
rs1004151296 Chr.7: 103074948-103074948 on Build GRCh38
rs777601972 Chr.7: 103074932-103074932 on Build GRCh38
rs879215566 Chr.7: 103074839-103074839 on Build GRCh38
rs974785044 Chr.7: 103074885-103074885 on Build GRCh38
rs7797873 Chr.7: 103074903-103074903 on Build GRCh38
rs1023639546 Chr.7: 103074975-103074975 on Build GRCh38
rs891180377 Chr.7: 103075022-103075022 on Build GRCh38
rs913540610 Chr.7: 103075013-103075013 on Build GRCh38
rs561686465 Chr.7: 103074898-103074898 on Build GRCh38
rs565762426 Chr.7: 103075018-103075018 on Build GRCh38
rs932472953 Chr.7: 103075037-103075037 on Build GRCh38
rs957799214 Chr.7: 103075009-103075009 on Build GRCh38
rs908033053 Chr.7: 103074902-103074902 on Build GRCh38
rs950265773 Chr.7: 103074746-103074746 on Build GRCh38
rs529402253 Chr.7: 103075183-103075183 on Build GRCh38
rs940077169 Chr.7: 103075043-103075043 on Build GRCh38
rs546106574 Chr.7: 103075137-103075137 on Build GRCh38
rs1008649781 Chr.7: 103074943-103074943 on Build GRCh38
rs970250056 Chr.7: 103074976-103074976 on Build GRCh38
rs950069067 Chr.7: 103075193-103075193 on Build GRCh38
rs953940959 Chr.7: 103075161-103075161 on Build GRCh38
rs564163859 Chr.7: 103074732-103074732 on Build GRCh38
rs987836623 Chr.7: 103074916-103074916 on Build GRCh38
rs575507046 Chr.7: 103075125-103075125 on Build GRCh38
rs927509851 Chr.7: 103075007-103075007 on Build GRCh38
rs937628959 Chr.7: 103074923-103074923 on Build GRCh38

View Full Product Details