Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 123617469-123617985 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASB15
Entrez Gene ID
142685
Gene Name
ankyrin repeat and SOCS box containing 15
Gene Aliases
-
UniGene
Hs.83916
Gene Chromosome Location
Chr.7: 123567003-123637900 on Build GRCh38
Transcripts
NM_001290258.1 NM_080928.4 XM_005250146.4 XM_011515816.2 XM_011515817.2 XM_011515818.2 XM_011515819.2 XM_011515820.2 XM_011515821.2 XM_017011756.1 XM_017011757.1 XM_017011758.1 NR_111922.1

Gene Symbol
LOC102724555
Entrez Gene ID
102724555
Gene Name
uncharacterized LOC102724555
Gene Aliases
-
UniGene
Hs.83916
Gene Chromosome Location
Chr.7: 123614161-123624957 on Build GRCh38
Transcripts
NM_001290258.1 NM_080928.4 XM_005250146.4 XM_011515816.2 XM_011515817.2 XM_011515818.2 XM_011515819.2 XM_011515820.2 XM_011515821.2 XM_017011756.1 XM_017011757.1 XM_017011758.1 NR_111922.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6175853 c.394G>T p.E132* Chr.7: 123617680-123617680 on Build GRCh38
4401528 c.311G>A p.W104* Chr.7: 123617597-123617597 on Build GRCh38
5902832 c.347C>T p.T116I Chr.7: 123617633-123617633 on Build GRCh38
351503 c.447G>C p.L149L Chr.7: 123617733-123617733 on Build GRCh38
6108289 c.323_324CC>AA p.T108K Chr.7: 123617609-123617610 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1002159136 Chr.7: 123617525-123617525 on Build GRCh38
rs531275250 Chr.7: 123617588-123617588 on Build GRCh38
rs770479939 Chr.7: 123617707-123617707 on Build GRCh38
rs903774254 Chr.7: 123617522-123617522 on Build GRCh38
rs903722978 Chr.7: 123617816-123617816 on Build GRCh38
rs779859329 Chr.7: 123617747-123617748 on Build GRCh38
rs771720344 Chr.7: 123617714-123617714 on Build GRCh38
rs76164370 Chr.7: 123617781-123617781 on Build GRCh38
rs774084795 Chr.7: 123617593-123617594 on Build GRCh38
rs971860300 Chr.7: 123617614-123617614 on Build GRCh38
rs140968272 Chr.7: 123617727-123617727 on Build GRCh38
rs765229732 Chr.7: 123617755-123617755 on Build GRCh38
rs144825349 Chr.7: 123617624-123617624 on Build GRCh38
rs151090515 Chr.7: 123617616-123617616 on Build GRCh38
rs200997209 Chr.7: 123617742-123617742 on Build GRCh38
rs767502624 Chr.7: 123617543-123617543 on Build GRCh38
rs770385759 Chr.7: 123617583-123617583 on Build GRCh38
rs993748466 Chr.7: 123617902-123617902 on Build GRCh38
rs757454184 Chr.7: 123617665-123617665 on Build GRCh38
rs547756180 Chr.7: 123617753-123617753 on Build GRCh38
rs552306792 Chr.7: 123617498-123617498 on Build GRCh38
rs375860525 Chr.7: 123617680-123617680 on Build GRCh38
rs1024175411 Chr.7: 123617578-123617578 on Build GRCh38
rs755943388 Chr.7: 123617748-123617748 on Build GRCh38
rs148590103 Chr.7: 123617801-123617801 on Build GRCh38
rs984533020 Chr.7: 123617960-123617960 on Build GRCh38
rs1026467094 Chr.7: 123617615-123617615 on Build GRCh38
rs907061507 Chr.7: 123617866-123617866 on Build GRCh38
rs374275126 Chr.7: 123617949-123617949 on Build GRCh38
rs539651699 Chr.7: 123617767-123617767 on Build GRCh38
rs982951019 Chr.7: 123617663-123617663 on Build GRCh38
rs1012824848 Chr.7: 123617830-123617830 on Build GRCh38
rs761393352 Chr.7: 123617591-123617591 on Build GRCh38
rs377376910 Chr.7: 123617550-123617550 on Build GRCh38
rs370687850 Chr.7: 123617557-123617557 on Build GRCh38
rs540229052 Chr.7: 123617825-123617825 on Build GRCh38
rs943637203 Chr.7: 123617652-123617652 on Build GRCh38
rs1055078048 Chr.7: 123617839-123617839 on Build GRCh38
rs61742146 Chr.7: 123617699-123617699 on Build GRCh38
rs754985132 Chr.7: 123617515-123617515 on Build GRCh38
rs770583211 Chr.7: 123617734-123617734 on Build GRCh38
rs895141594 Chr.7: 123617827-123617827 on Build GRCh38
rs184731284 Chr.7: 123617653-123617653 on Build GRCh38
rs749743490 Chr.7: 123617711-123617711 on Build GRCh38
rs776335852 Chr.7: 123617738-123617738 on Build GRCh38
rs569352950 Chr.7: 123617787-123617787 on Build GRCh38
rs562253475 Chr.7: 123617507-123617507 on Build GRCh38
rs766106829 Chr.7: 123617637-123617637 on Build GRCh38
rs367869561 Chr.7: 123617640-123617640 on Build GRCh38
rs760314827 Chr.7: 123617635-123617635 on Build GRCh38
rs747531826 Chr.7: 123617932-123617932 on Build GRCh38
rs1012751524 Chr.7: 123617542-123617542 on Build GRCh38
rs371457160 Chr.7: 123617651-123617651 on Build GRCh38
rs537979784 Chr.7: 123617793-123617793 on Build GRCh38
rs574779619 Chr.7: 123617814-123617814 on Build GRCh38
rs554046872 Chr.7: 123617855-123617855 on Build GRCh38
rs570759152 Chr.7: 123617760-123617760 on Build GRCh38
rs867990135 Chr.7: 123617700-123617700 on Build GRCh38
rs374782660 Chr.7: 123617582-123617582 on Build GRCh38
rs373700675 Chr.7: 123617719-123617719 on Build GRCh38
rs759506891 Chr.7: 123617641-123617641 on Build GRCh38
rs746205048 Chr.7: 123617685-123617685 on Build GRCh38
rs746384568 Chr.7: 123617565-123617565 on Build GRCh38
rs867744447 Chr.7: 123617639-123617639 on Build GRCh38
rs750198433 Chr.7: 123617722-123617723 on Build GRCh38
rs765141636 Chr.7: 123617648-123617648 on Build GRCh38
rs748273303 Chr.7: 123617763-123617763 on Build GRCh38
rs764116233 Chr.7: 123617690-123617690 on Build GRCh38
rs1024624033 Chr.7: 123617840-123617840 on Build GRCh38
rs976740508 Chr.7: 123617657-123617657 on Build GRCh38
rs780702181 Chr.7: 123617709-123617709 on Build GRCh38
rs1055055609 Chr.7: 123617806-123617806 on Build GRCh38
rs1033617583 Chr.7: 123617504-123617504 on Build GRCh38

View Full Product Details