Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 491207-491732 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TDRP
Entrez Gene ID
157695
Gene Name
testis development related protein
Gene Aliases
C8orf42, Inm01, TDRP1, TDRP2
UniGene
Hs.289293
Gene Chromosome Location
Chr.8: 489790-545781 on Build GRCh38
Transcripts
NM_001256113.1 NM_175075.4 XM_006716232.3 XM_017013113.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7308676 c.566delA p.Y189fs*4 Chr.8: 491603-491603 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs552162583 Chr.8: 491619-491619 on Build GRCh38
rs911497956 Chr.8: 491609-491609 on Build GRCh38
rs990962131 Chr.8: 491442-491442 on Build GRCh38
rs925272256 Chr.8: 491282-491282 on Build GRCh38
rs970343419 Chr.8: 491452-491452 on Build GRCh38
rs11988112 Chr.8: 491234-491234 on Build GRCh38
rs1026200975 Chr.8: 491305-491305 on Build GRCh38
rs930059089 Chr.8: 491685-491685 on Build GRCh38
rs142219802 Chr.8: 491259-491259 on Build GRCh38
rs551856780 Chr.8: 491370-491370 on Build GRCh38
rs746940221 Chr.8: 491428-491428 on Build GRCh38
rs908495488 Chr.8: 491600-491600 on Build GRCh38
rs901267333 Chr.8: 491492-491492 on Build GRCh38
rs942822371 Chr.8: 491640-491640 on Build GRCh38
rs191489067 Chr.8: 491394-491394 on Build GRCh38
rs775651308 Chr.8: 491621-491621 on Build GRCh38
rs994146000 Chr.8: 491426-491426 on Build GRCh38
rs577137504 Chr.8: 491527-491527 on Build GRCh38
rs921214687 Chr.8: 491327-491327 on Build GRCh38
rs901139359 Chr.8: 491409-491409 on Build GRCh38
rs1053831238 Chr.8: 491248-491248 on Build GRCh38
rs573002997 Chr.8: 491256-491256 on Build GRCh38
rs529995061 Chr.8: 491322-491322 on Build GRCh38
rs1054695545 Chr.8: 491498-491498 on Build GRCh38
rs943366526 Chr.8: 491296-491296 on Build GRCh38
rs936789567 Chr.8: 491366-491366 on Build GRCh38
rs559432066 Chr.8: 491671-491671 on Build GRCh38
rs528191210 Chr.8: 491692-491692 on Build GRCh38
rs1030656609 Chr.8: 491594-491594 on Build GRCh38
rs990111727 Chr.8: 491604-491604 on Build GRCh38
rs543618561 Chr.8: 491582-491582 on Build GRCh38
rs984589485 Chr.8: 491323-491323 on Build GRCh38
rs1001053597 Chr.8: 491244-491244 on Build GRCh38
rs1013809955 Chr.8: 491424-491424 on Build GRCh38
rs1025111674 Chr.8: 491555-491555 on Build GRCh38
rs549835774 Chr.8: 491326-491326 on Build GRCh38
rs937734209 Chr.8: 491666-491666 on Build GRCh38
rs974342108 Chr.8: 491643-491643 on Build GRCh38
rs760172591 Chr.8: 491252-491253 on Build GRCh38
rs542001962 Chr.8: 491551-491551 on Build GRCh38
rs990624152 Chr.8: 491333-491333 on Build GRCh38
rs568607672 Chr.8: 491465-491465 on Build GRCh38
rs191073565 Chr.8: 491566-491566 on Build GRCh38
rs1041166073 Chr.8: 491405-491405 on Build GRCh38
rs748976639 Chr.8: 491524-491524 on Build GRCh38
rs79607057 Chr.8: 491458-491458 on Build GRCh38
rs186219064 Chr.8: 491548-491548 on Build GRCh38
rs1047040569 Chr.8: 491521-491521 on Build GRCh38
rs114789539 Chr.8: 491545-491545 on Build GRCh38
rs563700618 Chr.8: 491237-491237 on Build GRCh38
rs1012084548 Chr.8: 491549-491549 on Build GRCh38
rs761794846 Chr.8: 491669-491669 on Build GRCh38
rs771939611 Chr.8: 491300-491300 on Build GRCh38
rs976952279 Chr.8: 491647-491647 on Build GRCh38
rs868569925 Chr.8: 491324-491324 on Build GRCh38
rs796430037 Chr.8: 491332-491332 on Build GRCh38
rs181646868 Chr.8: 491462-491462 on Build GRCh38
rs942896897 Chr.8: 491480-491480 on Build GRCh38
rs960858560 Chr.8: 491240-491240 on Build GRCh38
rs758481608 Chr.8: 491238-491238 on Build GRCh38
rs970458022 Chr.8: 491574-491574 on Build GRCh38
rs1033885872 Chr.8: 491395-491395 on Build GRCh38
rs1006128800 Chr.8: 491434-491434 on Build GRCh38
rs1033897158 Chr.8: 491239-491239 on Build GRCh38
rs1020032933 Chr.8: 491275-491275 on Build GRCh38
rs1042640486 Chr.8: 491355-491355 on Build GRCh38
rs917554513 Chr.8: 491347-491347 on Build GRCh38
rs949059728 Chr.8: 491348-491348 on Build GRCh38
rs552222062 Chr.8: 491437-491437 on Build GRCh38
rs537731178 Chr.8: 491487-491487 on Build GRCh38
rs546625541 Chr.8: 491580-491580 on Build GRCh38
rs898122776 Chr.8: 491386-491386 on Build GRCh38
rs532450503 Chr.8: 491616-491616 on Build GRCh38
rs929942903 Chr.8: 491520-491520 on Build GRCh38
rs1001473041 Chr.8: 491543-491543 on Build GRCh38
rs902791516 Chr.8: 491689-491689 on Build GRCh38
rs1042683149 Chr.8: 491235-491235 on Build GRCh38
rs774079376 Chr.8: 491665-491665 on Build GRCh38
rs1047158097 Chr.8: 491699-491699 on Build GRCh38
rs896167889 Chr.8: 491518-491518 on Build GRCh38
rs1038121938 Chr.8: 491470-491472 on Build GRCh38
rs1024746165 Chr.8: 491446-491446 on Build GRCh38
rs937665716 Chr.8: 491494-491494 on Build GRCh38
rs557406697 Chr.8: 491502-491502 on Build GRCh38
rs957722938 Chr.8: 491598-491598 on Build GRCh38
rs924192912 Chr.8: 491359-491359 on Build GRCh38
rs117177894 Chr.8: 491430-491430 on Build GRCh38
rs1064416 Chr.8: 491457-491457 on Build GRCh38
rs917624044 Chr.8: 491233-491233 on Build GRCh38
rs763918349 Chr.8: 491644-491644 on Build GRCh38
rs766707641 Chr.8: 491312-491312 on Build GRCh38
rs563407496 Chr.8: 491589-491589 on Build GRCh38
rs908339406 Chr.8: 491464-491464 on Build GRCh38
rs74451014 Chr.8: 491540-491540 on Build GRCh38
rs560964278 Chr.8: 491262-491262 on Build GRCh38
rs772511967 Chr.8: 491534-491534 on Build GRCh38
rs933875414 Chr.8: 491460-491460 on Build GRCh38
rs911368264 Chr.8: 491468-491468 on Build GRCh38
rs763211191 Chr.8: 491635-491635 on Build GRCh38
rs997377257 Chr.8: 491298-491298 on Build GRCh38
rs34318716 Chr.8: 491429-491429 on Build GRCh38
rs950642247 Chr.8: 491432-491432 on Build GRCh38
rs759556093 Chr.8: 491607-491607 on Build GRCh38
rs965433403 Chr.8: 491278-491278 on Build GRCh38
rs1042636196 Chr.8: 491687-491687 on Build GRCh38
rs950529416 Chr.8: 491313-491313 on Build GRCh38
rs1007369646 Chr.8: 491398-491398 on Build GRCh38
rs898274436 Chr.8: 491483-491483 on Build GRCh38
rs959258053 Chr.8: 491439-491439 on Build GRCh38

View Full Product Details