Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 161171618-161172124 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B4GALT3
Entrez Gene ID
8703
Gene Name
beta-1,4-galactosyltransferase 3
Gene Aliases
beta4Gal-T3
UniGene
Hs.321231
Gene Chromosome Location
Chr.1: 161171310-161177968 on Build GRCh38
Transcripts
NM_001199873.1 NM_001199874.1 NM_003779.3 XM_005245566.1 XM_011510093.1 XM_017002714.1 XM_005245291.3 XM_005245295.3 XM_006711402.2 XM_006711403.2 XM_006711406.2 XM_011509663.2 XM_011509664.1 XM_011509665.2 XM_011509666.2 XM_011509667.2 XM_011509668.2 XM_011509669.1 XM_011509670.2 XM_011509673.2 XM_011509674.2 XM_011509676.1 XM_011509677.1 XM_011509678.1 XM_011509679.1 XM_011509681.1 XM_011509682.1 XM_017001561.1 XM_017001562.1 XM_017001563.1 XM_017001564.1 XM_017001570.1 XM_017001571.1

Gene Symbol
PPOX
Entrez Gene ID
5498
Gene Name
protoporphyrinogen oxidase
Gene Aliases
PPO, V290M, VP
UniGene
Hs.517373
Gene Chromosome Location
Chr.1: 161165728-161178277 on Build GRCh38
Transcripts
NM_001199873.1 NM_001199874.1 NM_003779.3 XM_005245566.1 XM_011510093.1 XM_017002714.1 XM_005245291.3 XM_005245295.3 XM_006711402.2 XM_006711403.2 XM_006711406.2 XM_011509663.2 XM_011509664.1 XM_011509665.2 XM_011509666.2 XM_011509667.2 XM_011509668.2 XM_011509669.1 XM_011509670.2 XM_011509673.2 XM_011509674.2 XM_011509676.1 XM_011509677.1 XM_011509678.1 XM_011509679.1 XM_011509681.1 XM_011509682.1 XM_017001561.1 XM_017001562.1 XM_017001563.1 XM_017001564.1 XM_017001570.1 XM_017001571.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6059051 c.1097G>A p.R366H Chr.1: 161171901-161171901 on Build GRCh38
898498 c.1069T>C p.Y357H Chr.1: 161171929-161171929 on Build GRCh38
898497 c.1115G>A p.R372H Chr.1: 161171883-161171883 on Build GRCh38
4439765 c.1124delC p.P375fs*>19 Chr.1: 161171874-161171874 on Build GRCh38
898496 c.1182A>G p.*394W Chr.1: 161171816-161171816 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780404324 Chr.1: 161172072-161172072 on Build GRCh38
rs747011978 Chr.1: 161171942-161171942 on Build GRCh38
rs368972108 Chr.1: 161171810-161171810 on Build GRCh38
rs917341171 Chr.1: 161171703-161171703 on Build GRCh38
rs887281727 Chr.1: 161171761-161171761 on Build GRCh38
rs762986792 Chr.1: 161171962-161171962 on Build GRCh38
rs747156753 Chr.1: 161172076-161172076 on Build GRCh38
rs764451166 Chr.1: 161171913-161171913 on Build GRCh38
rs1047712763 Chr.1: 161171747-161171747 on Build GRCh38
rs371077103 Chr.1: 161171830-161171830 on Build GRCh38
rs145419100 Chr.1: 161171782-161171782 on Build GRCh38
rs367875334 Chr.1: 161171919-161171919 on Build GRCh38
rs950130600 Chr.1: 161171704-161171704 on Build GRCh38
rs757586249 Chr.1: 161171927-161171927 on Build GRCh38
rs775022821 Chr.1: 161172030-161172030 on Build GRCh38
rs774816977 Chr.1: 161171865-161171865 on Build GRCh38
rs767239787 Chr.1: 161172033-161172033 on Build GRCh38
rs894169144 Chr.1: 161171794-161171794 on Build GRCh38
rs778200053 Chr.1: 161171851-161171851 on Build GRCh38
rs750006771 Chr.1: 161171832-161171832 on Build GRCh38
rs760228954 Chr.1: 161171808-161171810 on Build GRCh38
rs146478151 Chr.1: 161171922-161171922 on Build GRCh38
rs762438234 Chr.1: 161172025-161172025 on Build GRCh38
rs766374908 Chr.1: 161171967-161171967 on Build GRCh38
rs763965479 Chr.1: 161171815-161171815 on Build GRCh38
rs563551027 Chr.1: 161171857-161171857 on Build GRCh38
rs779921161 Chr.1: 161171834-161171834 on Build GRCh38
rs111614721 Chr.1: 161171901-161171901 on Build GRCh38
rs765233285 Chr.1: 161172099-161172115 on Build GRCh38
rs144909171 Chr.1: 161171836-161171836 on Build GRCh38
rs41270033 Chr.1: 161171883-161171883 on Build GRCh38
rs6779 Chr.1: 161171642-161171642 on Build GRCh38
rs754526253 Chr.1: 161171841-161171841 on Build GRCh38
rs775657671 Chr.1: 161171806-161171806 on Build GRCh38
rs143618194 Chr.1: 161171847-161171852 on Build GRCh38
rs371235014 Chr.1: 161171890-161171890 on Build GRCh38
rs190810180 Chr.1: 161172053-161172053 on Build GRCh38
rs780215020 Chr.1: 161171934-161171934 on Build GRCh38
rs150162843 Chr.1: 161171996-161171996 on Build GRCh38
rs954647388 Chr.1: 161171852-161171852 on Build GRCh38
rs745986761 Chr.1: 161171867-161171867 on Build GRCh38
rs781536903 Chr.1: 161172100-161172100 on Build GRCh38
rs746359457 Chr.1: 161171931-161171931 on Build GRCh38
rs751099833 Chr.1: 161172013-161172013 on Build GRCh38
rs776689921 Chr.1: 161171950-161171950 on Build GRCh38
rs775814722 Chr.1: 161171877-161171877 on Build GRCh38
rs776264769 Chr.1: 161171946-161171947 on Build GRCh38
rs757954359 Chr.1: 161171833-161171833 on Build GRCh38
rs767892217 Chr.1: 161171705-161171705 on Build GRCh38
rs1047298806 Chr.1: 161171726-161171726 on Build GRCh38
rs779461856 Chr.1: 161171929-161171929 on Build GRCh38
rs771490999 Chr.1: 161171862-161171862 on Build GRCh38
rs773080802 Chr.1: 161171959-161171959 on Build GRCh38
rs1057311925 Chr.1: 161172010-161172010 on Build GRCh38
rs761907963 Chr.1: 161172091-161172092 on Build GRCh38
rs995818944 Chr.1: 161171818-161171818 on Build GRCh38
rs771570356 Chr.1: 161171800-161171801 on Build GRCh38
rs764183520 Chr.1: 161171881-161171881 on Build GRCh38
rs367859921 Chr.1: 161171903-161171903 on Build GRCh38
rs941553438 Chr.1: 161172038-161172038 on Build GRCh38
rs374151902 Chr.1: 161171776-161171776 on Build GRCh38
rs750965486 Chr.1: 161171915-161171915 on Build GRCh38
rs987716260 Chr.1: 161171656-161171656 on Build GRCh38
rs187196663 Chr.1: 161171956-161171956 on Build GRCh38
rs768221056 Chr.1: 161171855-161171856 on Build GRCh38
rs762612920 Chr.1: 161171895-161171895 on Build GRCh38
rs987495575 Chr.1: 161171673-161171673 on Build GRCh38
rs765309135 Chr.1: 161171885-161171885 on Build GRCh38
rs768971975 Chr.1: 161171949-161171949 on Build GRCh38
rs556696668 Chr.1: 161171695-161171695 on Build GRCh38
rs752286852 Chr.1: 161172034-161172034 on Build GRCh38
rs367641577 Chr.1: 161171880-161171880 on Build GRCh38
rs1046907837 Chr.1: 161171944-161171944 on Build GRCh38
rs767774729 Chr.1: 161171783-161171783 on Build GRCh38
rs769702808 Chr.1: 161171958-161171958 on Build GRCh38
rs372601868 Chr.1: 161172054-161172054 on Build GRCh38
rs375516892 Chr.1: 161172099-161172099 on Build GRCh38
rs910514435 Chr.1: 161171694-161171694 on Build GRCh38
rs150052876 Chr.1: 161171702-161171702 on Build GRCh38
rs898410918 Chr.1: 161171697-161171697 on Build GRCh38
rs114361795 Chr.1: 161171664-161171664 on Build GRCh38
rs771271911 Chr.1: 161171773-161171773 on Build GRCh38
rs866970689 Chr.1: 161171711-161171711 on Build GRCh38
rs753511474 Chr.1: 161171829-161171829 on Build GRCh38
rs937337169 Chr.1: 161171700-161171700 on Build GRCh38
rs939987098 Chr.1: 161171846-161171847 on Build GRCh38
rs374982935 Chr.1: 161171907-161171907 on Build GRCh38
rs749571924 Chr.1: 161171861-161171861 on Build GRCh38
rs377086271 Chr.1: 161171882-161171882 on Build GRCh38
rs775163116 Chr.1: 161171766-161171767 on Build GRCh38
rs775492179 Chr.1: 161171879-161171879 on Build GRCh38
rs950219674 Chr.1: 161171979-161171979 on Build GRCh38
rs978681066 Chr.1: 161171680-161171680 on Build GRCh38
rs967738055 Chr.1: 161171676-161171676 on Build GRCh38

View Full Product Details