Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 161626067-161626570 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FCGR3B
Entrez Gene ID
2215
Gene Name
Fc fragment of IgG receptor IIIb
Gene Aliases
CD16, CD16b, FCG3, FCGR3, FCR-10, FCRIII, FCRIIIb
UniGene
Hs.372679
Gene Chromosome Location
Chr.1: 161623196-161631963 on Build GRCh38
Transcripts
NM_000570.4 NM_001244753.1 NM_001271035.1 NM_001271036.1 NM_001271037.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2087302 c.393G>T p.W131C Chr.1: 161626329-161626329 on Build GRCh38
6499382 c.457C>T p.H153Y Chr.1: 161626265-161626265 on Build GRCh38
4439578 c.333C>T p.L111L Chr.1: 161626389-161626389 on Build GRCh38
2155542 c.380G>A p.R127K Chr.1: 161626342-161626342 on Build GRCh38
463278 c.519G>A p.R173R Chr.1: 161626203-161626203 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780470413 Chr.1: 161626374-161626374 on Build GRCh38
rs188904436 Chr.1: 161626220-161626220 on Build GRCh38
rs569831380 Chr.1: 161626445-161626445 on Build GRCh38
rs748393493 Chr.1: 161626443-161626443 on Build GRCh38
rs753862608 Chr.1: 161626145-161626145 on Build GRCh38
rs780479891 Chr.1: 161626428-161626428 on Build GRCh38
rs71632959 Chr.1: 161626282-161626282 on Build GRCh38
rs774669456 Chr.1: 161626119-161626119 on Build GRCh38
rs531328236 Chr.1: 161626290-161626290 on Build GRCh38
rs199850917 Chr.1: 161626431-161626431 on Build GRCh38
rs761937129 Chr.1: 161626306-161626306 on Build GRCh38
rs766024902 Chr.1: 161626404-161626404 on Build GRCh38
rs376613160 Chr.1: 161626219-161626219 on Build GRCh38
rs539705232 Chr.1: 161626424-161626424 on Build GRCh38
rs935979857 Chr.1: 161626164-161626164 on Build GRCh38
rs367924998 Chr.1: 161626097-161626097 on Build GRCh38
rs752445833 Chr.1: 161626417-161626417 on Build GRCh38
rs371517248 Chr.1: 161626214-161626214 on Build GRCh38
rs751319014 Chr.1: 161626333-161626333 on Build GRCh38
rs754401520 Chr.1: 161626260-161626260 on Build GRCh38
rs371730223 Chr.1: 161626098-161626098 on Build GRCh38
rs539692915 Chr.1: 161626106-161626106 on Build GRCh38
rs372653045 Chr.1: 161626522-161626522 on Build GRCh38
rs749692570 Chr.1: 161626279-161626279 on Build GRCh38
rs373862616 Chr.1: 161626265-161626265 on Build GRCh38
rs752240620 Chr.1: 161626341-161626341 on Build GRCh38
rs776781080 Chr.1: 161626303-161626303 on Build GRCh38
rs747138881 Chr.1: 161626376-161626376 on Build GRCh38
rs796343414 Chr.1: 161626237-161626237 on Build GRCh38
rs767638126 Chr.1: 161626124-161626124 on Build GRCh38
rs771581225 Chr.1: 161626281-161626281 on Build GRCh38
rs767195235 Chr.1: 161626413-161626413 on Build GRCh38
rs745540495 Chr.1: 161626105-161626105 on Build GRCh38
rs71632958 Chr.1: 161626250-161626250 on Build GRCh38
rs1010720514 Chr.1: 161626203-161626203 on Build GRCh38
rs547946059 Chr.1: 161626310-161626310 on Build GRCh38
rs757115083 Chr.1: 161626096-161626096 on Build GRCh38
rs748627593 Chr.1: 161626110-161626110 on Build GRCh38
rs780789100 Chr.1: 161626278-161626278 on Build GRCh38
rs4047527 Chr.1: 161626417-161626418 on Build GRCh38
rs373599391 Chr.1: 161626090-161626090 on Build GRCh38
rs754664371 Chr.1: 161626324-161626324 on Build GRCh38
rs755754121 Chr.1: 161626351-161626351 on Build GRCh38
rs774982858 Chr.1: 161626109-161626109 on Build GRCh38
rs768548987 Chr.1: 161626302-161626302 on Build GRCh38
rs368509879 Chr.1: 161626131-161626131 on Build GRCh38
rs767346542 Chr.1: 161626202-161626202 on Build GRCh38
rs570696778 Chr.1: 161626383-161626383 on Build GRCh38
rs763728267 Chr.1: 161626422-161626422 on Build GRCh38
rs747393520 Chr.1: 161626430-161626430 on Build GRCh38
rs766880686 Chr.1: 161626326-161626326 on Build GRCh38
rs779540050 Chr.1: 161626358-161626358 on Build GRCh38
rs754764388 Chr.1: 161626370-161626375 on Build GRCh38
rs769914012 Chr.1: 161626391-161626391 on Build GRCh38
rs866086184 Chr.1: 161626370-161626370 on Build GRCh38
rs755635433 Chr.1: 161626418-161626418 on Build GRCh38
rs750006226 Chr.1: 161626167-161626167 on Build GRCh38
rs375009248 Chr.1: 161626111-161626111 on Build GRCh38
rs751135498 Chr.1: 161626318-161626318 on Build GRCh38
rs200914601 Chr.1: 161626238-161626238 on Build GRCh38
rs969588337 Chr.1: 161626414-161626414 on Build GRCh38
rs759308120 Chr.1: 161626412-161626412 on Build GRCh38
rs536913894 Chr.1: 161626457-161626457 on Build GRCh38
rs418215 Chr.1: 161626493-161626493 on Build GRCh38
rs779652750 Chr.1: 161626246-161626246 on Build GRCh38
rs764950507 Chr.1: 161626233-161626233 on Build GRCh38
rs371070567 Chr.1: 161626316-161626316 on Build GRCh38
rs372772013 Chr.1: 161626193-161626193 on Build GRCh38
rs372411058 Chr.1: 161626395-161626395 on Build GRCh38
rs372871192 Chr.1: 161626221-161626221 on Build GRCh38
rs750202857 Chr.1: 161626235-161626235 on Build GRCh38
rs776782096 Chr.1: 161626379-161626379 on Build GRCh38
rs367634824 Chr.1: 161626200-161626200 on Build GRCh38
rs568134732 Chr.1: 161626312-161626312 on Build GRCh38
rs563492801 Chr.1: 161626283-161626283 on Build GRCh38
rs200215055 Chr.1: 161626196-161626196 on Build GRCh38
rs763200211 Chr.1: 161626116-161626116 on Build GRCh38
rs571234639 Chr.1: 161626439-161626439 on Build GRCh38
rs767873532 Chr.1: 161626431-161626432 on Build GRCh38
rs762914235 Chr.1: 161626401-161626401 on Build GRCh38
rs114169903 Chr.1: 161626242-161626242 on Build GRCh38
rs980675129 Chr.1: 161626473-161626473 on Build GRCh38
rs763905220 Chr.1: 161626138-161626138 on Build GRCh38
rs756952234 Chr.1: 161626150-161626150 on Build GRCh38
rs71632957 Chr.1: 161626224-161626224 on Build GRCh38
rs1029222881 Chr.1: 161626240-161626240 on Build GRCh38
rs758058086 Chr.1: 161626177-161626177 on Build GRCh38
rs376472862 Chr.1: 161626227-161626227 on Build GRCh38
rs779736569 Chr.1: 161626182-161626182 on Build GRCh38
rs376493908 Chr.1: 161626291-161626291 on Build GRCh38
rs990133852 Chr.1: 161626548-161626548 on Build GRCh38
rs746397505 Chr.1: 161626365-161626365 on Build GRCh38
rs773840391 Chr.1: 161626114-161626114 on Build GRCh38
rs772612155 Chr.1: 161626288-161626288 on Build GRCh38
rs758796084 Chr.1: 161626371-161626371 on Build GRCh38
rs766818878 Chr.1: 161626118-161626118 on Build GRCh38
rs748192789 Chr.1: 161626386-161626386 on Build GRCh38
rs527319298 Chr.1: 161626315-161626315 on Build GRCh38
rs760987440 Chr.1: 161626295-161626295 on Build GRCh38
rs535629418 Chr.1: 161626474-161626474 on Build GRCh38
rs200319084 Chr.1: 161626378-161626378 on Build GRCh38
rs760874126 Chr.1: 161626135-161626135 on Build GRCh38
rs576621422 Chr.1: 161626161-161626161 on Build GRCh38
rs746500816 Chr.1: 161626188-161626188 on Build GRCh38
rs774209738 Chr.1: 161626411-161626411 on Build GRCh38

View Full Product Details