Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 103375618-103376088 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTHRC1
Entrez Gene ID
115908
Gene Name
collagen triple helix repeat containing 1
Gene Aliases
-
UniGene
Hs.405614
Gene Chromosome Location
Chr.8: 103371515-103383005 on Build GRCh38
Transcripts
NM_001256099.1 NM_138455.3 XM_011516824.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5142921 c.206G>A p.R69Q Chr.8: 103375835-103375835 on Build GRCh38
1094471 c.355G>T p.D119Y Chr.8: 103375942-103375942 on Build GRCh38
1158113 c.167T>C p.L56S Chr.8: 103375754-103375754 on Build GRCh38
4958220 c.309C>A p.G103G Chr.8: 103375938-103375938 on Build GRCh38
4936364 c.274T>A p.Y92N Chr.8: 103375903-103375903 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs145881116 Chr.8: 103375834-103375834 on Build GRCh38
rs759365776 Chr.8: 103375792-103375792 on Build GRCh38
rs983881049 Chr.8: 103375897-103375897 on Build GRCh38
rs940782594 Chr.8: 103375681-103375681 on Build GRCh38
rs771956187 Chr.8: 103375908-103375908 on Build GRCh38
rs187299788 Chr.8: 103375785-103375785 on Build GRCh38
rs750617028 Chr.8: 103375890-103375891 on Build GRCh38
rs905171722 Chr.8: 103375704-103375704 on Build GRCh38
rs756369288 Chr.8: 103375875-103375875 on Build GRCh38
rs868291260 Chr.8: 103375904-103375904 on Build GRCh38
rs781710038 Chr.8: 103375846-103375846 on Build GRCh38
rs113614259 Chr.8: 103375782-103375782 on Build GRCh38
rs772011261 Chr.8: 103376024-103376024 on Build GRCh38
rs754082574 Chr.8: 103376022-103376033 on Build GRCh38
rs540047490 Chr.8: 103376038-103376039 on Build GRCh38
rs761583037 Chr.8: 103376054-103376054 on Build GRCh38
rs748382570 Chr.8: 103375780-103375780 on Build GRCh38
rs146609679 Chr.8: 103375959-103375959 on Build GRCh38
rs779856790 Chr.8: 103376057-103376057 on Build GRCh38
rs780649365 Chr.8: 103375725-103375725 on Build GRCh38
rs149006532 Chr.8: 103375864-103375864 on Build GRCh38
rs374084026 Chr.8: 103375958-103375958 on Build GRCh38
rs776397159 Chr.8: 103376050-103376050 on Build GRCh38
rs767061518 Chr.8: 103375835-103375835 on Build GRCh38
rs745828427 Chr.8: 103375902-103375902 on Build GRCh38
rs988716539 Chr.8: 103376028-103376028 on Build GRCh38
rs546224866 Chr.8: 103376039-103376039 on Build GRCh38
rs775361593 Chr.8: 103375913-103375913 on Build GRCh38
rs149662990 Chr.8: 103375807-103375807 on Build GRCh38
rs775098206 Chr.8: 103375797-103375797 on Build GRCh38
rs747437502 Chr.8: 103375767-103375767 on Build GRCh38
rs373203731 Chr.8: 103375811-103375811 on Build GRCh38
rs531041475 Chr.8: 103375724-103375724 on Build GRCh38
rs946948667 Chr.8: 103375694-103375694 on Build GRCh38
rs191688677 Chr.8: 103376034-103376034 on Build GRCh38
rs745879862 Chr.8: 103376006-103376006 on Build GRCh38
rs539717711 Chr.8: 103376021-103376021 on Build GRCh38
rs908393007 Chr.8: 103375899-103375899 on Build GRCh38
rs767275808 Chr.8: 103375796-103375796 on Build GRCh38
rs770966489 Chr.8: 103375881-103375881 on Build GRCh38
rs753401752 Chr.8: 103375812-103375812 on Build GRCh38
rs757420039 Chr.8: 103375993-103375993 on Build GRCh38
rs909298683 Chr.8: 103375651-103375651 on Build GRCh38
rs779752692 Chr.8: 103375956-103375956 on Build GRCh38
rs755392681 Chr.8: 103375770-103375770 on Build GRCh38
rs749448992 Chr.8: 103375880-103375880 on Build GRCh38
rs377662370 Chr.8: 103375972-103375972 on Build GRCh38
rs768415647 Chr.8: 103375922-103375922 on Build GRCh38
rs755445506 Chr.8: 103375843-103375843 on Build GRCh38
rs1050683688 Chr.8: 103375756-103375756 on Build GRCh38
rs199777983 Chr.8: 103376048-103376048 on Build GRCh38
rs750236918 Chr.8: 103375645-103375645 on Build GRCh38
rs777473565 Chr.8: 103376061-103376064 on Build GRCh38
rs915387656 Chr.8: 103376038-103376038 on Build GRCh38
rs752084339 Chr.8: 103375842-103375842 on Build GRCh38
rs778093567 Chr.8: 103375988-103375988 on Build GRCh38
rs761452801 Chr.8: 103375954-103375954 on Build GRCh38
rs781609744 Chr.8: 103375773-103375773 on Build GRCh38
rs889397221 Chr.8: 103375762-103375762 on Build GRCh38
rs756380687 Chr.8: 103375960-103375961 on Build GRCh38
rs376472345 Chr.8: 103375866-103375866 on Build GRCh38
rs1033593807 Chr.8: 103375860-103375860 on Build GRCh38
rs901866266 Chr.8: 103375718-103375718 on Build GRCh38
rs972142813 Chr.8: 103375936-103375936 on Build GRCh38
rs369008583 Chr.8: 103375786-103375786 on Build GRCh38
rs549472586 Chr.8: 103375990-103375990 on Build GRCh38
rs145838701 Chr.8: 103375878-103375878 on Build GRCh38
rs1036287767 Chr.8: 103375717-103375717 on Build GRCh38
rs72124849 Chr.8: 103376061-103376065 on Build GRCh38
rs753158525 Chr.8: 103375960-103375960 on Build GRCh38
rs780939033 Chr.8: 103375647-103375647 on Build GRCh38
rs778995847 Chr.8: 103375994-103375994 on Build GRCh38
rs778869312 Chr.8: 103375888-103375888 on Build GRCh38
rs147175675 Chr.8: 103375781-103375781 on Build GRCh38
rs1001036925 Chr.8: 103375775-103375775 on Build GRCh38
rs6468870 Chr.8: 103376048-103376048 on Build GRCh38
rs753249581 Chr.8: 103375709-103375709 on Build GRCh38
rs753984040 Chr.8: 103375991-103375991 on Build GRCh38
rs201468963 Chr.8: 103375746-103375746 on Build GRCh38
rs761868024 Chr.8: 103375704-103375705 on Build GRCh38
rs760330177 Chr.8: 103375917-103375917 on Build GRCh38
rs776145170 Chr.8: 103375946-103375946 on Build GRCh38
rs767655755 Chr.8: 103375709-103375719 on Build GRCh38
rs764820491 Chr.8: 103375957-103375957 on Build GRCh38
rs763680532 Chr.8: 103375696-103375696 on Build GRCh38
rs779842417 Chr.8: 103376027-103376027 on Build GRCh38

View Full Product Details