Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 138593928-138594435 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL22A1
Entrez Gene ID
169044
Gene Name
collagen type XXII alpha 1
Gene Aliases
-
UniGene
Hs.117169
Gene Chromosome Location
Chr.8: 138588235-138914087 on Build GRCh38
Transcripts
NM_152888.2 XM_011516883.2 XM_011516884.2 XM_011516885.2 XM_011516886.2 XM_011516887.2 XM_011516889.2 XM_017013150.1 XM_017013151.1 XM_017013152.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7400618 c.4615+7G>C p.? Chr.8: 138594010-138594010 on Build GRCh38
5721263 c.4578G>A p.G1526G Chr.8: 138594054-138594054 on Build GRCh38
3897095 c.4543G>A p.G1515S Chr.8: 138594089-138594089 on Build GRCh38
4406144 c.4505C>A p.P1502Q Chr.8: 138594127-138594127 on Build GRCh38
1623649 c.4558C>A p.P1520T Chr.8: 138594074-138594074 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs987286819 Chr.8: 138594040-138594040 on Build GRCh38
rs547467071 Chr.8: 138594405-138594405 on Build GRCh38
rs771428130 Chr.8: 138594099-138594099 on Build GRCh38
rs757031031 Chr.8: 138594166-138594166 on Build GRCh38
rs765161418 Chr.8: 138594201-138594201 on Build GRCh38
rs545551660 Chr.8: 138594331-138594331 on Build GRCh38
rs775611424 Chr.8: 138594122-138594122 on Build GRCh38
rs574775105 Chr.8: 138594269-138594269 on Build GRCh38
rs554915061 Chr.8: 138594252-138594252 on Build GRCh38
rs1029387217 Chr.8: 138594292-138594292 on Build GRCh38
rs931304758 Chr.8: 138594089-138594089 on Build GRCh38
rs895660454 Chr.8: 138593980-138593980 on Build GRCh38
rs149118857 Chr.8: 138594119-138594119 on Build GRCh38
rs1042708033 Chr.8: 138593958-138593958 on Build GRCh38
rs576957094 Chr.8: 138594311-138594311 on Build GRCh38
rs750755891 Chr.8: 138594236-138594236 on Build GRCh38
rs773869516 Chr.8: 138594239-138594239 on Build GRCh38
rs1032473216 Chr.8: 138594132-138594132 on Build GRCh38
rs775571353 Chr.8: 138594202-138594202 on Build GRCh38
rs200663003 Chr.8: 138594220-138594220 on Build GRCh38
rs376814137 Chr.8: 138594090-138594090 on Build GRCh38
rs761127995 Chr.8: 138594240-138594240 on Build GRCh38
rs759469460 Chr.8: 138594074-138594074 on Build GRCh38
rs781554664 Chr.8: 138593952-138593952 on Build GRCh38
rs751164353 Chr.8: 138594210-138594210 on Build GRCh38
rs893543952 Chr.8: 138594380-138594380 on Build GRCh38
rs756451546 Chr.8: 138594247-138594247 on Build GRCh38
rs753349571 Chr.8: 138594013-138594013 on Build GRCh38
rs375352490 Chr.8: 138593987-138593987 on Build GRCh38
rs758615300 Chr.8: 138593991-138593991 on Build GRCh38
rs1047038618 Chr.8: 138594261-138594261 on Build GRCh38
rs574848231 Chr.8: 138593969-138593969 on Build GRCh38
rs748343987 Chr.8: 138594191-138594191 on Build GRCh38
rs886965860 Chr.8: 138594411-138594411 on Build GRCh38
rs747408977 Chr.8: 138594071-138594071 on Build GRCh38
rs368931383 Chr.8: 138594153-138594153 on Build GRCh38
rs759483374 Chr.8: 138594157-138594157 on Build GRCh38
rs770253828 Chr.8: 138594121-138594121 on Build GRCh38
rs905369385 Chr.8: 138594360-138594362 on Build GRCh38
rs111264173 Chr.8: 138593974-138593974 on Build GRCh38
rs10097507 Chr.8: 138594363-138594363 on Build GRCh38
rs372973189 Chr.8: 138594014-138594014 on Build GRCh38
rs933372446 Chr.8: 138594127-138594127 on Build GRCh38
rs752188788 Chr.8: 138594092-138594092 on Build GRCh38
rs115499018 Chr.8: 138594192-138594192 on Build GRCh38
rs561595229 Chr.8: 138594095-138594095 on Build GRCh38
rs1009268762 Chr.8: 138594380-138594380 on Build GRCh38
rs749715113 Chr.8: 138594392-138594392 on Build GRCh38
rs933180755 Chr.8: 138594023-138594023 on Build GRCh38
rs756608501 Chr.8: 138594173-138594173 on Build GRCh38
rs72727814 Chr.8: 138594184-138594184 on Build GRCh38
rs768819909 Chr.8: 138594123-138594123 on Build GRCh38
rs765117772 Chr.8: 138594160-138594160 on Build GRCh38
rs769315745 Chr.8: 138594131-138594131 on Build GRCh38
rs771513929 Chr.8: 138594072-138594072 on Build GRCh38
rs369056975 Chr.8: 138594004-138594004 on Build GRCh38
rs763685563 Chr.8: 138594208-138594208 on Build GRCh38
rs759682018 Chr.8: 138593982-138593982 on Build GRCh38
rs763212911 Chr.8: 138594078-138594078 on Build GRCh38
rs1013736262 Chr.8: 138594106-138594106 on Build GRCh38
rs1032133568 Chr.8: 138594375-138594377 on Build GRCh38
rs781478424 Chr.8: 138594114-138594114 on Build GRCh38
rs762086962 Chr.8: 138594133-138594133 on Build GRCh38
rs780000347 Chr.8: 138594025-138594025 on Build GRCh38
rs540385225 Chr.8: 138593970-138593970 on Build GRCh38
rs369311158 Chr.8: 138594187-138594187 on Build GRCh38
rs146594952 Chr.8: 138594381-138594381 on Build GRCh38
rs753241668 Chr.8: 138594033-138594033 on Build GRCh38
rs748451665 Chr.8: 138594162-138594162 on Build GRCh38
rs780546094 Chr.8: 138594251-138594251 on Build GRCh38
rs776810948 Chr.8: 138593979-138593979 on Build GRCh38
rs767109882 Chr.8: 138594221-138594221 on Build GRCh38
rs778350816 Chr.8: 138594053-138594053 on Build GRCh38
rs10097485 Chr.8: 138594296-138594296 on Build GRCh38
rs951294502 Chr.8: 138594052-138594052 on Build GRCh38
rs200938042 Chr.8: 138593986-138593986 on Build GRCh38
rs201934935 Chr.8: 138594165-138594165 on Build GRCh38
rs145074537 Chr.8: 138594077-138594077 on Build GRCh38
rs767287920 Chr.8: 138593998-138593998 on Build GRCh38
rs987928364 Chr.8: 138594310-138594310 on Build GRCh38
rs780451617 Chr.8: 138594175-138594175 on Build GRCh38
rs138310793 Chr.8: 138594163-138594163 on Build GRCh38
rs763052328 Chr.8: 138594207-138594207 on Build GRCh38
rs768730192 Chr.8: 138594186-138594186 on Build GRCh38
rs531509761 Chr.8: 138594081-138594081 on Build GRCh38
rs946311449 Chr.8: 138594161-138594161 on Build GRCh38
rs866028615 Chr.8: 138594117-138594117 on Build GRCh38
rs200310171 Chr.8: 138594085-138594085 on Build GRCh38
rs759271482 Chr.8: 138594199-138594199 on Build GRCh38
rs747819697 Chr.8: 138594067-138594067 on Build GRCh38
rs202020964 Chr.8: 138593992-138593992 on Build GRCh38
rs201591565 Chr.8: 138594037-138594037 on Build GRCh38
rs774064214 Chr.8: 138593966-138593966 on Build GRCh38
rs547734171 Chr.8: 138594083-138594083 on Build GRCh38
rs949837363 Chr.8: 138594403-138594403 on Build GRCh38
rs186880766 Chr.8: 138594391-138594391 on Build GRCh38
rs949656791 Chr.8: 138594329-138594329 on Build GRCh38
rs760488061 Chr.8: 138594082-138594082 on Build GRCh38
rs866909847 Chr.8: 138594183-138594183 on Build GRCh38
rs887080965 Chr.8: 138594284-138594284 on Build GRCh38
rs10110076 Chr.8: 138594286-138594286 on Build GRCh38
rs776446118 Chr.8: 138594194-138594194 on Build GRCh38
rs185123006 Chr.8: 138594068-138594068 on Build GRCh38
rs139744602 Chr.8: 138594353-138594353 on Build GRCh38
rs764343646 Chr.8: 138594080-138594080 on Build GRCh38
rs777206646 Chr.8: 138594073-138594073 on Build GRCh38
rs201551033 Chr.8: 138594170-138594170 on Build GRCh38
rs375061236 Chr.8: 138593968-138593968 on Build GRCh38
rs766146519 Chr.8: 138594146-138594146 on Build GRCh38
rs200298766 Chr.8: 138594094-138594094 on Build GRCh38
rs144526158 Chr.8: 138594359-138594359 on Build GRCh38
rs781598213 Chr.8: 138594169-138594169 on Build GRCh38
rs988065247 Chr.8: 138594013-138594013 on Build GRCh38

View Full Product Details