Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 143589620-143590123 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EEF1D
Entrez Gene ID
1936
Gene Name
eukaryotic translation elongation factor 1 delta
Gene Aliases
EF-1D, EF1D, FP1047
UniGene
Hs.333388
Gene Chromosome Location
Chr.8: 143579697-143597675 on Build GRCh38
Transcripts
NM_001130053.2 NM_001130055.2 NM_001130056.2 NM_001130057.2 NM_001195203.1 NM_001289950.1 NM_001317743.1 NM_001960.4 NM_032378.4 XM_005250820.3 XM_005250823.1 XM_006716519.3 XM_006716520.3 XM_006716522.1 XM_006716523.1 XM_006716524.1 XM_006716525.1 XM_011516905.2 XM_011516906.2 XM_017013168.1 XM_017013169.1 XM_017013170.1 XM_017013171.1 XM_017013172.1 XM_017013173.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5484962 c.165C>T p.D55D Chr.8: 143589917-143589917 on Build GRCh38
4969708 c.177C>T p.D59D Chr.8: 143589905-143589905 on Build GRCh38
4844845 c.318C>A p.L106L Chr.8: 143589764-143589764 on Build GRCh38
6651719 c.201C>G p.D67E Chr.8: 143589881-143589881 on Build GRCh38
7121646 c.462G>T p.V154V Chr.8: 143589620-143589620 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375729821 Chr.8: 143589905-143589905 on Build GRCh38
rs769069537 Chr.8: 143590043-143590043 on Build GRCh38
rs777455462 Chr.8: 143589926-143589926 on Build GRCh38
rs759488885 Chr.8: 143590070-143590070 on Build GRCh38
rs777561744 Chr.8: 143590019-143590019 on Build GRCh38
rs759137271 Chr.8: 143589746-143589746 on Build GRCh38
rs781535559 Chr.8: 143589986-143589986 on Build GRCh38
rs778891496 Chr.8: 143589972-143589972 on Build GRCh38
rs750543113 Chr.8: 143589881-143589887 on Build GRCh38
rs369940305 Chr.8: 143589799-143589799 on Build GRCh38
rs767687448 Chr.8: 143589999-143589999 on Build GRCh38
rs756961336 Chr.8: 143590096-143590096 on Build GRCh38
rs765310456 Chr.8: 143589814-143589814 on Build GRCh38
rs376735898 Chr.8: 143589950-143589950 on Build GRCh38
rs992012907 Chr.8: 143589844-143589844 on Build GRCh38
rs138931466 Chr.8: 143589836-143589836 on Build GRCh38
rs776985852 Chr.8: 143589939-143589939 on Build GRCh38
rs146677657 Chr.8: 143589775-143589775 on Build GRCh38
rs759744448 Chr.8: 143589878-143589878 on Build GRCh38
rs756032266 Chr.8: 143589894-143589894 on Build GRCh38
rs768145252 Chr.8: 143589794-143589794 on Build GRCh38
rs377595210 Chr.8: 143589992-143589992 on Build GRCh38
rs143180580 Chr.8: 143589773-143589773 on Build GRCh38
rs761305190 Chr.8: 143589868-143589868 on Build GRCh38
rs762354014 Chr.8: 143589793-143589793 on Build GRCh38
rs780333965 Chr.8: 143590029-143590029 on Build GRCh38
rs764663548 Chr.8: 143589691-143589691 on Build GRCh38
rs371315183 Chr.8: 143589967-143589967 on Build GRCh38
rs773933749 Chr.8: 143589855-143589855 on Build GRCh38
rs140837533 Chr.8: 143589787-143589787 on Build GRCh38
rs1024776639 Chr.8: 143589846-143589846 on Build GRCh38
rs773364490 Chr.8: 143590047-143590047 on Build GRCh38
rs61757370 Chr.8: 143589785-143589785 on Build GRCh38
rs762445559 Chr.8: 143589757-143589757 on Build GRCh38
rs370528981 Chr.8: 143590005-143590005 on Build GRCh38
rs1032810554 Chr.8: 143589718-143589718 on Build GRCh38
rs1014879713 Chr.8: 143589832-143589832 on Build GRCh38
rs768873142 Chr.8: 143589996-143589996 on Build GRCh38
rs372141530 Chr.8: 143589749-143589749 on Build GRCh38
rs760596856 Chr.8: 143589959-143589959 on Build GRCh38
rs370379043 Chr.8: 143589916-143589916 on Build GRCh38
rs747989092 Chr.8: 143589652-143589652 on Build GRCh38
rs774793383 Chr.8: 143589751-143589751 on Build GRCh38
rs780791045 Chr.8: 143590022-143590022 on Build GRCh38
rs754708414 Chr.8: 143589969-143589969 on Build GRCh38
rs771677695 Chr.8: 143589925-143589925 on Build GRCh38
rs573856271 Chr.8: 143589955-143589955 on Build GRCh38
rs772061445 Chr.8: 143589654-143589654 on Build GRCh38
rs373393123 Chr.8: 143589813-143589813 on Build GRCh38
rs568697177 Chr.8: 143589706-143589706 on Build GRCh38
rs191012991 Chr.8: 143590072-143590072 on Build GRCh38
rs540445790 Chr.8: 143589761-143589761 on Build GRCh38
rs754066458 Chr.8: 143589767-143589767 on Build GRCh38
rs747658631 Chr.8: 143589862-143589862 on Build GRCh38
rs749651006 Chr.8: 143590034-143590034 on Build GRCh38
rs1042372972 Chr.8: 143589965-143589965 on Build GRCh38
rs760515138 Chr.8: 143589671-143589671 on Build GRCh38
rs146096793 Chr.8: 143589800-143589800 on Build GRCh38
rs764684665 Chr.8: 143589681-143589681 on Build GRCh38
rs200364826 Chr.8: 143590095-143590095 on Build GRCh38
rs746432478 Chr.8: 143589991-143589991 on Build GRCh38
rs768210001 Chr.8: 143589852-143589852 on Build GRCh38
rs749391938 Chr.8: 143589727-143589727 on Build GRCh38
rs779748464 Chr.8: 143589847-143589847 on Build GRCh38
rs34365735 Chr.8: 143589678-143589678 on Build GRCh38
rs907837150 Chr.8: 143589884-143589884 on Build GRCh38
rs3812447 Chr.8: 143589786-143589786 on Build GRCh38
rs773923508 Chr.8: 143589796-143589796 on Build GRCh38
rs140176916 Chr.8: 143590046-143590046 on Build GRCh38
rs752271362 Chr.8: 143589682-143589682 on Build GRCh38
rs1029485601 Chr.8: 143589873-143589873 on Build GRCh38
rs35331982 Chr.8: 143589735-143589735 on Build GRCh38
rs753872753 Chr.8: 143589968-143589968 on Build GRCh38
rs921506699 Chr.8: 143590058-143590058 on Build GRCh38
rs752554217 Chr.8: 143590014-143590014 on Build GRCh38
rs757117888 Chr.8: 143589825-143589825 on Build GRCh38
rs752079200 Chr.8: 143589754-143589754 on Build GRCh38
rs747852224 Chr.8: 143589920-143589920 on Build GRCh38
rs962682372 Chr.8: 143590013-143590013 on Build GRCh38
rs763566376 Chr.8: 143589753-143589753 on Build GRCh38
rs3812448 Chr.8: 143589752-143589752 on Build GRCh38
rs759550819 Chr.8: 143590008-143590008 on Build GRCh38
rs752068501 Chr.8: 143589697-143589697 on Build GRCh38
rs752981152 Chr.8: 143589886-143589886 on Build GRCh38
rs779942339 Chr.8: 143589724-143589724 on Build GRCh38
rs758056631 Chr.8: 143590017-143590017 on Build GRCh38
rs776440433 Chr.8: 143589744-143589744 on Build GRCh38
rs146141808 Chr.8: 143590045-143590045 on Build GRCh38
rs969667105 Chr.8: 143589985-143589985 on Build GRCh38
rs754059781 Chr.8: 143589809-143589809 on Build GRCh38
rs780320871 Chr.8: 143589766-143589766 on Build GRCh38
rs770538454 Chr.8: 143589984-143589984 on Build GRCh38
rs368871686 Chr.8: 143589888-143589888 on Build GRCh38
rs372434997 Chr.8: 143589893-143589893 on Build GRCh38
rs201992595 Chr.8: 143590091-143590091 on Build GRCh38
rs374763009 Chr.8: 143589929-143589929 on Build GRCh38
rs775663686 Chr.8: 143589949-143589949 on Build GRCh38
rs142962150 Chr.8: 143590006-143590006 on Build GRCh38
rs764114491 Chr.8: 143589951-143589951 on Build GRCh38
rs779843665 Chr.8: 143589904-143589904 on Build GRCh38
rs766882970 Chr.8: 143590098-143590098 on Build GRCh38
rs148849920 Chr.8: 143589781-143589781 on Build GRCh38
rs185413566 Chr.8: 143589896-143589896 on Build GRCh38
rs144264495 Chr.8: 143589973-143589973 on Build GRCh38
rs369086189 Chr.8: 143589699-143589699 on Build GRCh38
rs772817824 Chr.8: 143589664-143589664 on Build GRCh38
rs752786599 Chr.8: 143589815-143589815 on Build GRCh38
rs770874884 Chr.8: 143589776-143589776 on Build GRCh38
rs747247690 Chr.8: 143589917-143589917 on Build GRCh38
rs375747373 Chr.8: 143590086-143590086 on Build GRCh38
rs770199997 Chr.8: 143589945-143589945 on Build GRCh38
rs144750496 Chr.8: 143589994-143589994 on Build GRCh38
rs755911229 Chr.8: 143589845-143589845 on Build GRCh38
rs763307061 Chr.8: 143589687-143589687 on Build GRCh38
rs201319490 Chr.8: 143590087-143590087 on Build GRCh38
rs766220330 Chr.8: 143589672-143589672 on Build GRCh38
rs1004196937 Chr.8: 143589819-143589819 on Build GRCh38
rs774705087 Chr.8: 143589997-143589997 on Build GRCh38
rs773634871 Chr.8: 143589788-143589788 on Build GRCh38
rs376504350 Chr.8: 143589848-143589848 on Build GRCh38
rs34547530 Chr.8: 143589877-143589877 on Build GRCh38
rs746505007 Chr.8: 143589978-143589978 on Build GRCh38
rs756698312 Chr.8: 143590020-143590020 on Build GRCh38
rs148133232 Chr.8: 143589998-143589998 on Build GRCh38
rs936359272 Chr.8: 143589881-143589881 on Build GRCh38
rs776243280 Chr.8: 143589677-143589677 on Build GRCh38
rs924906711 Chr.8: 143589758-143589758 on Build GRCh38
rs759812478 Chr.8: 143589944-143589944 on Build GRCh38
rs773833658 Chr.8: 143589867-143589867 on Build GRCh38
rs746317133 Chr.8: 143590023-143590023 on Build GRCh38
rs112907282 Chr.8: 143589865-143589865 on Build GRCh38
rs745664828 Chr.8: 143589782-143589782 on Build GRCh38
rs777013173 Chr.8: 143589783-143589783 on Build GRCh38
rs777359937 Chr.8: 143589977-143589977 on Build GRCh38
rs746967313 Chr.8: 143589741-143589741 on Build GRCh38
rs756630296 Chr.8: 143590024-143590024 on Build GRCh38
rs765075693 Chr.8: 143590012-143590012 on Build GRCh38
rs777753354 Chr.8: 143589737-143589737 on Build GRCh38
rs375424802 Chr.8: 143589762-143589762 on Build GRCh38
rs150053229 Chr.8: 143589705-143589705 on Build GRCh38
rs765379444 Chr.8: 143589880-143589880 on Build GRCh38
rs770680615 Chr.8: 143589743-143589743 on Build GRCh38
rs868363652 Chr.8: 143589907-143589907 on Build GRCh38
rs766726716 Chr.8: 143589875-143589875 on Build GRCh38

View Full Product Details