Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 143868325-143868829 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPPK1
Entrez Gene ID
83481
Gene Name
epiplakin 1
Gene Aliases
EPIPL, EPIPL1
UniGene
Hs.200412
Gene Chromosome Location
Chr.8: 143857319-143879043 on Build GRCh38
Transcripts
NM_031308.3 XM_017013889.1 XM_017013890.1 XM_017013891.1 XM_017013892.1

SNP Detail

SNP ID Chromosome Location
rs782552852 Chr.8: 143868356-143868356 on Build GRCh38
rs569107828 Chr.8: 143868572-143868572 on Build GRCh38
rs782354255 Chr.8: 143868581-143868581 on Build GRCh38
rs191532254 Chr.8: 143868618-143868618 on Build GRCh38
rs782537971 Chr.8: 143868724-143868724 on Build GRCh38
rs1037437050 Chr.8: 143868348-143868348 on Build GRCh38
rs782104048 Chr.8: 143868558-143868558 on Build GRCh38
rs199851232 Chr.8: 143868354-143868354 on Build GRCh38
rs201538740 Chr.8: 143868565-143868565 on Build GRCh38
rs782526891 Chr.8: 143868557-143868557 on Build GRCh38
rs983173970 Chr.8: 143868791-143868791 on Build GRCh38
rs117131125 Chr.8: 143868370-143868370 on Build GRCh38
rs782003091 Chr.8: 143868390-143868390 on Build GRCh38
rs782603702 Chr.8: 143868371-143868371 on Build GRCh38
rs376180791 Chr.8: 143868465-143868465 on Build GRCh38
rs557769989 Chr.8: 143868614-143868614 on Build GRCh38
rs781830503 Chr.8: 143868489-143868489 on Build GRCh38
rs373737061 Chr.8: 143868626-143868626 on Build GRCh38
rs782309931 Chr.8: 143868762-143868762 on Build GRCh38
rs368146843 Chr.8: 143868705-143868705 on Build GRCh38
rs782126049 Chr.8: 143868481-143868481 on Build GRCh38
rs782435054 Chr.8: 143868379-143868379 on Build GRCh38
rs6558399 Chr.8: 143868735-143868735 on Build GRCh38
rs782728846 Chr.8: 143868692-143868692 on Build GRCh38
rs782805448 Chr.8: 143868773-143868773 on Build GRCh38
rs782231061 Chr.8: 143868373-143868373 on Build GRCh38
rs782817925 Chr.8: 143868361-143868361 on Build GRCh38
rs146005478 Chr.8: 143868744-143868744 on Build GRCh38
rs368863995 Chr.8: 143868560-143868560 on Build GRCh38
rs201357174 Chr.8: 143868454-143868454 on Build GRCh38
rs782143817 Chr.8: 143868767-143868767 on Build GRCh38
rs782458222 Chr.8: 143868635-143868635 on Build GRCh38
rs144286038 Chr.8: 143868532-143868532 on Build GRCh38
rs906336420 Chr.8: 143868541-143868541 on Build GRCh38
rs988104020 Chr.8: 143868426-143868426 on Build GRCh38
rs551340158 Chr.8: 143868809-143868809 on Build GRCh38
rs540029263 Chr.8: 143868381-143868381 on Build GRCh38
rs782578844 Chr.8: 143868568-143868568 on Build GRCh38
rs782051022 Chr.8: 143868699-143868699 on Build GRCh38
rs781882163 Chr.8: 143868493-143868493 on Build GRCh38
rs782125557 Chr.8: 143868690-143868690 on Build GRCh38
rs573015971 Chr.8: 143868664-143868664 on Build GRCh38
rs781892821 Chr.8: 143868637-143868637 on Build GRCh38
rs782364262 Chr.8: 143868647-143868647 on Build GRCh38
rs111431754 Chr.8: 143868499-143868499 on Build GRCh38
rs782529811 Chr.8: 143868622-143868622 on Build GRCh38
rs35301416 Chr.8: 143868659-143868660 on Build GRCh38
rs782727181 Chr.8: 143868795-143868795 on Build GRCh38
rs781810446 Chr.8: 143868410-143868410 on Build GRCh38
rs782064600 Chr.8: 143868675-143868675 on Build GRCh38
rs782792511 Chr.8: 143868403-143868403 on Build GRCh38
rs781803355 Chr.8: 143868542-143868542 on Build GRCh38
rs782093131 Chr.8: 143868759-143868759 on Build GRCh38
rs148755111 Chr.8: 143868553-143868553 on Build GRCh38
rs370108979 Chr.8: 143868537-143868537 on Build GRCh38
rs964881215 Chr.8: 143868764-143868764 on Build GRCh38
rs114155737 Chr.8: 143868766-143868766 on Build GRCh38
rs782787968 Chr.8: 143868406-143868406 on Build GRCh38
rs782627172 Chr.8: 143868518-143868518 on Build GRCh38
rs182729035 Chr.8: 143868467-143868467 on Build GRCh38
rs782006083 Chr.8: 143868463-143868463 on Build GRCh38
rs529491804 Chr.8: 143868736-143868736 on Build GRCh38
rs782186468 Chr.8: 143868585-143868585 on Build GRCh38
rs373681462 Chr.8: 143868640-143868640 on Build GRCh38
rs782704465 Chr.8: 143868409-143868409 on Build GRCh38
rs782742738 Chr.8: 143868486-143868486 on Build GRCh38
rs190800981 Chr.8: 143868660-143868660 on Build GRCh38
rs782032010 Chr.8: 143868599-143868599 on Build GRCh38
rs781910932 Chr.8: 143868582-143868582 on Build GRCh38
rs781838496 Chr.8: 143868561-143868561 on Build GRCh38
rs372624077 Chr.8: 143868806-143868806 on Build GRCh38
rs782810532 Chr.8: 143868680-143868680 on Build GRCh38
rs782760991 Chr.8: 143868803-143868803 on Build GRCh38
rs782296613 Chr.8: 143868450-143868450 on Build GRCh38
rs782079467 Chr.8: 143868349-143868349 on Build GRCh38
rs782669255 Chr.8: 143868441-143868441 on Build GRCh38
rs781963854 Chr.8: 143868616-143868616 on Build GRCh38
rs782209179 Chr.8: 143868391-143868391 on Build GRCh38
rs6994315 Chr.8: 143868631-143868631 on Build GRCh38
rs781841573 Chr.8: 143868693-143868693 on Build GRCh38
rs782451931 Chr.8: 143868567-143868567 on Build GRCh38
rs532952345 Chr.8: 143868464-143868464 on Build GRCh38
rs201611695 Chr.8: 143868689-143868698 on Build GRCh38
rs782374942 Chr.8: 143868385-143868385 on Build GRCh38
rs999317576 Chr.8: 143868598-143868598 on Build GRCh38
rs927766632 Chr.8: 143868657-143868657 on Build GRCh38
rs782432341 Chr.8: 143868491-143868491 on Build GRCh38
rs782009256 Chr.8: 143868651-143868651 on Build GRCh38
rs373864197 Chr.8: 143868393-143868393 on Build GRCh38
rs1017745299 Chr.8: 143868758-143868758 on Build GRCh38
rs781881083 Chr.8: 143868717-143868717 on Build GRCh38
rs782404065 Chr.8: 143868592-143868592 on Build GRCh38
rs782800740 Chr.8: 143868475-143868475 on Build GRCh38
rs1054923745 Chr.8: 143868672-143868672 on Build GRCh38
rs782119648 Chr.8: 143868404-143868404 on Build GRCh38
rs575607968 Chr.8: 143868347-143868347 on Build GRCh38
rs566572612 Chr.8: 143868480-143868480 on Build GRCh38
rs782266342 Chr.8: 143868505-143868505 on Build GRCh38
rs782384260 Chr.8: 143868455-143868455 on Build GRCh38
rs910830625 Chr.8: 143868442-143868442 on Build GRCh38
rs782623089 Chr.8: 143868731-143868731 on Build GRCh38
rs186732658 Chr.8: 143868604-143868604 on Build GRCh38
rs782353672 Chr.8: 143868392-143868392 on Build GRCh38
rs782125308 Chr.8: 143868783-143868783 on Build GRCh38
rs782373090 Chr.8: 143868745-143868745 on Build GRCh38
rs782212513 Chr.8: 143868580-143868580 on Build GRCh38
rs201986967 Chr.8: 143868646-143868646 on Build GRCh38
rs373861393 Chr.8: 143868681-143868681 on Build GRCh38
rs894986070 Chr.8: 143868673-143868673 on Build GRCh38
rs782477999 Chr.8: 143868727-143868727 on Build GRCh38
rs267601818 Chr.8: 143868682-143868682 on Build GRCh38
rs781866214 Chr.8: 143868363-143868363 on Build GRCh38
rs782355139 Chr.8: 143868544-143868545 on Build GRCh38
rs782191264 Chr.8: 143868732-143868732 on Build GRCh38
rs782183903 Chr.8: 143868520-143868520 on Build GRCh38
rs375831338 Chr.8: 143868534-143868534 on Build GRCh38
rs527426145 Chr.8: 143868526-143868526 on Build GRCh38
rs367640883 Chr.8: 143868804-143868804 on Build GRCh38
rs191674254 Chr.8: 143868352-143868352 on Build GRCh38
rs368225830 Chr.8: 143868515-143868515 on Build GRCh38
rs782562143 Chr.8: 143868411-143868411 on Build GRCh38
rs781802200 Chr.8: 143868355-143868355 on Build GRCh38
rs545364230 Chr.8: 143868613-143868613 on Build GRCh38
rs781951311 Chr.8: 143868752-143868752 on Build GRCh38
rs781847075 Chr.8: 143868782-143868782 on Build GRCh38
rs201170631 Chr.8: 143868402-143868402 on Build GRCh38
rs782176868 Chr.8: 143868435-143868435 on Build GRCh38
rs370414140 Chr.8: 143868655-143868655 on Build GRCh38
rs201982226 Chr.8: 143868807-143868807 on Build GRCh38
rs782500204 Chr.8: 143868576-143868576 on Build GRCh38
rs782020246 Chr.8: 143868528-143868528 on Build GRCh38
rs782114813 Chr.8: 143868531-143868531 on Build GRCh38
rs782325076 Chr.8: 143868438-143868438 on Build GRCh38
rs954912892 Chr.8: 143868424-143868424 on Build GRCh38
rs782581659 Chr.8: 143868492-143868492 on Build GRCh38
rs782071034 Chr.8: 143868470-143868470 on Build GRCh38
rs782205400 Chr.8: 143868810-143868810 on Build GRCh38
rs544519274 Chr.8: 143868420-143868420 on Build GRCh38

View Full Product Details