Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 16409423-16409917 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BNC2
Entrez Gene ID
54796
Gene Name
basonuclin 2
Gene Aliases
BSN2
UniGene
Hs.656581
Gene Chromosome Location
Chr.9: 16409503-16870838 on Build GRCh38
Transcripts
NM_001317939.1 NM_001317940.1 NM_017637.5 XM_011517924.1 XM_017014816.1 XM_017014817.1 XM_017014818.1 XM_017014820.1 XM_017014822.1 XM_017014823.1 XM_017014824.1 XM_017014825.1 XM_017014829.1

SNP Detail

SNP ID Chromosome Location
rs954420903 Chr.9: 16409636-16409636 on Build GRCh38
rs572880463 Chr.9: 16409536-16409536 on Build GRCh38
rs917951110 Chr.9: 16409452-16409452 on Build GRCh38
rs967671057 Chr.9: 16409604-16409604 on Build GRCh38
rs751566326 Chr.9: 16409813-16409813 on Build GRCh38
rs557316005 Chr.9: 16409484-16409484 on Build GRCh38
rs966183289 Chr.9: 16409695-16409695 on Build GRCh38
rs1053582113 Chr.9: 16409564-16409564 on Build GRCh38
rs911673753 Chr.9: 16409658-16409658 on Build GRCh38
rs754843055 Chr.9: 16409873-16409873 on Build GRCh38
rs930929860 Chr.9: 16409453-16409453 on Build GRCh38
rs540297434 Chr.9: 16409574-16409574 on Build GRCh38
rs895638605 Chr.9: 16409497-16409498 on Build GRCh38
rs544815767 Chr.9: 16409650-16409650 on Build GRCh38
rs903018153 Chr.9: 16409887-16409887 on Build GRCh38
rs539544317 Chr.9: 16409615-16409615 on Build GRCh38
rs1049506255 Chr.9: 16409457-16409457 on Build GRCh38
rs973149532 Chr.9: 16409674-16409674 on Build GRCh38
rs905202536 Chr.9: 16409472-16409472 on Build GRCh38
rs556257053 Chr.9: 16409654-16409654 on Build GRCh38
rs761592476 Chr.9: 16409493-16409493 on Build GRCh38
rs554989663 Chr.9: 16409875-16409875 on Build GRCh38
rs576150288 Chr.9: 16409719-16409719 on Build GRCh38
rs963495278 Chr.9: 16409653-16409653 on Build GRCh38
rs896549693 Chr.9: 16409855-16409855 on Build GRCh38
rs926606052 Chr.9: 16409792-16409792 on Build GRCh38
rs949383058 Chr.9: 16409465-16409465 on Build GRCh38
rs762481770 Chr.9: 16409715-16409715 on Build GRCh38
rs75992648 Chr.9: 16409638-16409638 on Build GRCh38
rs926635066 Chr.9: 16409845-16409845 on Build GRCh38
rs1056453553 Chr.9: 16409823-16409823 on Build GRCh38
rs150867944 Chr.9: 16409769-16409769 on Build GRCh38
rs892282931 Chr.9: 16409586-16409586 on Build GRCh38
rs1000712890 Chr.9: 16409541-16409541 on Build GRCh38
rs930857948 Chr.9: 16409741-16409741 on Build GRCh38
rs142148839 Chr.9: 16409616-16409616 on Build GRCh38
rs539088830 Chr.9: 16409474-16409474 on Build GRCh38
rs983042655 Chr.9: 16409749-16409749 on Build GRCh38
rs775685234 Chr.9: 16409447-16409447 on Build GRCh38
rs1041906828 Chr.9: 16409874-16409874 on Build GRCh38
rs1049323777 Chr.9: 16409754-16409754 on Build GRCh38
rs918137289 Chr.9: 16409760-16409760 on Build GRCh38
rs981257022 Chr.9: 16409464-16409464 on Build GRCh38
rs1000125197 Chr.9: 16409610-16409610 on Build GRCh38
rs1045436947 Chr.9: 16409814-16409814 on Build GRCh38
rs200219996 Chr.9: 16409803-16409808 on Build GRCh38

View Full Product Details