Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 34722946-34723448 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM205A
Entrez Gene ID
259308
Gene Name
family with sequence similarity 205 member A
Gene Aliases
C9orf144B
UniGene
Hs.522093
Gene Chromosome Location
Chr.9: 34723053-34729538 on Build GRCh38
Transcripts
NM_001141917.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1134317 c.3908C>T p.S1303F Chr.9: 34723332-34723332 on Build GRCh38
5659774 c.3992C>A p.P1331Q Chr.9: 34723248-34723248 on Build GRCh38
6294115 c.3797C>A p.T1266K Chr.9: 34723443-34723443 on Build GRCh38
4491890 c.3882C>T p.S1294S Chr.9: 34723358-34723358 on Build GRCh38
5890413 c.3889G>A p.D1297N Chr.9: 34723351-34723351 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374244221 Chr.9: 34723254-34723254 on Build GRCh38
rs769461122 Chr.9: 34723117-34723117 on Build GRCh38
rs758533151 Chr.9: 34723282-34723282 on Build GRCh38
rs190400553 Chr.9: 34723281-34723281 on Build GRCh38
rs938712929 Chr.9: 34723375-34723375 on Build GRCh38
rs542039696 Chr.9: 34723264-34723264 on Build GRCh38
rs780947814 Chr.9: 34723300-34723300 on Build GRCh38
rs751471024 Chr.9: 34723000-34723000 on Build GRCh38
rs565452547 Chr.9: 34723359-34723359 on Build GRCh38
rs928661981 Chr.9: 34723196-34723196 on Build GRCh38
rs7047839 Chr.9: 34723197-34723197 on Build GRCh38
rs1043247443 Chr.9: 34723243-34723243 on Build GRCh38
rs61732416 Chr.9: 34723342-34723342 on Build GRCh38
rs950582120 Chr.9: 34723140-34723140 on Build GRCh38
rs117225869 Chr.9: 34723027-34723027 on Build GRCh38
rs773916038 Chr.9: 34723352-34723352 on Build GRCh38
rs370804526 Chr.9: 34723229-34723229 on Build GRCh38
rs552249182 Chr.9: 34723386-34723386 on Build GRCh38
rs984602390 Chr.9: 34723158-34723158 on Build GRCh38
rs367568564 Chr.9: 34723226-34723226 on Build GRCh38
rs2297132 Chr.9: 34723184-34723184 on Build GRCh38
rs745702833 Chr.9: 34723317-34723317 on Build GRCh38
rs755938714 Chr.9: 34723333-34723333 on Build GRCh38
rs10814138 Chr.9: 34723038-34723038 on Build GRCh38
rs747553525 Chr.9: 34723353-34723353 on Build GRCh38
rs374781990 Chr.9: 34723341-34723341 on Build GRCh38
rs375920889 Chr.9: 34723391-34723391 on Build GRCh38
rs142157224 Chr.9: 34723153-34723153 on Build GRCh38
rs1053391635 Chr.9: 34723218-34723218 on Build GRCh38
rs980635030 Chr.9: 34723343-34723343 on Build GRCh38
rs538479960 Chr.9: 34722979-34722979 on Build GRCh38
rs759995803 Chr.9: 34723394-34723394 on Build GRCh38
rs531395284 Chr.9: 34723261-34723261 on Build GRCh38
rs762588338 Chr.9: 34723182-34723182 on Build GRCh38
rs527603144 Chr.9: 34723286-34723286 on Build GRCh38
rs753851740 Chr.9: 34723208-34723208 on Build GRCh38
rs1017350339 Chr.9: 34723270-34723270 on Build GRCh38
rs1017694958 Chr.9: 34723015-34723015 on Build GRCh38
rs546555671 Chr.9: 34723296-34723296 on Build GRCh38
rs919157003 Chr.9: 34723048-34723048 on Build GRCh38
rs947624054 Chr.9: 34723405-34723405 on Build GRCh38
rs541568272 Chr.9: 34723052-34723052 on Build GRCh38
rs368078669 Chr.9: 34723418-34723418 on Build GRCh38
rs1025907802 Chr.9: 34723294-34723294 on Build GRCh38
rs1043319691 Chr.9: 34723407-34723407 on Build GRCh38
rs367965087 Chr.9: 34723269-34723269 on Build GRCh38
rs928719759 Chr.9: 34723374-34723374 on Build GRCh38
rs909026433 Chr.9: 34723181-34723181 on Build GRCh38
rs950654969 Chr.9: 34723309-34723309 on Build GRCh38
rs757235713 Chr.9: 34723299-34723299 on Build GRCh38
rs775757882 Chr.9: 34723424-34723424 on Build GRCh38
rs569375586 Chr.9: 34723402-34723402 on Build GRCh38
rs372666371 Chr.9: 34723345-34723345 on Build GRCh38
rs201220429 Chr.9: 34723372-34723372 on Build GRCh38
rs777420676 Chr.9: 34722975-34722975 on Build GRCh38
rs200184020 Chr.9: 34723361-34723361 on Build GRCh38
rs532422895 Chr.9: 34723351-34723351 on Build GRCh38

View Full Product Details