Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 35660682-35661180 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGEF39
Entrez Gene ID
84904
Gene Name
Rho guanine nucleotide exchange factor 39
Gene Aliases
C9orf100
UniGene
Hs.534579
Gene Chromosome Location
Chr.9: 35658875-35665281 on Build GRCh38
Transcripts
NM_032818.2 XM_011518057.2 XM_017015225.1 XM_017015227.1 NM_001195200.1 NM_001195201.1 NM_001195217.1 NM_174923.2 XM_005251403.4

Gene Symbol
CCDC107
Entrez Gene ID
203260
Gene Name
coiled-coil domain containing 107
Gene Aliases
PSEC0222
UniGene
Hs.745107
Gene Chromosome Location
Chr.9: 35658282-35661503 on Build GRCh38
Transcripts
NM_032818.2 XM_011518057.2 XM_017015225.1 XM_017015227.1 NM_001195200.1 NM_001195201.1 NM_001195217.1 NM_174923.2 XM_005251403.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6621254 c.494C>T p.P165L Chr.9: 35660829-35660829 on Build GRCh38
6837410 c.461G>T p.W154L Chr.9: 35660796-35660796 on Build GRCh38
3907188 c.749G>A p.R250Q Chr.9: 35661084-35661084 on Build GRCh38
223327 c.535G>A p.G179R Chr.9: 35660870-35660870 on Build GRCh38
6621253 c.583T>C p.F195L Chr.9: 35660918-35660918 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776277883 Chr.9: 35660711-35660711 on Build GRCh38
rs756761846 Chr.9: 35661084-35661084 on Build GRCh38
rs10441686 Chr.9: 35661091-35661091 on Build GRCh38
rs756667454 Chr.9: 35660896-35660897 on Build GRCh38
rs561444311 Chr.9: 35660712-35660712 on Build GRCh38
rs1042864491 Chr.9: 35661052-35661052 on Build GRCh38
rs763119314 Chr.9: 35661133-35661133 on Build GRCh38
rs374363949 Chr.9: 35660816-35660816 on Build GRCh38
rs777404170 Chr.9: 35661103-35661104 on Build GRCh38
rs750964618 Chr.9: 35660897-35660898 on Build GRCh38
rs757681889 Chr.9: 35660783-35660783 on Build GRCh38
rs748224933 Chr.9: 35661123-35661123 on Build GRCh38
rs534657208 Chr.9: 35661001-35661004 on Build GRCh38
rs748919859 Chr.9: 35660910-35660910 on Build GRCh38
rs770497181 Chr.9: 35660917-35660917 on Build GRCh38
rs748763882 Chr.9: 35660997-35660997 on Build GRCh38
rs188379610 Chr.9: 35660876-35660876 on Build GRCh38
rs754479840 Chr.9: 35660975-35660977 on Build GRCh38
rs373051260 Chr.9: 35660786-35660786 on Build GRCh38
rs10441685 Chr.9: 35661060-35661060 on Build GRCh38
rs754250887 Chr.9: 35660756-35660756 on Build GRCh38
rs763277783 Chr.9: 35661065-35661065 on Build GRCh38
rs377274752 Chr.9: 35661148-35661148 on Build GRCh38
rs895168551 Chr.9: 35660766-35660766 on Build GRCh38
rs778200263 Chr.9: 35660753-35660753 on Build GRCh38
rs145670939 Chr.9: 35660830-35660830 on Build GRCh38
rs769809480 Chr.9: 35661125-35661125 on Build GRCh38
rs147165051 Chr.9: 35660994-35660994 on Build GRCh38
rs770806021 Chr.9: 35660815-35660815 on Build GRCh38
rs113536720 Chr.9: 35661097-35661097 on Build GRCh38
rs778499991 Chr.9: 35661009-35661009 on Build GRCh38
rs747714583 Chr.9: 35660991-35660993 on Build GRCh38
rs879048977 Chr.9: 35661078-35661078 on Build GRCh38
rs753194449 Chr.9: 35661149-35661149 on Build GRCh38
rs747669830 Chr.9: 35660801-35660801 on Build GRCh38
rs746904340 Chr.9: 35661056-35661056 on Build GRCh38
rs767373279 Chr.9: 35660823-35660823 on Build GRCh38
rs201575851 Chr.9: 35661066-35661066 on Build GRCh38
rs756924613 Chr.9: 35660998-35660998 on Build GRCh38
rs144841450 Chr.9: 35660920-35660920 on Build GRCh38
rs752815799 Chr.9: 35660826-35660826 on Build GRCh38
rs752108004 Chr.9: 35660984-35660984 on Build GRCh38
rs773360401 Chr.9: 35661126-35661126 on Build GRCh38
rs372621134 Chr.9: 35661094-35661094 on Build GRCh38
rs41298216 Chr.9: 35661147-35661147 on Build GRCh38
rs199991115 Chr.9: 35661058-35661058 on Build GRCh38
rs140449147 Chr.9: 35660787-35660787 on Build GRCh38
rs142365691 Chr.9: 35660847-35660847 on Build GRCh38
rs879031515 Chr.9: 35660963-35660963 on Build GRCh38
rs768717463 Chr.9: 35661057-35661057 on Build GRCh38
rs2275420 Chr.9: 35660904-35660904 on Build GRCh38
rs780613843 Chr.9: 35660796-35660796 on Build GRCh38
rs769419069 Chr.9: 35660806-35660806 on Build GRCh38
rs746812364 Chr.9: 35661115-35661115 on Build GRCh38
rs1017031441 Chr.9: 35660914-35660914 on Build GRCh38
rs764380931 Chr.9: 35660843-35660843 on Build GRCh38
rs774688219 Chr.9: 35661143-35661143 on Build GRCh38
rs761267468 Chr.9: 35661153-35661153 on Build GRCh38
rs565195727 Chr.9: 35660829-35660829 on Build GRCh38
rs766678660 Chr.9: 35661067-35661067 on Build GRCh38
rs199818206 Chr.9: 35661031-35661031 on Build GRCh38
rs747649230 Chr.9: 35660893-35660893 on Build GRCh38
rs779926968 Chr.9: 35661098-35661098 on Build GRCh38
rs779751434 Chr.9: 35660962-35660962 on Build GRCh38
rs756350610 Chr.9: 35660745-35660745 on Build GRCh38
rs71521284 Chr.9: 35661017-35661017 on Build GRCh38
rs765492244 Chr.9: 35661061-35661061 on Build GRCh38
rs775450547 Chr.9: 35661036-35661036 on Build GRCh38
rs767597591 Chr.9: 35660732-35660732 on Build GRCh38
rs778711357 Chr.9: 35660981-35660983 on Build GRCh38
rs751950189 Chr.9: 35661070-35661070 on Build GRCh38
rs747003370 Chr.9: 35661115-35661115 on Build GRCh38
rs145124138 Chr.9: 35660884-35660884 on Build GRCh38
rs772485080 Chr.9: 35660795-35660795 on Build GRCh38
rs374125478 Chr.9: 35660948-35660948 on Build GRCh38
rs35921814 Chr.9: 35661010-35661010 on Build GRCh38
rs750784999 Chr.9: 35660965-35660965 on Build GRCh38
rs753387364 Chr.9: 35661083-35661083 on Build GRCh38
rs766836308 Chr.9: 35660983-35660983 on Build GRCh38
rs3174266 Chr.9: 35660719-35660719 on Build GRCh38
rs561977892 Chr.9: 35661119-35661119 on Build GRCh38
rs149423190 Chr.9: 35660918-35660918 on Build GRCh38
rs769329190 Chr.9: 35660902-35660902 on Build GRCh38
rs766597639 Chr.9: 35660964-35660964 on Build GRCh38
rs1339374 Chr.9: 35660993-35660993 on Build GRCh38
rs764221631 Chr.9: 35660947-35660947 on Build GRCh38
rs758752401 Chr.9: 35660977-35660977 on Build GRCh38
rs755422129 Chr.9: 35661071-35661071 on Build GRCh38
rs753385503 Chr.9: 35660968-35660968 on Build GRCh38
rs773343294 Chr.9: 35661064-35661064 on Build GRCh38
rs768700936 Chr.9: 35661116-35661116 on Build GRCh38
rs202093963 Chr.9: 35660807-35660807 on Build GRCh38
rs371820543 Chr.9: 35660926-35660926 on Build GRCh38
rs141349284 Chr.9: 35660875-35660875 on Build GRCh38
rs745464083 Chr.9: 35661092-35661092 on Build GRCh38
rs367950013 Chr.9: 35660855-35660855 on Build GRCh38
rs762343169 Chr.9: 35660868-35660868 on Build GRCh38
rs752832075 Chr.9: 35660740-35660740 on Build GRCh38
rs767970537 Chr.9: 35661072-35661072 on Build GRCh38
rs756504155 Chr.9: 35660955-35660955 on Build GRCh38
rs533224381 Chr.9: 35660821-35660821 on Build GRCh38
rs151243859 Chr.9: 35660958-35660958 on Build GRCh38
rs766603469 Chr.9: 35661135-35661135 on Build GRCh38
rs147969979 Chr.9: 35660921-35660921 on Build GRCh38

View Full Product Details