Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 36215325-36215865 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNE
Entrez Gene ID
10020
Gene Name
glucosamine (UDP-N-acetyl)-2-epimerase/N-acetylmannosamine kinase
Gene Aliases
DMRV, GLCNE, IBM2, NM, Uae1
UniGene
Hs.5920
Gene Chromosome Location
Chr.9: 36214441-36277056 on Build GRCh38
Transcripts
NM_001128227.2 NM_001190383.1 NM_001190384.1 NM_001190388.1 NM_005476.5 XM_017014167.1 XM_017014168.1

SNP Detail

SNP ID Chromosome Location
rs190727651 Chr.9: 36215730-36215730 on Build GRCh38
rs886063918 Chr.9: 36215671-36215671 on Build GRCh38
rs867558953 Chr.9: 36215726-36215726 on Build GRCh38
rs559424901 Chr.9: 36215386-36215386 on Build GRCh38
rs983349971 Chr.9: 36215596-36215596 on Build GRCh38
rs537436586 Chr.9: 36215827-36215827 on Build GRCh38
rs977758584 Chr.9: 36215572-36215572 on Build GRCh38
rs16933086 Chr.9: 36215584-36215584 on Build GRCh38
rs568921977 Chr.9: 36215825-36215825 on Build GRCh38
rs959053325 Chr.9: 36215513-36215513 on Build GRCh38
rs780300628 Chr.9: 36215743-36215743 on Build GRCh38
rs958344546 Chr.9: 36215387-36215387 on Build GRCh38
rs992534852 Chr.9: 36215393-36215393 on Build GRCh38
rs528538484 Chr.9: 36215391-36215391 on Build GRCh38
rs567820407 Chr.9: 36215686-36215686 on Build GRCh38
rs1033416837 Chr.9: 36215448-36215448 on Build GRCh38
rs1001017679 Chr.9: 36215771-36215771 on Build GRCh38
rs916906578 Chr.9: 36215399-36215399 on Build GRCh38
rs760418417 Chr.9: 36215428-36215428 on Build GRCh38
rs1000654782 Chr.9: 36215413-36215413 on Build GRCh38
rs7044157 Chr.9: 36215539-36215539 on Build GRCh38
rs751849626 Chr.9: 36215745-36215745 on Build GRCh38
rs41277097 Chr.9: 36215637-36215637 on Build GRCh38
rs909083594 Chr.9: 36215607-36215607 on Build GRCh38
rs1053779509 Chr.9: 36215697-36215697 on Build GRCh38
rs761447556 Chr.9: 36215790-36215790 on Build GRCh38
rs995029905 Chr.9: 36215833-36215833 on Build GRCh38
rs894910208 Chr.9: 36215804-36215804 on Build GRCh38
rs1034130570 Chr.9: 36215350-36215350 on Build GRCh38
rs1005111949 Chr.9: 36215761-36215762 on Build GRCh38
rs552282953 Chr.9: 36215780-36215780 on Build GRCh38
rs763907827 Chr.9: 36215514-36215514 on Build GRCh38
rs963168796 Chr.9: 36215614-36215614 on Build GRCh38
rs921366466 Chr.9: 36215657-36215657 on Build GRCh38
rs750509496 Chr.9: 36215700-36215700 on Build GRCh38
rs544309845 Chr.9: 36215435-36215435 on Build GRCh38
rs530317139 Chr.9: 36215687-36215687 on Build GRCh38
rs1024615466 Chr.9: 36215565-36215565 on Build GRCh38
rs185870650 Chr.9: 36215705-36215705 on Build GRCh38
rs775057401 Chr.9: 36215356-36215356 on Build GRCh38
rs538774065 Chr.9: 36215764-36215764 on Build GRCh38

View Full Product Details