Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 121098244-121098699 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNTRL
Entrez Gene ID
11064
Gene Name
centriolin
Gene Aliases
CEP1, CEP110, FAN, bA165P4.1
UniGene
Hs.653263
Gene Chromosome Location
Chr.9: 121074778-121177610 on Build GRCh38
Transcripts
NM_007018.4 XM_005251679.3 XM_006716940.2 XM_006716944.3 XM_011518166.1 XM_011518167.1 XM_011518168.1 XM_011518169.2 XM_011518170.1 XM_011518172.2 XM_017014223.1 XM_017014224.1 XM_017014225.1 XM_017014227.1 XM_017014228.1 XM_017014229.1 XM_017014230.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5632655 c.673C>G p.L225V Chr.9: 121098437-121098437 on Build GRCh38
3847526 c.665C>A p.S222Y Chr.9: 121098429-121098429 on Build GRCh38
5359179 c.655G>T p.D219Y Chr.9: 121098419-121098419 on Build GRCh38
3903680 c.760C>T p.P254S Chr.9: 121098524-121098524 on Build GRCh38
7310035 c.645A>G p.K215K Chr.9: 121098409-121098409 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1006910628 Chr.9: 121098327-121098327 on Build GRCh38
rs1002169317 Chr.9: 121098637-121098637 on Build GRCh38
rs888966131 Chr.9: 121098633-121098633 on Build GRCh38
rs1016900699 Chr.9: 121098661-121098661 on Build GRCh38
rs971665424 Chr.9: 121098372-121098372 on Build GRCh38
rs933224324 Chr.9: 121098525-121098525 on Build GRCh38
rs759552851 Chr.9: 121098339-121098339 on Build GRCh38
rs892525330 Chr.9: 121098547-121098547 on Build GRCh38
rs372485353 Chr.9: 121098522-121098522 on Build GRCh38
rs143362843 Chr.9: 121098418-121098418 on Build GRCh38
rs552995922 Chr.9: 121098460-121098460 on Build GRCh38
rs745960037 Chr.9: 121098557-121098557 on Build GRCh38
rs1024954689 Chr.9: 121098279-121098279 on Build GRCh38
rs1005980041 Chr.9: 121098634-121098634 on Build GRCh38
rs760483028 Chr.9: 121098468-121098468 on Build GRCh38
rs772286609 Chr.9: 121098562-121098562 on Build GRCh38
rs779021409 Chr.9: 121098420-121098420 on Build GRCh38
rs753046483 Chr.9: 121098391-121098391 on Build GRCh38
rs749536840 Chr.9: 121098412-121098412 on Build GRCh38
rs181161584 Chr.9: 121098350-121098350 on Build GRCh38
rs1016950140 Chr.9: 121098329-121098329 on Build GRCh38
rs376506159 Chr.9: 121098379-121098379 on Build GRCh38
rs370222223 Chr.9: 121098507-121098507 on Build GRCh38
rs113096477 Chr.9: 121098311-121098311 on Build GRCh38
rs778143798 Chr.9: 121098509-121098509 on Build GRCh38
rs144772143 Chr.9: 121098401-121098401 on Build GRCh38
rs556749701 Chr.9: 121098606-121098606 on Build GRCh38
rs113016876 Chr.9: 121098551-121098551 on Build GRCh38
rs772122904 Chr.9: 121098439-121098439 on Build GRCh38
rs745984126 Chr.9: 121098431-121098431 on Build GRCh38
rs749923533 Chr.9: 121098497-121098497 on Build GRCh38
rs150932215 Chr.9: 121098471-121098471 on Build GRCh38
rs761284046 Chr.9: 121098591-121098592 on Build GRCh38
rs761172082 Chr.9: 121098374-121098374 on Build GRCh38
rs746967651 Chr.9: 121098605-121098605 on Build GRCh38
rs781697307 Chr.9: 121098389-121098389 on Build GRCh38
rs369416576 Chr.9: 121098340-121098340 on Build GRCh38
rs776572660 Chr.9: 121098275-121098275 on Build GRCh38
rs753205863 Chr.9: 121098505-121098505 on Build GRCh38
rs768588394 Chr.9: 121098469-121098469 on Build GRCh38
rs764835008 Chr.9: 121098483-121098483 on Build GRCh38
rs954468769 Chr.9: 121098448-121098448 on Build GRCh38
rs773903978 Chr.9: 121098581-121098581 on Build GRCh38
rs369969301 Chr.9: 121098430-121098430 on Build GRCh38
rs770995226 Chr.9: 121098527-121098527 on Build GRCh38
rs751968056 Chr.9: 121098383-121098383 on Build GRCh38
rs753549081 Chr.9: 121098371-121098371 on Build GRCh38
rs979045287 Chr.9: 121098502-121098502 on Build GRCh38
rs1050243350 Chr.9: 121098590-121098590 on Build GRCh38
rs949499576 Chr.9: 121098583-121098583 on Build GRCh38
rs905438674 Chr.9: 121098333-121098333 on Build GRCh38
rs761555576 Chr.9: 121098481-121098481 on Build GRCh38
rs373021722 Chr.9: 121098363-121098363 on Build GRCh38
rs10818504 Chr.9: 121098411-121098411 on Build GRCh38
rs755476575 Chr.9: 121098388-121098388 on Build GRCh38
rs779062127 Chr.9: 121098530-121098530 on Build GRCh38
rs776452294 Chr.9: 121098611-121098611 on Build GRCh38
rs754197021 Chr.9: 121098520-121098520 on Build GRCh38
rs780084959 Chr.9: 121098584-121098584 on Build GRCh38

View Full Product Details