Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 21497676-21498224 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNKSR2
Entrez Gene ID
22866
Gene Name
connector enhancer of kinase suppressor of Ras 2
Gene Aliases
CNK2, KSR2, MAGUIN
UniGene
Hs.555917
Gene Chromosome Location
Chr.X: 21374335-21654695 on Build GRCh38
Transcripts
NM_001168647.1 NM_001168648.1 NM_001168649.1 NM_014927.3 XM_011545471.2 XM_011545472.2 XM_017029358.1

SNP Detail

SNP ID Chromosome Location
rs777852949 Chr.X: 21497761-21497761 on Build GRCh38
rs771593465 Chr.X: 21497748-21497748 on Build GRCh38
rs996753964 Chr.X: 21497775-21497775 on Build GRCh38
rs915888623 Chr.X: 21498003-21498003 on Build GRCh38
rs776199536 Chr.X: 21497844-21497844 on Build GRCh38
rs1035852539 Chr.X: 21497754-21497754 on Build GRCh38
rs981645975 Chr.X: 21498198-21498198 on Build GRCh38
rs942867835 Chr.X: 21497948-21497948 on Build GRCh38
rs754824253 Chr.X: 21497760-21497760 on Build GRCh38
rs1047522563 Chr.X: 21498078-21498078 on Build GRCh38
rs372333253 Chr.X: 21497850-21497850 on Build GRCh38
rs779011548 Chr.X: 21497783-21497783 on Build GRCh38
rs758464289 Chr.X: 21497896-21497896 on Build GRCh38
rs750379127 Chr.X: 21497782-21497782 on Build GRCh38
rs760248628 Chr.X: 21498001-21498001 on Build GRCh38
rs779864503 Chr.X: 21497897-21497897 on Build GRCh38
rs930048317 Chr.X: 21498076-21498076 on Build GRCh38
rs888433972 Chr.X: 21498086-21498086 on Build GRCh38
rs768363034 Chr.X: 21497984-21497984 on Build GRCh38
rs780004363 Chr.X: 21497755-21497755 on Build GRCh38
rs749461022 Chr.X: 21497766-21497766 on Build GRCh38
rs182700450 Chr.X: 21497718-21497718 on Build GRCh38
rs957441272 Chr.X: 21497961-21497961 on Build GRCh38
rs977289951 Chr.X: 21497907-21497907 on Build GRCh38
rs12387312 Chr.X: 21497898-21497898 on Build GRCh38
rs922685959 Chr.X: 21498016-21498016 on Build GRCh38
rs771539354 Chr.X: 21497911-21497911 on Build GRCh38
rs978971032 Chr.X: 21497908-21497908 on Build GRCh38
rs955181209 Chr.X: 21497886-21497886 on Build GRCh38
rs751136115 Chr.X: 21497986-21497989 on Build GRCh38
rs778038278 Chr.X: 21498103-21498103 on Build GRCh38
rs751332354 Chr.X: 21497757-21497757 on Build GRCh38
rs769002421 Chr.X: 21497853-21497853 on Build GRCh38
rs1001326329 Chr.X: 21497728-21497728 on Build GRCh38
rs775035431 Chr.X: 21497892-21497892 on Build GRCh38
rs941953241 Chr.X: 21498180-21498180 on Build GRCh38
rs1031509221 Chr.X: 21497957-21497957 on Build GRCh38
rs771081623 Chr.X: 21497781-21497781 on Build GRCh38
rs911426205 Chr.X: 21497909-21497909 on Build GRCh38
rs1029611950 Chr.X: 21497777-21497777 on Build GRCh38
rs745987326 Chr.X: 21497795-21497795 on Build GRCh38
rs772687544 Chr.X: 21497822-21497822 on Build GRCh38

View Full Product Details