Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 21526627-21527146 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNKSR2
Entrez Gene ID
22866
Gene Name
connector enhancer of kinase suppressor of Ras 2
Gene Aliases
CNK2, KSR2, MAGUIN
UniGene
Hs.555917
Gene Chromosome Location
Chr.X: 21374335-21654695 on Build GRCh38
Transcripts
NM_001168647.1 NM_001168648.1 NM_001168649.1 NM_014927.3 XM_011545471.2 XM_011545472.2 XM_017029358.1

SNP Detail

SNP ID Chromosome Location
rs113629606 Chr.X: 21526803-21526804 on Build GRCh38
rs781772926 Chr.X: 21526749-21526749 on Build GRCh38
rs773153437 Chr.X: 21526789-21526790 on Build GRCh38
rs760601692 Chr.X: 21526826-21526829 on Build GRCh38
rs965114773 Chr.X: 21527075-21527075 on Build GRCh38
rs748534053 Chr.X: 21526828-21526828 on Build GRCh38
rs903528777 Chr.X: 21526721-21526721 on Build GRCh38
rs756551525 Chr.X: 21526763-21526763 on Build GRCh38
rs759498814 Chr.X: 21526897-21526897 on Build GRCh38
rs780816555 Chr.X: 21526678-21526678 on Build GRCh38
rs376856886 Chr.X: 21526786-21526788 on Build GRCh38
rs192590228 Chr.X: 21526772-21526772 on Build GRCh38
rs753703685 Chr.X: 21527015-21527015 on Build GRCh38
rs369645688 Chr.X: 21526838-21526838 on Build GRCh38
rs760662472 Chr.X: 21527005-21527005 on Build GRCh38
rs774222501 Chr.X: 21526840-21526840 on Build GRCh38
rs5904570 Chr.X: 21526921-21526921 on Build GRCh38
rs772981322 Chr.X: 21526766-21526766 on Build GRCh38
rs773818782 Chr.X: 21526896-21526896 on Build GRCh38
rs763920799 Chr.X: 21527012-21527012 on Build GRCh38
rs767474886 Chr.X: 21526904-21526904 on Build GRCh38
rs1027939182 Chr.X: 21526842-21526842 on Build GRCh38
rs747740203 Chr.X: 21526717-21526717 on Build GRCh38
rs376627210 Chr.X: 21527016-21527016 on Build GRCh38
rs770999019 Chr.X: 21526786-21526787 on Build GRCh38
rs754491959 Chr.X: 21526660-21526660 on Build GRCh38
rs186685084 Chr.X: 21527118-21527118 on Build GRCh38
rs970481526 Chr.X: 21527066-21527066 on Build GRCh38
rs1016321787 Chr.X: 21527056-21527056 on Build GRCh38
rs1043446489 Chr.X: 21526702-21526702 on Build GRCh38
rs375204104 Chr.X: 21526837-21526837 on Build GRCh38
rs373075473 Chr.X: 21526903-21526903 on Build GRCh38
rs980110282 Chr.X: 21527077-21527077 on Build GRCh38
rs769315443 Chr.X: 21526731-21526731 on Build GRCh38
rs953615357 Chr.X: 21526917-21526917 on Build GRCh38
rs750360837 Chr.X: 21526818-21526823 on Build GRCh38
rs912893013 Chr.X: 21527121-21527121 on Build GRCh38
rs926563288 Chr.X: 21527078-21527078 on Build GRCh38
rs373350088 Chr.X: 21526830-21526830 on Build GRCh38
rs912039815 Chr.X: 21527072-21527072 on Build GRCh38
rs779405213 Chr.X: 21526787-21526789 on Build GRCh38
rs182089918 Chr.X: 21526786-21526786 on Build GRCh38
rs766751823 Chr.X: 21527036-21527036 on Build GRCh38
rs904437552 Chr.X: 21526655-21526655 on Build GRCh38
rs775354143 Chr.X: 21526905-21526905 on Build GRCh38
rs748412392 Chr.X: 21526752-21526752 on Build GRCh38
rs962031244 Chr.X: 21526787-21526795 on Build GRCh38

View Full Product Details