Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 40597148-40597676 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP6AP2
Entrez Gene ID
10159
Gene Name
ATPase H+ transporting accessory protein 2
Gene Aliases
APT6M8-9, ATP6IP2, ATP6M8-9, ELDF10, HT028, M8-9, MRXE, MRXSH, MSTP009, PRR, RENR, XMRE, XPDS
UniGene
Hs.495960
Gene Chromosome Location
Chr.X: 40580964-40606637 on Build GRCh38
Transcripts
NM_005765.2

SNP Detail

SNP ID Chromosome Location
rs756186125 Chr.X: 40597394-40597394 on Build GRCh38
rs745734335 Chr.X: 40597584-40597584 on Build GRCh38
rs774980742 Chr.X: 40597283-40597283 on Build GRCh38
rs193191654 Chr.X: 40597227-40597227 on Build GRCh38
rs749077028 Chr.X: 40597239-40597239 on Build GRCh38
rs200933554 Chr.X: 40597217-40597217 on Build GRCh38
rs764281230 Chr.X: 40597455-40597455 on Build GRCh38
rs142175071 Chr.X: 40597333-40597333 on Build GRCh38
rs963044963 Chr.X: 40597604-40597604 on Build GRCh38
rs768196393 Chr.X: 40597377-40597377 on Build GRCh38
rs770593270 Chr.X: 40597251-40597251 on Build GRCh38
rs772002758 Chr.X: 40597272-40597272 on Build GRCh38
rs560852897 Chr.X: 40597528-40597528 on Build GRCh38
rs760027081 Chr.X: 40597626-40597626 on Build GRCh38
rs745748841 Chr.X: 40597263-40597263 on Build GRCh38
rs778703690 Chr.X: 40597253-40597253 on Build GRCh38
rs146371390 Chr.X: 40597593-40597593 on Build GRCh38
rs184634552 Chr.X: 40597585-40597585 on Build GRCh38
rs138458908 Chr.X: 40597599-40597599 on Build GRCh38
rs397518480 Chr.X: 40597293-40597293 on Build GRCh38
rs761547273 Chr.X: 40597336-40597336 on Build GRCh38
rs138952430 Chr.X: 40597305-40597305 on Build GRCh38
rs746499876 Chr.X: 40597594-40597594 on Build GRCh38
rs775818303 Chr.X: 40597646-40597646 on Build GRCh38
rs1012021824 Chr.X: 40597341-40597341 on Build GRCh38
rs761302416 Chr.X: 40597600-40597600 on Build GRCh38
rs376011929 Chr.X: 40597290-40597290 on Build GRCh38
rs766788880 Chr.X: 40597363-40597363 on Build GRCh38
rs752801515 Chr.X: 40597392-40597392 on Build GRCh38
rs372993268 Chr.X: 40597602-40597602 on Build GRCh38
rs778612011 Chr.X: 40597576-40597576 on Build GRCh38
rs767999388 Chr.X: 40597597-40597597 on Build GRCh38
rs369015249 Chr.X: 40597514-40597514 on Build GRCh38
rs768104799 Chr.X: 40597636-40597636 on Build GRCh38
rs768335960 Chr.X: 40597320-40597320 on Build GRCh38
rs121918521 Chr.X: 40597269-40597269 on Build GRCh38
rs142013283 Chr.X: 40597620-40597620 on Build GRCh38
rs761053156 Chr.X: 40597228-40597228 on Build GRCh38
rs146546768 Chr.X: 40597215-40597215 on Build GRCh38
rs970665566 Chr.X: 40597443-40597443 on Build GRCh38
rs866200097 Chr.X: 40597247-40597247 on Build GRCh38
rs757515632 Chr.X: 40597573-40597573 on Build GRCh38
rs759926863 Chr.X: 40597199-40597199 on Build GRCh38
rs1025250054 Chr.X: 40597413-40597413 on Build GRCh38
rs760115306 Chr.X: 40597366-40597366 on Build GRCh38

View Full Product Details