Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 101617794-101618302 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARMCX6
Entrez Gene ID
54470
Gene Name
armadillo repeat containing, X-linked 6
Gene Aliases
GASP10
UniGene
Hs.83530
Gene Chromosome Location
Chr.X: 101615118-101618001 on Build GRCh38
Transcripts
NM_001009584.1 NM_001184768.1 NM_019007.3 NR_033669.1 NR_033670.1 XM_011530976.2

SNP Detail

SNP ID Chromosome Location
rs953856958 Chr.X: 101617820-101617820 on Build GRCh38
rs939903781 Chr.X: 101617940-101617940 on Build GRCh38
rs1055129861 Chr.X: 101617960-101617960 on Build GRCh38
rs1000161830 Chr.X: 101618087-101618087 on Build GRCh38
rs1032598483 Chr.X: 101618107-101618107 on Build GRCh38
rs191104829 Chr.X: 101618197-101618197 on Build GRCh38
rs1047026727 Chr.X: 101618070-101618070 on Build GRCh38
rs902690962 Chr.X: 101618073-101618073 on Build GRCh38
rs1028058122 Chr.X: 101618207-101618207 on Build GRCh38
rs919884406 Chr.X: 101617955-101617955 on Build GRCh38
rs782094229 Chr.X: 101618095-101618095 on Build GRCh38
rs931323001 Chr.X: 101617958-101617958 on Build GRCh38
rs895272143 Chr.X: 101617990-101617990 on Build GRCh38
rs912468030 Chr.X: 101617938-101617938 on Build GRCh38
rs972696773 Chr.X: 101617949-101617949 on Build GRCh38
rs781831805 Chr.X: 101618015-101618015 on Build GRCh38
rs986656502 Chr.X: 101617827-101617827 on Build GRCh38
rs782058785 Chr.X: 101617914-101617914 on Build GRCh38
rs782722655 Chr.X: 101618006-101618006 on Build GRCh38
rs892770352 Chr.X: 101618190-101618190 on Build GRCh38
rs146022941 Chr.X: 101618175-101618175 on Build GRCh38
rs953806262 Chr.X: 101618217-101618217 on Build GRCh38
rs1028082637 Chr.X: 101617817-101617817 on Build GRCh38

View Full Product Details