Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 102750145-102750685 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BHLHB9
Entrez Gene ID
80823
Gene Name
basic helix-loop-helix domain containing, class B, 9
Gene Aliases
GASP3, p60TRP
UniGene
Hs.4276
Gene Chromosome Location
Chr.X: 102720714-102752441 on Build GRCh38
Transcripts
NM_001142524.1 NM_001142525.1 NM_001142526.1 NM_001142527.1 NM_001142528.1 NM_001142529.1 NM_001142530.1 NM_030639.2

SNP Detail

SNP ID Chromosome Location
rs747241738 Chr.X: 102750177-102750177 on Build GRCh38
rs377472532 Chr.X: 102750635-102750635 on Build GRCh38
rs771105705 Chr.X: 102750184-102750184 on Build GRCh38
rs184569966 Chr.X: 102750234-102750234 on Build GRCh38
rs777580372 Chr.X: 102750375-102750375 on Build GRCh38
rs763469511 Chr.X: 102750208-102750208 on Build GRCh38
rs754503542 Chr.X: 102750282-102750282 on Build GRCh38
rs868545592 Chr.X: 102750517-102750517 on Build GRCh38
rs759050072 Chr.X: 102750581-102750581 on Build GRCh38
rs111938109 Chr.X: 102750399-102750399 on Build GRCh38
rs758358614 Chr.X: 102750620-102750620 on Build GRCh38
rs777668450 Chr.X: 102750627-102750627 on Build GRCh38
rs757072249 Chr.X: 102750657-102750657 on Build GRCh38
rs749362154 Chr.X: 102750207-102750207 on Build GRCh38
rs751398086 Chr.X: 102750634-102750634 on Build GRCh38
rs778771568 Chr.X: 102750289-102750289 on Build GRCh38
rs1016631657 Chr.X: 102750313-102750313 on Build GRCh38
rs762235437 Chr.X: 102750520-102750520 on Build GRCh38
rs764804966 Chr.X: 102750594-102750594 on Build GRCh38
rs765705848 Chr.X: 102750191-102750191 on Build GRCh38
rs746760363 Chr.X: 102750381-102750381 on Build GRCh38
rs377700426 Chr.X: 102750326-102750326 on Build GRCh38
rs766011623 Chr.X: 102750553-102750553 on Build GRCh38
rs764567044 Chr.X: 102750219-102750219 on Build GRCh38
rs763559247 Chr.X: 102750470-102750470 on Build GRCh38
rs371291879 Chr.X: 102750385-102750385 on Build GRCh38
rs148746991 Chr.X: 102750438-102750438 on Build GRCh38
rs56344702 Chr.X: 102750227-102750227 on Build GRCh38
rs202050941 Chr.X: 102750308-102750308 on Build GRCh38
rs374677851 Chr.X: 102750421-102750421 on Build GRCh38
rs753513839 Chr.X: 102750563-102750563 on Build GRCh38
rs775658452 Chr.X: 102750443-102750443 on Build GRCh38
rs753327427 Chr.X: 102750280-102750280 on Build GRCh38
rs202018067 Chr.X: 102750190-102750190 on Build GRCh38
rs769236546 Chr.X: 102750491-102750491 on Build GRCh38
rs766031453 Chr.X: 102750266-102750266 on Build GRCh38
rs774904470 Chr.X: 102750505-102750505 on Build GRCh38
rs775654938 Chr.X: 102750462-102750462 on Build GRCh38
rs776599844 Chr.X: 102750186-102750186 on Build GRCh38
rs763988408 Chr.X: 102750498-102750499 on Build GRCh38
rs189497492 Chr.X: 102750255-102750255 on Build GRCh38
rs752167408 Chr.X: 102750600-102750600 on Build GRCh38

View Full Product Details