Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 104190529-104191062 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM199X
Entrez Gene ID
139231
Gene Name
family with sequence similarity 199, X-linked
Gene Aliases
CXorf39
UniGene
Hs.496535
Gene Chromosome Location
Chr.X: 104157074-104195902 on Build GRCh38
Transcripts
NM_207318.3 XM_005262079.2 XM_017029270.1

SNP Detail

SNP ID Chromosome Location
rs375965807 Chr.X: 104190587-104190587 on Build GRCh38
rs34923235 Chr.X: 104190901-104190902 on Build GRCh38
rs916949376 Chr.X: 104190877-104190877 on Build GRCh38
rs782005669 Chr.X: 104190732-104190732 on Build GRCh38
rs996972205 Chr.X: 104191023-104191023 on Build GRCh38
rs782560692 Chr.X: 104191037-104191037 on Build GRCh38
rs782725220 Chr.X: 104190683-104190683 on Build GRCh38
rs1055880252 Chr.X: 104190873-104190873 on Build GRCh38
rs184731658 Chr.X: 104190596-104190596 on Build GRCh38
rs781958394 Chr.X: 104190651-104190651 on Build GRCh38
rs782189324 Chr.X: 104190588-104190588 on Build GRCh38
rs997273576 Chr.X: 104190648-104190648 on Build GRCh38
rs920346598 Chr.X: 104190837-104190837 on Build GRCh38
rs200453477 Chr.X: 104190694-104190695 on Build GRCh38
rs782098006 Chr.X: 104190671-104190671 on Build GRCh38
rs201856884 Chr.X: 104190583-104190583 on Build GRCh38
rs781842532 Chr.X: 104190869-104190869 on Build GRCh38
rs17332064 Chr.X: 104190937-104190937 on Build GRCh38
rs34417303 Chr.X: 104190730-104190731 on Build GRCh38
rs903402423 Chr.X: 104190938-104190938 on Build GRCh38
rs936157237 Chr.X: 104190948-104190948 on Build GRCh38
rs373939792 Chr.X: 104190741-104190741 on Build GRCh38
rs949733428 Chr.X: 104190910-104190910 on Build GRCh38
rs973175169 Chr.X: 104190734-104190734 on Build GRCh38
rs189259259 Chr.X: 104190968-104190968 on Build GRCh38
rs113449164 Chr.X: 104190913-104190913 on Build GRCh38
rs782730314 Chr.X: 104190950-104190950 on Build GRCh38
rs907796725 Chr.X: 104190705-104190705 on Build GRCh38
rs782756155 Chr.X: 104190685-104190685 on Build GRCh38
rs781898880 Chr.X: 104191005-104191005 on Build GRCh38
rs782147712 Chr.X: 104190847-104190847 on Build GRCh38
rs899929033 Chr.X: 104190982-104190982 on Build GRCh38
rs955546295 Chr.X: 104190681-104190681 on Build GRCh38

View Full Product Details