Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 111298406-111298913 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCX
Entrez Gene ID
1641
Gene Name
doublecortin
Gene Aliases
DBCN, DC, LISX, SCLH, XLIS
UniGene
Hs.34780
Gene Chromosome Location
Chr.X: 111293779-111412232 on Build GRCh38
Transcripts
NM_000555.3 NM_001195553.1 NM_178151.2 NM_178152.2 NM_178153.2 XM_011530878.2 XM_011530879.2 XM_011530880.2 XM_017029312.1

SNP Detail

SNP ID Chromosome Location
rs1031881202 Chr.X: 111298874-111298874 on Build GRCh38
rs934822149 Chr.X: 111298464-111298464 on Build GRCh38
rs921678386 Chr.X: 111298820-111298820 on Build GRCh38
rs774114666 Chr.X: 111298741-111298741 on Build GRCh38
rs764951400 Chr.X: 111298875-111298875 on Build GRCh38
rs1027990516 Chr.X: 111298666-111298666 on Build GRCh38
rs895155868 Chr.X: 111298637-111298637 on Build GRCh38
rs772248282 Chr.X: 111298803-111298803 on Build GRCh38
rs1008466633 Chr.X: 111298858-111298858 on Build GRCh38
rs989801126 Chr.X: 111298839-111298839 on Build GRCh38
rs770695600 Chr.X: 111298795-111298795 on Build GRCh38
rs996263545 Chr.X: 111298724-111298724 on Build GRCh38
rs756957490 Chr.X: 111298545-111298545 on Build GRCh38
rs900221691 Chr.X: 111298708-111298708 on Build GRCh38
rs747982479 Chr.X: 111298754-111298754 on Build GRCh38
rs955629325 Chr.X: 111298778-111298778 on Build GRCh38
rs186702259 Chr.X: 111298533-111298533 on Build GRCh38
rs914643276 Chr.X: 111298569-111298569 on Build GRCh38
rs902646225 Chr.X: 111298432-111298432 on Build GRCh38
rs948758055 Chr.X: 111298595-111298595 on Build GRCh38
rs938548561 Chr.X: 111298638-111298638 on Build GRCh38
rs779628574 Chr.X: 111298699-111298699 on Build GRCh38
rs772015393 Chr.X: 111298586-111298586 on Build GRCh38
rs1029914690 Chr.X: 111298757-111298757 on Build GRCh38
rs1012220358 Chr.X: 111298641-111298641 on Build GRCh38
rs749112256 Chr.X: 111298707-111298707 on Build GRCh38
rs755529139 Chr.X: 111298688-111298688 on Build GRCh38
rs185796489 Chr.X: 111298823-111298823 on Build GRCh38
rs1047672876 Chr.X: 111298646-111298646 on Build GRCh38
rs781041715 Chr.X: 111298627-111298627 on Build GRCh38
rs193043715 Chr.X: 111298628-111298628 on Build GRCh38
rs776472621 Chr.X: 111298807-111298807 on Build GRCh38
rs146909118 Chr.X: 111298735-111298735 on Build GRCh38
rs1042568880 Chr.X: 111298884-111298884 on Build GRCh38
rs760911437 Chr.X: 111298882-111298882 on Build GRCh38
rs765456325 Chr.X: 111298467-111298467 on Build GRCh38
rs915483325 Chr.X: 111298860-111298860 on Build GRCh38

View Full Product Details