Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 220698119-220698610 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf115
Entrez Gene ID
79762
Gene Name
chromosome 1 open reading frame 115
Gene Aliases
-
UniGene
Hs.519839
Gene Chromosome Location
Chr.1: 220690286-220699157 on Build GRCh38
Transcripts
NM_024709.4

SNP Detail

SNP ID Chromosome Location
rs750553554 Chr.1: 220698422-220698422 on Build GRCh38
rs991867554 Chr.1: 220698544-220698544 on Build GRCh38
rs937352549 Chr.1: 220698517-220698517 on Build GRCh38
rs903340296 Chr.1: 220698572-220698572 on Build GRCh38
rs1018193941 Chr.1: 220698405-220698405 on Build GRCh38
rs962912431 Chr.1: 220698424-220698424 on Build GRCh38
rs1025407220 Chr.1: 220698452-220698452 on Build GRCh38
rs561461497 Chr.1: 220698548-220698548 on Build GRCh38
rs909505909 Chr.1: 220698328-220698328 on Build GRCh38
rs1011757131 Chr.1: 220698262-220698262 on Build GRCh38
rs1037332813 Chr.1: 220698372-220698372 on Build GRCh38
rs140194598 Chr.1: 220698323-220698323 on Build GRCh38
rs941039219 Chr.1: 220698329-220698329 on Build GRCh38
rs890790392 Chr.1: 220698391-220698391 on Build GRCh38
rs534201644 Chr.1: 220698286-220698286 on Build GRCh38
rs1003167019 Chr.1: 220698573-220698573 on Build GRCh38
rs75023424 Chr.1: 220698496-220698496 on Build GRCh38
rs558919728 Chr.1: 220698162-220698162 on Build GRCh38
rs538001905 Chr.1: 220698209-220698209 on Build GRCh38
rs981477018 Chr.1: 220698469-220698469 on Build GRCh38
rs146117059 Chr.1: 220698246-220698246 on Build GRCh38
rs999259286 Chr.1: 220698377-220698377 on Build GRCh38
rs554082904 Chr.1: 220698298-220698298 on Build GRCh38
rs577094112 Chr.1: 220698311-220698311 on Build GRCh38
rs1056040109 Chr.1: 220698233-220698233 on Build GRCh38
rs144442301 Chr.1: 220698196-220698196 on Build GRCh38
rs186575018 Chr.1: 220698378-220698378 on Build GRCh38
rs747240063 Chr.1: 220698549-220698549 on Build GRCh38
rs576186377 Chr.1: 220698433-220698433 on Build GRCh38
rs902283523 Chr.1: 220698342-220698342 on Build GRCh38
rs367621013 Chr.1: 220698534-220698534 on Build GRCh38
rs927470857 Chr.1: 220698288-220698291 on Build GRCh38
rs889853357 Chr.1: 220698183-220698183 on Build GRCh38
rs184813132 Chr.1: 220698358-220698358 on Build GRCh38
rs1006920224 Chr.1: 220698464-220698464 on Build GRCh38
rs933936943 Chr.1: 220698409-220698409 on Build GRCh38
rs1007258274 Chr.1: 220698403-220698403 on Build GRCh38
rs957425018 Chr.1: 220698221-220698221 on Build GRCh38
rs981595686 Chr.1: 220698282-220698282 on Build GRCh38
rs893084728 Chr.1: 220698484-220698484 on Build GRCh38
rs951487453 Chr.1: 220698468-220698468 on Build GRCh38
rs79758674 Chr.1: 220698232-220698232 on Build GRCh38
rs775522467 Chr.1: 220698530-220698530 on Build GRCh38
rs1041933649 Chr.1: 220698552-220698552 on Build GRCh38
rs539200668 Chr.1: 220698142-220698142 on Build GRCh38

View Full Product Details