Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 226990974-226991479 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDC42BPA
Entrez Gene ID
8476
Gene Name
CDC42 binding protein kinase alpha
Gene Aliases
MRCK, MRCKA, PK428
UniGene
Hs.35433
Gene Chromosome Location
Chr.1: 226989865-227318492 on Build GRCh38
Transcripts
NM_003607.3 NM_014826.4 XM_005273321.3 XM_005273322.3 XM_005273324.3 XM_011544308.2 XM_017002578.1

SNP Detail

SNP ID Chromosome Location
rs754297250 Chr.1: 226991259-226991259 on Build GRCh38
rs930124601 Chr.1: 226991420-226991420 on Build GRCh38
rs941853123 Chr.1: 226991375-226991375 on Build GRCh38
rs985732740 Chr.1: 226991326-226991326 on Build GRCh38
rs59349491 Chr.1: 226991092-226991092 on Build GRCh38
rs925548865 Chr.1: 226991020-226991020 on Build GRCh38
rs954801600 Chr.1: 226991272-226991272 on Build GRCh38
rs1047909700 Chr.1: 226991362-226991362 on Build GRCh38
rs1045287 Chr.1: 226991261-226991261 on Build GRCh38
rs184569762 Chr.1: 226991014-226991014 on Build GRCh38
rs1045289 Chr.1: 226991012-226991012 on Build GRCh38
rs1045285 Chr.1: 226991353-226991353 on Build GRCh38
rs1040187611 Chr.1: 226991075-226991075 on Build GRCh38
rs1047338737 Chr.1: 226991421-226991421 on Build GRCh38
rs950408823 Chr.1: 226991171-226991171 on Build GRCh38
rs1024946634 Chr.1: 226991457-226991457 on Build GRCh38
rs1045290 Chr.1: 226990999-226990999 on Build GRCh38
rs746585347 Chr.1: 226991119-226991119 on Build GRCh38
rs72762309 Chr.1: 226991125-226991125 on Build GRCh38
rs867978715 Chr.1: 226991451-226991451 on Build GRCh38
rs370137574 Chr.1: 226991117-226991117 on Build GRCh38
rs936694802 Chr.1: 226991315-226991315 on Build GRCh38
rs189941160 Chr.1: 226991402-226991402 on Build GRCh38
rs747308120 Chr.1: 226991373-226991373 on Build GRCh38
rs942976323 Chr.1: 226991054-226991054 on Build GRCh38
rs1045286 Chr.1: 226991336-226991336 on Build GRCh38
rs958412926 Chr.1: 226991297-226991297 on Build GRCh38
rs978033372 Chr.1: 226991278-226991278 on Build GRCh38
rs1050402403 Chr.1: 226991023-226991023 on Build GRCh38
rs923909790 Chr.1: 226991015-226991015 on Build GRCh38
rs769448679 Chr.1: 226991390-226991390 on Build GRCh38
rs923836182 Chr.1: 226991292-226991296 on Build GRCh38
rs746461038 Chr.1: 226991042-226991042 on Build GRCh38
rs537006146 Chr.1: 226991393-226991393 on Build GRCh38
rs895049943 Chr.1: 226991412-226991412 on Build GRCh38
rs1047539949 Chr.1: 226991107-226991107 on Build GRCh38
rs566009419 Chr.1: 226991198-226991198 on Build GRCh38
rs771896216 Chr.1: 226991072-226991073 on Build GRCh38
rs2989946 Chr.1: 226991247-226991247 on Build GRCh38
rs1032523515 Chr.1: 226991295-226991295 on Build GRCh38
rs779079429 Chr.1: 226991265-226991265 on Build GRCh38
rs550050829 Chr.1: 226991039-226991039 on Build GRCh38
rs943051205 Chr.1: 226991341-226991341 on Build GRCh38
rs576980963 Chr.1: 226991418-226991418 on Build GRCh38
rs901586792 Chr.1: 226991082-226991082 on Build GRCh38
rs774649797 Chr.1: 226991358-226991358 on Build GRCh38
rs3768431 Chr.1: 226991346-226991346 on Build GRCh38
rs1012865998 Chr.1: 226991091-226991091 on Build GRCh38
rs115381746 Chr.1: 226991435-226991435 on Build GRCh38
rs895187500 Chr.1: 226991156-226991156 on Build GRCh38
rs748688682 Chr.1: 226991198-226991199 on Build GRCh38
rs936956766 Chr.1: 226991022-226991022 on Build GRCh38
rs1053896450 Chr.1: 226991022-226991024 on Build GRCh38

View Full Product Details