Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 228102178-228102692 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf35
Entrez Gene ID
79169
Gene Name
chromosome 1 open reading frame 35
Gene Aliases
MMTAG2
UniGene
Hs.445952
Gene Chromosome Location
Chr.1: 228100727-228103462 on Build GRCh38
Transcripts
NM_024319.3 NR_130975.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4615185 c.273_280delCACGGGCC p.T92fs*86 Chr.1: 228102654-228102661 on Build GRCh38
1295953 c.404G>C p.R135P Chr.1: 228102353-228102353 on Build GRCh38
4028855 c.382G>A p.V128M Chr.1: 228102375-228102375 on Build GRCh38
5983755 c.275C>T p.T92M Chr.1: 228102659-228102659 on Build GRCh38
7358300 c.425A>G p.K142R Chr.1: 228102332-228102332 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369194534 Chr.1: 228102324-228102324 on Build GRCh38
rs781755647 Chr.1: 228102600-228102601 on Build GRCh38
rs947959471 Chr.1: 228102608-228102608 on Build GRCh38
rs773858150 Chr.1: 228102281-228102281 on Build GRCh38
rs867326277 Chr.1: 228102482-228102482 on Build GRCh38
rs768292808 Chr.1: 228102370-228102370 on Build GRCh38
rs777092961 Chr.1: 228102593-228102593 on Build GRCh38
rs1037634609 Chr.1: 228102288-228102288 on Build GRCh38
rs1030007746 Chr.1: 228102305-228102305 on Build GRCh38
rs370772720 Chr.1: 228102638-228102638 on Build GRCh38
rs570632683 Chr.1: 228102491-228102491 on Build GRCh38
rs778552072 Chr.1: 228102375-228102375 on Build GRCh38
rs777713372 Chr.1: 228102261-228102261 on Build GRCh38
rs747640027 Chr.1: 228102459-228102459 on Build GRCh38
rs777130616 Chr.1: 228102290-228102290 on Build GRCh38
rs755939861 Chr.1: 228102430-228102430 on Build GRCh38
rs781000351 Chr.1: 228102339-228102339 on Build GRCh38
rs763126994 Chr.1: 228102321-228102321 on Build GRCh38
rs772251124 Chr.1: 228102630-228102630 on Build GRCh38
rs201723208 Chr.1: 228102598-228102598 on Build GRCh38
rs768239498 Chr.1: 228102279-228102279 on Build GRCh38
rs1011133799 Chr.1: 228102198-228102198 on Build GRCh38
rs376871646 Chr.1: 228102413-228102413 on Build GRCh38
rs776662344 Chr.1: 228102636-228102636 on Build GRCh38
rs867187913 Chr.1: 228102521-228102521 on Build GRCh38
rs757053740 Chr.1: 228102265-228102265 on Build GRCh38
rs942727658 Chr.1: 228102354-228102354 on Build GRCh38
rs765301504 Chr.1: 228102204-228102204 on Build GRCh38
rs923204543 Chr.1: 228102417-228102417 on Build GRCh38
rs750285905 Chr.1: 228102423-228102423 on Build GRCh38
rs61825155 Chr.1: 228102332-228102332 on Build GRCh38
rs757934128 Chr.1: 228102466-228102466 on Build GRCh38
rs550471447 Chr.1: 228102471-228102471 on Build GRCh38
rs750207588 Chr.1: 228102344-228102344 on Build GRCh38
rs754125957 Chr.1: 228102199-228102199 on Build GRCh38
rs753286442 Chr.1: 228102199-228102200 on Build GRCh38
rs747390709 Chr.1: 228102516-228102518 on Build GRCh38
rs779654391 Chr.1: 228102439-228102439 on Build GRCh38
rs566468092 Chr.1: 228102571-228102571 on Build GRCh38
rs377443672 Chr.1: 228102522-228102522 on Build GRCh38
rs955546434 Chr.1: 228102258-228102258 on Build GRCh38
rs763032189 Chr.1: 228102416-228102416 on Build GRCh38
rs769453961 Chr.1: 228102269-228102269 on Build GRCh38
rs555368495 Chr.1: 228102624-228102624 on Build GRCh38
rs746296593 Chr.1: 228102484-228102484 on Build GRCh38
rs540925037 Chr.1: 228102232-228102232 on Build GRCh38
rs892140484 Chr.1: 228102412-228102412 on Build GRCh38
rs748160830 Chr.1: 228102610-228102620 on Build GRCh38
rs754673815 Chr.1: 228102443-228102443 on Build GRCh38
rs755813107 Chr.1: 228102349-228102349 on Build GRCh38
rs975448763 Chr.1: 228102233-228102233 on Build GRCh38
rs759969526 Chr.1: 228102401-228102401 on Build GRCh38
rs768840354 Chr.1: 228102518-228102518 on Build GRCh38
rs778607073 Chr.1: 228102456-228102456 on Build GRCh38
rs964177292 Chr.1: 228102432-228102432 on Build GRCh38
rs752413496 Chr.1: 228102659-228102659 on Build GRCh38
rs760706045 Chr.1: 228102632-228102632 on Build GRCh38
rs765004577 Chr.1: 228102585-228102585 on Build GRCh38
rs780513010 Chr.1: 228102601-228102601 on Build GRCh38
rs764220937 Chr.1: 228102419-228102419 on Build GRCh38
rs746448908 Chr.1: 228102597-228102597 on Build GRCh38
rs947909071 Chr.1: 228102301-228102301 on Build GRCh38
rs578158660 Chr.1: 228102223-228102223 on Build GRCh38
rs758317136 Chr.1: 228102215-228102215 on Build GRCh38
rs982769714 Chr.1: 228102262-228102262 on Build GRCh38
rs1039400723 Chr.1: 228102218-228102218 on Build GRCh38
rs955445962 Chr.1: 228102513-228102513 on Build GRCh38
rs530647448 Chr.1: 228102426-228102426 on Build GRCh38
rs754727266 Chr.1: 228102580-228102580 on Build GRCh38
rs773212690 Chr.1: 228102539-228102539 on Build GRCh38
rs558022354 Chr.1: 228102640-228102640 on Build GRCh38
rs771534451 Chr.1: 228102398-228102398 on Build GRCh38
rs749671377 Chr.1: 228102604-228102604 on Build GRCh38
rs779475958 Chr.1: 228102271-228102271 on Build GRCh38
rs528084635 Chr.1: 228102295-228102295 on Build GRCh38
rs746911412 Chr.1: 228102264-228102264 on Build GRCh38
rs145743393 Chr.1: 228102350-228102350 on Build GRCh38
rs757989264 Chr.1: 228102589-228102589 on Build GRCh38
rs774504299 Chr.1: 228102520-228102520 on Build GRCh38
rs766262947 Chr.1: 228102567-228102567 on Build GRCh38
rs752792503 Chr.1: 228102207-228102207 on Build GRCh38
rs755468078 Chr.1: 228102538-228102539 on Build GRCh38
rs758155115 Chr.1: 228102285-228102292 on Build GRCh38
rs756850969 Chr.1: 228102204-228102205 on Build GRCh38
rs997002286 Chr.1: 228102473-228102473 on Build GRCh38
rs755215152 Chr.1: 228102203-228102203 on Build GRCh38
rs371201234 Chr.1: 228102555-228102555 on Build GRCh38
rs373623617 Chr.1: 228102586-228102586 on Build GRCh38
rs867593559 Chr.1: 228102538-228102538 on Build GRCh38
rs901749116 Chr.1: 228102219-228102219 on Build GRCh38
rs368673303 Chr.1: 228102500-228102500 on Build GRCh38
rs762065505 Chr.1: 228102270-228102271 on Build GRCh38
rs780516497 Chr.1: 228102317-228102320 on Build GRCh38
rs747586817 Chr.1: 228102393-228102393 on Build GRCh38
rs770016363 Chr.1: 228102404-228102404 on Build GRCh38
rs767426726 Chr.1: 228102530-228102530 on Build GRCh38
rs920409852 Chr.1: 228102292-228102292 on Build GRCh38
rs1011907889 Chr.1: 228102272-228102272 on Build GRCh38
rs751604659 Chr.1: 228102420-228102420 on Build GRCh38
rs902280027 Chr.1: 228102248-228102248 on Build GRCh38
rs906248662 Chr.1: 228102328-228102328 on Build GRCh38
rs768965232 Chr.1: 228102611-228102611 on Build GRCh38
rs2295995 Chr.1: 228102275-228102275 on Build GRCh38
rs922747896 Chr.1: 228102255-228102255 on Build GRCh38
rs1042253104 Chr.1: 228102431-228102431 on Build GRCh38
rs1000703488 Chr.1: 228102474-228102474 on Build GRCh38
rs752814685 Chr.1: 228102304-228102304 on Build GRCh38
rs753584462 Chr.1: 228102440-228102440 on Build GRCh38
rs1012348704 Chr.1: 228102579-228102579 on Build GRCh38
rs769962246 Chr.1: 228102621-228102622 on Build GRCh38
rs184854491 Chr.1: 228102512-228102512 on Build GRCh38
rs780910425 Chr.1: 228102266-228102266 on Build GRCh38
rs765476215 Chr.1: 228102298-228102298 on Build GRCh38
rs761205135 Chr.1: 228102282-228102282 on Build GRCh38
rs771479442 Chr.1: 228102286-228102286 on Build GRCh38
rs984295845 Chr.1: 228102619-228102619 on Build GRCh38
rs899673663 Chr.1: 228102213-228102213 on Build GRCh38
rs200603347 Chr.1: 228102634-228102634 on Build GRCh38
rs373950653 Chr.1: 228102353-228102353 on Build GRCh38
rs764775944 Chr.1: 228102260-228102261 on Build GRCh38
rs774557625 Chr.1: 228102623-228102623 on Build GRCh38

View Full Product Details