Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 14899370-14899884 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCLRE1C
Entrez Gene ID
64421
Gene Name
DNA cross-link repair 1C
Gene Aliases
A-SCID, DCLREC1C, RS-SCID, SCIDA, SNM1C
UniGene
Hs.655932
Gene Chromosome Location
Chr.10: 14897853-14954432 on Build GRCh38
Transcripts
XM_017016556.1 XM_017016559.1 XM_017016560.1 NM_001193424.1 NM_001193425.1 NM_001193426.1 NM_001193427.1 NM_024670.3 NR_034181.1 XM_006717503.3 XM_011519662.2 XM_017016637.1

Gene Symbol
SUV39H2
Entrez Gene ID
79723
Gene Name
suppressor of variegation 3-9 homolog 2
Gene Aliases
KMT1B
UniGene
Hs.554883
Gene Chromosome Location
Chr.10: 14878783-14904315 on Build GRCh38
Transcripts
XM_017016556.1 XM_017016559.1 XM_017016560.1 NM_001193424.1 NM_001193425.1 NM_001193426.1 NM_001193427.1 NM_024670.3 NR_034181.1 XM_006717503.3 XM_011519662.2 XM_017016637.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1347020 c.956C>T p.A319V Chr.10: 14899645-14899645 on Build GRCh38
4449716 c.670-1G>A p.? Chr.10: 14899538-14899538 on Build GRCh38
5050058 c.909G>A p.T303T Chr.10: 14899598-14899598 on Build GRCh38
5050060 c.919G>A p.D307N Chr.10: 14899608-14899608 on Build GRCh38
7191387 c.868G>T p.A290S Chr.10: 14899557-14899557 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374384196 Chr.10: 14899715-14899715 on Build GRCh38
rs772448390 Chr.10: 14899700-14899700 on Build GRCh38
rs183140822 Chr.10: 14899610-14899610 on Build GRCh38
rs375402354 Chr.10: 14899736-14899736 on Build GRCh38
rs17353856 Chr.10: 14899655-14899655 on Build GRCh38
rs188371957 Chr.10: 14899800-14899800 on Build GRCh38
rs371193897 Chr.10: 14899466-14899466 on Build GRCh38
rs765430780 Chr.10: 14899613-14899613 on Build GRCh38
rs147233152 Chr.10: 14899734-14899734 on Build GRCh38
rs369186882 Chr.10: 14899661-14899661 on Build GRCh38
rs1017597310 Chr.10: 14899793-14899793 on Build GRCh38
rs147278353 Chr.10: 14899676-14899676 on Build GRCh38
rs771360187 Chr.10: 14899721-14899721 on Build GRCh38
rs1009576660 Chr.10: 14899757-14899757 on Build GRCh38
rs201406856 Chr.10: 14899699-14899699 on Build GRCh38
rs1056233424 Chr.10: 14899695-14899695 on Build GRCh38
rs1006084833 Chr.10: 14899775-14899775 on Build GRCh38
rs752873117 Chr.10: 14899624-14899624 on Build GRCh38
rs761163033 Chr.10: 14899579-14899579 on Build GRCh38
rs543776803 Chr.10: 14899566-14899566 on Build GRCh38
rs776944452 Chr.10: 14899723-14899723 on Build GRCh38
rs745322597 Chr.10: 14899522-14899522 on Build GRCh38
rs1029271379 Chr.10: 14899848-14899848 on Build GRCh38
rs150887083 Chr.10: 14899456-14899456 on Build GRCh38
rs185751453 Chr.10: 14899470-14899470 on Build GRCh38
rs769296049 Chr.10: 14899530-14899530 on Build GRCh38
rs979002845 Chr.10: 14899428-14899429 on Build GRCh38
rs770484753 Chr.10: 14899510-14899510 on Build GRCh38
rs34659846 Chr.10: 14899699-14899700 on Build GRCh38
rs148667087 Chr.10: 14899611-14899611 on Build GRCh38
rs373066474 Chr.10: 14899509-14899509 on Build GRCh38
rs769240981 Chr.10: 14899693-14899693 on Build GRCh38
rs911580226 Chr.10: 14899529-14899529 on Build GRCh38
rs747364188 Chr.10: 14899822-14899822 on Build GRCh38
rs777408336 Chr.10: 14899503-14899503 on Build GRCh38
rs773785311 Chr.10: 14899714-14899714 on Build GRCh38
rs776401267 Chr.10: 14899521-14899521 on Build GRCh38
rs780583220 Chr.10: 14899497-14899498 on Build GRCh38
rs768113481 Chr.10: 14899568-14899568 on Build GRCh38
rs138209139 Chr.10: 14899488-14899488 on Build GRCh38
rs773447856 Chr.10: 14899571-14899571 on Build GRCh38
rs541394387 Chr.10: 14899745-14899745 on Build GRCh38
rs758378629 Chr.10: 14899637-14899637 on Build GRCh38
rs190174096 Chr.10: 14899598-14899598 on Build GRCh38
rs745546710 Chr.10: 14899685-14899685 on Build GRCh38
rs1030436677 Chr.10: 14899826-14899826 on Build GRCh38
rs550100723 Chr.10: 14899835-14899835 on Build GRCh38
rs112796645 Chr.10: 14899533-14899533 on Build GRCh38
rs372658702 Chr.10: 14899809-14899809 on Build GRCh38
rs777832782 Chr.10: 14899646-14899646 on Build GRCh38
rs34488711 Chr.10: 14899714-14899715 on Build GRCh38
rs141509653 Chr.10: 14899424-14899424 on Build GRCh38
rs757086288 Chr.10: 14899660-14899660 on Build GRCh38
rs546943726 Chr.10: 14899468-14899470 on Build GRCh38
rs969077561 Chr.10: 14899802-14899802 on Build GRCh38
rs769125245 Chr.10: 14899696-14899697 on Build GRCh38
rs987167769 Chr.10: 14899515-14899515 on Build GRCh38
rs35032339 Chr.10: 14899698-14899699 on Build GRCh38
rs749739907 Chr.10: 14899582-14899584 on Build GRCh38
rs748128649 Chr.10: 14899491-14899491 on Build GRCh38
rs758218478 Chr.10: 14899498-14899498 on Build GRCh38

View Full Product Details