Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 43155207-43155717 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSGALNACT2
Entrez Gene ID
55454
Gene Name
chondroitin sulfate N-acetylgalactosaminyltransferase 2
Gene Aliases
CHGN2, ChGn-2, GALNACT-2, GALNACT2, PRO0082, beta4GalNAcT
UniGene
Hs.657569
Gene Chromosome Location
Chr.10: 43138445-43185308 on Build GRCh38
Transcripts
NM_001319654.1 NM_001319656.1 NM_018590.4 NR_135077.1 XM_011539938.1 XM_017016388.1 XM_017016389.1 XM_017016390.1 XM_017016391.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
363612 c.101G>T p.C34F Chr.10: 43155250-43155250 on Build GRCh38
6066196 c.183A>T p.L61L Chr.10: 43155332-43155332 on Build GRCh38
7178401 c.338A>C p.K113T Chr.10: 43155487-43155487 on Build GRCh38
6129692 c.478G>T p.E160* Chr.10: 43155627-43155627 on Build GRCh38
3670534 c.67T>A p.C23S Chr.10: 43155216-43155216 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs549097243 Chr.10: 43155249-43155249 on Build GRCh38
rs758213307 Chr.10: 43155509-43155509 on Build GRCh38
rs751349856 Chr.10: 43155585-43155585 on Build GRCh38
rs763376840 Chr.10: 43155378-43155378 on Build GRCh38
rs759334130 Chr.10: 43155588-43155588 on Build GRCh38
rs770410222 Chr.10: 43155363-43155363 on Build GRCh38
rs753345504 Chr.10: 43155409-43155409 on Build GRCh38
rs567494904 Chr.10: 43155279-43155279 on Build GRCh38
rs755203690 Chr.10: 43155533-43155533 on Build GRCh38
rs772362661 Chr.10: 43155679-43155679 on Build GRCh38
rs772770109 Chr.10: 43155581-43155581 on Build GRCh38
rs895740907 Chr.10: 43155339-43155339 on Build GRCh38
rs774640408 Chr.10: 43155564-43155564 on Build GRCh38
rs757053801 Chr.10: 43155330-43155330 on Build GRCh38
rs1035454945 Chr.10: 43155316-43155316 on Build GRCh38
rs756215710 Chr.10: 43155556-43155556 on Build GRCh38
rs41298781 Chr.10: 43155467-43155467 on Build GRCh38
rs372035294 Chr.10: 43155269-43155269 on Build GRCh38
rs542751915 Chr.10: 43155417-43155417 on Build GRCh38
rs370010974 Chr.10: 43155608-43155608 on Build GRCh38
rs755223401 Chr.10: 43155432-43155432 on Build GRCh38
rs368933539 Chr.10: 43155484-43155484 on Build GRCh38
rs745652732 Chr.10: 43155341-43155341 on Build GRCh38
rs762476511 Chr.10: 43155361-43155361 on Build GRCh38
rs766082270 Chr.10: 43155340-43155342 on Build GRCh38
rs767875885 Chr.10: 43155601-43155601 on Build GRCh38
rs577874758 Chr.10: 43155314-43155314 on Build GRCh38
rs893250141 Chr.10: 43155653-43155653 on Build GRCh38
rs749692632 Chr.10: 43155562-43155562 on Build GRCh38
rs200632295 Chr.10: 43155582-43155582 on Build GRCh38
rs773759812 Chr.10: 43155373-43155373 on Build GRCh38
rs780096763 Chr.10: 43155343-43155343 on Build GRCh38
rs765139917 Chr.10: 43155418-43155418 on Build GRCh38
rs557258013 Chr.10: 43155551-43155551 on Build GRCh38
rs778750683 Chr.10: 43155333-43155333 on Build GRCh38
rs759317317 Chr.10: 43155294-43155294 on Build GRCh38
rs937053844 Chr.10: 43155656-43155656 on Build GRCh38
rs768557337 Chr.10: 43155351-43155351 on Build GRCh38
rs907082267 Chr.10: 43155622-43155622 on Build GRCh38
rs776941721 Chr.10: 43155690-43155690 on Build GRCh38
rs778439473 Chr.10: 43155560-43155560 on Build GRCh38
rs201853037 Chr.10: 43155643-43155643 on Build GRCh38
rs1001338798 Chr.10: 43155315-43155315 on Build GRCh38
rs753717102 Chr.10: 43155328-43155328 on Build GRCh38
rs1043224506 Chr.10: 43155302-43155302 on Build GRCh38
rs747131975 Chr.10: 43155345-43155345 on Build GRCh38
rs769224623 Chr.10: 43155579-43155579 on Build GRCh38
rs374031130 Chr.10: 43155565-43155565 on Build GRCh38
rs781479052 Chr.10: 43155444-43155444 on Build GRCh38
rs755541801 Chr.10: 43155309-43155309 on Build GRCh38
rs758077647 Chr.10: 43155428-43155428 on Build GRCh38
rs771143238 Chr.10: 43155466-43155466 on Build GRCh38
rs772390120 Chr.10: 43155342-43155342 on Build GRCh38
rs374840357 Chr.10: 43155449-43155449 on Build GRCh38
rs779446866 Chr.10: 43155672-43155672 on Build GRCh38
rs144936502 Chr.10: 43155459-43155459 on Build GRCh38
rs200474280 Chr.10: 43155538-43155538 on Build GRCh38
rs781135524 Chr.10: 43155491-43155491 on Build GRCh38
rs781532114 Chr.10: 43155535-43155535 on Build GRCh38
rs748214670 Chr.10: 43155448-43155448 on Build GRCh38
rs772516385 Chr.10: 43155298-43155298 on Build GRCh38
rs41298783 Chr.10: 43155494-43155494 on Build GRCh38
rs771795425 Chr.10: 43155323-43155323 on Build GRCh38
rs750261794 Chr.10: 43155425-43155425 on Build GRCh38
rs746251791 Chr.10: 43155678-43155678 on Build GRCh38
rs370782457 Chr.10: 43155651-43155651 on Build GRCh38
rs150062393 Chr.10: 43155392-43155392 on Build GRCh38
rs199746535 Chr.10: 43155288-43155288 on Build GRCh38
rs773817131 Chr.10: 43155458-43155458 on Build GRCh38
rs776228932 Chr.10: 43155571-43155571 on Build GRCh38
rs201378409 Chr.10: 43155289-43155289 on Build GRCh38
rs915731392 Chr.10: 43155492-43155492 on Build GRCh38
rs150048164 Chr.10: 43155430-43155430 on Build GRCh38
rs200443486 Chr.10: 43155353-43155353 on Build GRCh38
rs774957310 Chr.10: 43155383-43155383 on Build GRCh38
rs753722603 Chr.10: 43155577-43155578 on Build GRCh38
rs763445891 Chr.10: 43155402-43155402 on Build GRCh38
rs149042915 Chr.10: 43155496-43155496 on Build GRCh38
rs752910250 Chr.10: 43155605-43155605 on Build GRCh38
rs375066016 Chr.10: 43155301-43155301 on Build GRCh38
rs778065650 Chr.10: 43155623-43155623 on Build GRCh38
rs771464642 Chr.10: 43155563-43155563 on Build GRCh38
rs375255101 Chr.10: 43155663-43155663 on Build GRCh38
rs763792861 Chr.10: 43155480-43155480 on Build GRCh38
rs761324012 Chr.10: 43155572-43155572 on Build GRCh38
rs776953921 Chr.10: 43155273-43155273 on Build GRCh38
rs267602488 Chr.10: 43155266-43155266 on Build GRCh38
rs769235430 Chr.10: 43155268-43155268 on Build GRCh38
rs944230917 Chr.10: 43155263-43155263 on Build GRCh38
rs771503726 Chr.10: 43155642-43155642 on Build GRCh38
rs760993595 Chr.10: 43155314-43155315 on Build GRCh38
rs199602683 Chr.10: 43155379-43155379 on Build GRCh38
rs766149579 Chr.10: 43155526-43155526 on Build GRCh38
rs947061762 Chr.10: 43155290-43155290 on Build GRCh38
rs989093066 Chr.10: 43155506-43155506 on Build GRCh38
rs368373201 Chr.10: 43155429-43155429 on Build GRCh38
rs751355565 Chr.10: 43155532-43155532 on Build GRCh38
rs202041692 Chr.10: 43155501-43155501 on Build GRCh38
rs760315975 Chr.10: 43155475-43155475 on Build GRCh38

View Full Product Details