Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 72011403-72011902 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHST3
Entrez Gene ID
9469
Gene Name
carbohydrate sulfotransferase 3
Gene Aliases
C6ST, C6ST1, HSD
UniGene
Hs.158304
Gene Chromosome Location
Chr.10: 71964362-72013564 on Build GRCh38
Transcripts
NM_004273.4 XM_006718075.3 XM_011540369.2

SNP Detail

SNP ID Chromosome Location
rs529402579 Chr.10: 72011558-72011558 on Build GRCh38
rs982011982 Chr.10: 72011482-72011482 on Build GRCh38
rs548082422 Chr.10: 72011829-72011829 on Build GRCh38
rs542056241 Chr.10: 72011427-72011427 on Build GRCh38
rs76750860 Chr.10: 72011561-72011561 on Build GRCh38
rs550027128 Chr.10: 72011511-72011511 on Build GRCh38
rs964636198 Chr.10: 72011822-72011822 on Build GRCh38
rs75962504 Chr.10: 72011772-72011772 on Build GRCh38
rs545967470 Chr.10: 72011716-72011716 on Build GRCh38
rs539580815 Chr.10: 72011743-72011743 on Build GRCh38
rs868390331 Chr.10: 72011575-72011575 on Build GRCh38
rs145507314 Chr.10: 72011493-72011493 on Build GRCh38
rs573559460 Chr.10: 72011445-72011445 on Build GRCh38
rs564572994 Chr.10: 72011862-72011862 on Build GRCh38
rs1018806913 Chr.10: 72011811-72011811 on Build GRCh38
rs974091740 Chr.10: 72011498-72011498 on Build GRCh38
rs943926044 Chr.10: 72011546-72011546 on Build GRCh38
rs911831901 Chr.10: 72011569-72011569 on Build GRCh38
rs895434732 Chr.10: 72011715-72011715 on Build GRCh38
rs902658685 Chr.10: 72011615-72011615 on Build GRCh38
rs1044689463 Chr.10: 72011699-72011699 on Build GRCh38
rs12172746 Chr.10: 72011524-72011524 on Build GRCh38
rs1035291799 Chr.10: 72011492-72011492 on Build GRCh38
rs1042195597 Chr.10: 72011559-72011559 on Build GRCh38
rs193204538 Chr.10: 72011838-72011838 on Build GRCh38
rs12250549 Chr.10: 72011777-72011777 on Build GRCh38
rs762284234 Chr.10: 72011728-72011728 on Build GRCh38
rs577353046 Chr.10: 72011602-72011602 on Build GRCh38
rs886047180 Chr.10: 72011855-72011855 on Build GRCh38
rs902691502 Chr.10: 72011454-72011454 on Build GRCh38
rs764442442 Chr.10: 72011630-72011630 on Build GRCh38
rs1039042493 Chr.10: 72011583-72011583 on Build GRCh38
rs886047179 Chr.10: 72011638-72011638 on Build GRCh38
rs950620753 Chr.10: 72011553-72011553 on Build GRCh38
rs1042524555 Chr.10: 72011437-72011437 on Build GRCh38
rs912387007 Chr.10: 72011527-72011527 on Build GRCh38
rs1022589622 Chr.10: 72011727-72011727 on Build GRCh38
rs1003738314 Chr.10: 72011626-72011626 on Build GRCh38
rs148431635 Chr.10: 72011763-72011763 on Build GRCh38
rs572091260 Chr.10: 72011490-72011490 on Build GRCh38
rs566156519 Chr.10: 72011641-72011641 on Build GRCh38
rs751765701 Chr.10: 72011649-72011649 on Build GRCh38
rs1013596149 Chr.10: 72011640-72011640 on Build GRCh38
rs915232825 Chr.10: 72011590-72011590 on Build GRCh38
rs74145519 Chr.10: 72011510-72011510 on Build GRCh38
rs560145890 Chr.10: 72011428-72011428 on Build GRCh38
rs769807215 Chr.10: 72011535-72011535 on Build GRCh38
rs556456859 Chr.10: 72011878-72011878 on Build GRCh38
rs763081087 Chr.10: 72011604-72011604 on Build GRCh38
rs563954537 Chr.10: 72011496-72011496 on Build GRCh38
rs963381667 Chr.10: 72011733-72011733 on Build GRCh38
rs932573838 Chr.10: 72011522-72011522 on Build GRCh38
rs187587634 Chr.10: 72011600-72011600 on Build GRCh38

View Full Product Details