Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 73234679-73235193 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM149B1
Entrez Gene ID
317662
Gene Name
family with sequence similarity 149 member B1
Gene Aliases
KIAA0974
UniGene
Hs.408577
Gene Chromosome Location
Chr.10: 73168090-73242181 on Build GRCh38
Transcripts
NM_173348.1 XM_005269744.2 XM_005269745.2 XM_005269747.3 XM_011539737.2 XM_017016164.1 XM_017016165.1 XM_017016166.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6272652 c.1366G>T p.D456Y Chr.10: 73234830-73234830 on Build GRCh38
7426501 c.1370C>T p.S457L Chr.10: 73234834-73234834 on Build GRCh38
5012173 c.1459G>A p.G487R Chr.10: 73234923-73234923 on Build GRCh38
6518008 c.1420C>A p.P474T Chr.10: 73234884-73234884 on Build GRCh38
6260482 c.1427G>T p.G476V Chr.10: 73234891-73234891 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1033785277 Chr.10: 73234830-73234830 on Build GRCh38
rs774250509 Chr.10: 73234904-73234904 on Build GRCh38
rs1015543742 Chr.10: 73235128-73235128 on Build GRCh38
rs753061386 Chr.10: 73234873-73234873 on Build GRCh38
rs529722258 Chr.10: 73235111-73235111 on Build GRCh38
rs573526295 Chr.10: 73234937-73234937 on Build GRCh38
rs11814139 Chr.10: 73235129-73235129 on Build GRCh38
rs772279017 Chr.10: 73235107-73235107 on Build GRCh38
rs746797586 Chr.10: 73235055-73235056 on Build GRCh38
rs11819288 Chr.10: 73234860-73234860 on Build GRCh38
rs1034527860 Chr.10: 73234747-73234747 on Build GRCh38
rs1003429334 Chr.10: 73234739-73234739 on Build GRCh38
rs907409958 Chr.10: 73234934-73234934 on Build GRCh38
rs1042629481 Chr.10: 73234990-73234990 on Build GRCh38
rs140932169 Chr.10: 73235121-73235121 on Build GRCh38
rs1003135479 Chr.10: 73234940-73234940 on Build GRCh38
rs972448583 Chr.10: 73235019-73235019 on Build GRCh38
rs759716392 Chr.10: 73235153-73235153 on Build GRCh38
rs746770486 Chr.10: 73235160-73235160 on Build GRCh38
rs971688342 Chr.10: 73234717-73234717 on Build GRCh38
rs771002500 Chr.10: 73234860-73234861 on Build GRCh38
rs1009522816 Chr.10: 73234770-73234770 on Build GRCh38
rs768535982 Chr.10: 73234813-73234813 on Build GRCh38
rs776858653 Chr.10: 73235097-73235097 on Build GRCh38
rs892793497 Chr.10: 73234887-73234887 on Build GRCh38
rs941024653 Chr.10: 73234866-73234866 on Build GRCh38
rs182236885 Chr.10: 73234933-73234933 on Build GRCh38
rs566576470 Chr.10: 73235054-73235054 on Build GRCh38
rs1051773279 Chr.10: 73234722-73234722 on Build GRCh38
rs11819307 Chr.10: 73234985-73234985 on Build GRCh38
rs909209417 Chr.10: 73234821-73234821 on Build GRCh38
rs958765363 Chr.10: 73234831-73234831 on Build GRCh38
rs745995251 Chr.10: 73235073-73235073 on Build GRCh38
rs968027354 Chr.10: 73234798-73234798 on Build GRCh38
rs191145472 Chr.10: 73234856-73234856 on Build GRCh38
rs756720458 Chr.10: 73234905-73234905 on Build GRCh38
rs543277642 Chr.10: 73234910-73234910 on Build GRCh38
rs531961915 Chr.10: 73234984-73234984 on Build GRCh38
rs763027521 Chr.10: 73234832-73234832 on Build GRCh38
rs895264775 Chr.10: 73235056-73235056 on Build GRCh38
rs2241968 Chr.10: 73234755-73234755 on Build GRCh38
rs191041721 Chr.10: 73234986-73234986 on Build GRCh38
rs796588524 Chr.10: 73234881-73234881 on Build GRCh38
rs540559624 Chr.10: 73234973-73234973 on Build GRCh38
rs879208223 Chr.10: 73235122-73235122 on Build GRCh38
rs903838392 Chr.10: 73235004-73235004 on Build GRCh38
rs923297265 Chr.10: 73235031-73235031 on Build GRCh38
rs921053690 Chr.10: 73234916-73234916 on Build GRCh38
rs562279953 Chr.10: 73235058-73235058 on Build GRCh38
rs778281633 Chr.10: 73234829-73234829 on Build GRCh38
rs112273035 Chr.10: 73234836-73234836 on Build GRCh38
rs766353650 Chr.10: 73234852-73234852 on Build GRCh38
rs539543745 Chr.10: 73234794-73234794 on Build GRCh38
rs749780762 Chr.10: 73234855-73234855 on Build GRCh38
rs932806638 Chr.10: 73234954-73234954 on Build GRCh38
rs61742710 Chr.10: 73234862-73234862 on Build GRCh38
rs756850934 Chr.10: 73234966-73234966 on Build GRCh38
rs771223827 Chr.10: 73235075-73235075 on Build GRCh38
rs1000896327 Chr.10: 73235037-73235037 on Build GRCh38
rs991366825 Chr.10: 73234834-73234834 on Build GRCh38
rs185843840 Chr.10: 73234976-73234976 on Build GRCh38
rs374684689 Chr.10: 73234853-73234853 on Build GRCh38
rs867530426 Chr.10: 73235117-73235117 on Build GRCh38
rs942567906 Chr.10: 73235119-73235119 on Build GRCh38
rs984906713 Chr.10: 73234810-73234810 on Build GRCh38
rs1013161764 Chr.10: 73235074-73235074 on Build GRCh38
rs774701845 Chr.10: 73234867-73234867 on Build GRCh38

View Full Product Details