Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 74314358-74314887 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADK
Entrez Gene ID
132
Gene Name
adenosine kinase
Gene Aliases
AK
UniGene
Hs.656586
Gene Chromosome Location
Chr.10: 74151185-74709303 on Build GRCh38
Transcripts
NM_001123.3 NM_001202449.1 NM_001202450.1 NM_006721.3 XM_017015698.1 XM_017015700.1 XM_017015701.1 XM_017015702.1 XM_017015704.1 XM_017015705.1 XM_017015706.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4775515 c.271C>T p.Q91* Chr.10: 74314743-74314743 on Build GRCh38
5420419 c.211A>G p.K71E Chr.10: 74314683-74314683 on Build GRCh38
5371123 c.197T>C p.F66S Chr.10: 74314669-74314669 on Build GRCh38
242857 c.261T>G p.I87M Chr.10: 74314733-74314733 on Build GRCh38
1349112 c.226G>A p.E76K Chr.10: 74314698-74314698 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755324323 Chr.10: 74314729-74314729 on Build GRCh38
rs958133477 Chr.10: 74314456-74314456 on Build GRCh38
rs187598971 Chr.10: 74314573-74314573 on Build GRCh38
rs941122285 Chr.10: 74314596-74314596 on Build GRCh38
rs1001811211 Chr.10: 74314589-74314589 on Build GRCh38
rs756956429 Chr.10: 74314650-74314650 on Build GRCh38
rs144492760 Chr.10: 74314649-74314649 on Build GRCh38
rs892715717 Chr.10: 74314519-74314519 on Build GRCh38
rs942781825 Chr.10: 74314464-74314464 on Build GRCh38
rs546729163 Chr.10: 74314751-74314751 on Build GRCh38
rs761033679 Chr.10: 74314632-74314632 on Build GRCh38
rs759919169 Chr.10: 74314621-74314621 on Build GRCh38
rs909855808 Chr.10: 74314423-74314423 on Build GRCh38
rs201219225 Chr.10: 74314769-74314769 on Build GRCh38
rs928195106 Chr.10: 74314553-74314556 on Build GRCh38
rs752349730 Chr.10: 74314524-74314524 on Build GRCh38
rs779372765 Chr.10: 74314750-74314751 on Build GRCh38
rs780042061 Chr.10: 74314697-74314697 on Build GRCh38
rs140410201 Chr.10: 74314407-74314407 on Build GRCh38
rs763692124 Chr.10: 74314472-74314472 on Build GRCh38
rs975405319 Chr.10: 74314818-74314818 on Build GRCh38
rs575638744 Chr.10: 74314490-74314490 on Build GRCh38
rs759057753 Chr.10: 74314388-74314391 on Build GRCh38
rs966696204 Chr.10: 74314478-74314478 on Build GRCh38
rs747006315 Chr.10: 74314722-74314722 on Build GRCh38
rs533459835 Chr.10: 74314780-74314780 on Build GRCh38
rs779328407 Chr.10: 74314755-74314755 on Build GRCh38
rs201734294 Chr.10: 74314731-74314731 on Build GRCh38
rs556278270 Chr.10: 74314473-74314473 on Build GRCh38
rs967148036 Chr.10: 74314807-74314807 on Build GRCh38
rs959557425 Chr.10: 74314560-74314560 on Build GRCh38
rs752945358 Chr.10: 74314630-74314630 on Build GRCh38
rs142725894 Chr.10: 74314417-74314417 on Build GRCh38
rs1034765439 Chr.10: 74314591-74314591 on Build GRCh38
rs922724017 Chr.10: 74314854-74314854 on Build GRCh38
rs748691633 Chr.10: 74314792-74314792 on Build GRCh38
rs955447789 Chr.10: 74314861-74314861 on Build GRCh38
rs776060880 Chr.10: 74314796-74314796 on Build GRCh38
rs1017665892 Chr.10: 74314442-74314442 on Build GRCh38
rs561133949 Chr.10: 74314626-74314626 on Build GRCh38
rs369308554 Chr.10: 74314784-74314784 on Build GRCh38
rs375709878 Chr.10: 74314648-74314648 on Build GRCh38
rs975519450 Chr.10: 74314468-74314468 on Build GRCh38
rs367966707 Chr.10: 74314579-74314579 on Build GRCh38
rs191307870 Chr.10: 74314802-74314802 on Build GRCh38
rs1006663678 Chr.10: 74314431-74314431 on Build GRCh38
rs372669372 Chr.10: 74314694-74314694 on Build GRCh38
rs778258589 Chr.10: 74314687-74314687 on Build GRCh38
rs372679823 Chr.10: 74314768-74314768 on Build GRCh38
rs1036751579 Chr.10: 74314620-74314620 on Build GRCh38
rs201786575 Chr.10: 74314692-74314692 on Build GRCh38
rs946958620 Chr.10: 74314556-74314556 on Build GRCh38

View Full Product Details