Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 89206118-89206630 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CH25H
Entrez Gene ID
9023
Gene Name
cholesterol 25-hydroxylase
Gene Aliases
C25H
UniGene
Hs.47357
Gene Chromosome Location
Chr.10: 89205937-89207314 on Build GRCh38
Transcripts
NM_003956.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4016490 c.794G>A p.R265Q Chr.10: 89206499-89206499 on Build GRCh38
5702988 c.675_676delCA p.H225fs*22 Chr.10: 89206617-89206618 on Build GRCh38
5359865 c.677G>T p.R226I Chr.10: 89206616-89206616 on Build GRCh38
1187985 c.673C>T p.H225Y Chr.10: 89206620-89206620 on Build GRCh38
5371258 c.758A>G p.Y253C Chr.10: 89206535-89206535 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs551761521 Chr.10: 89206404-89206404 on Build GRCh38
rs754613437 Chr.10: 89206499-89206499 on Build GRCh38
rs73368297 Chr.10: 89206265-89206265 on Build GRCh38
rs769576953 Chr.10: 89206591-89206591 on Build GRCh38
rs1045813895 Chr.10: 89206468-89206468 on Build GRCh38
rs762877575 Chr.10: 89206595-89206595 on Build GRCh38
rs150599888 Chr.10: 89206478-89206478 on Build GRCh38
rs75732804 Chr.10: 89206216-89206216 on Build GRCh38
rs796889448 Chr.10: 89206360-89206360 on Build GRCh38
rs533405960 Chr.10: 89206392-89206392 on Build GRCh38
rs901319204 Chr.10: 89206344-89206344 on Build GRCh38
rs370786931 Chr.10: 89206582-89206582 on Build GRCh38
rs959203021 Chr.10: 89206311-89206311 on Build GRCh38
rs1044937789 Chr.10: 89206174-89206174 on Build GRCh38
rs746317434 Chr.10: 89206543-89206543 on Build GRCh38
rs975435086 Chr.10: 89206385-89206386 on Build GRCh38
rs555616432 Chr.10: 89206522-89206522 on Build GRCh38
rs944385073 Chr.10: 89206369-89206369 on Build GRCh38
rs757603807 Chr.10: 89206528-89206528 on Build GRCh38
rs528719427 Chr.10: 89206251-89206251 on Build GRCh38
rs776799067 Chr.10: 89206190-89206190 on Build GRCh38
rs760912477 Chr.10: 89206445-89206445 on Build GRCh38
rs746409622 Chr.10: 89206432-89206432 on Build GRCh38
rs761762740 Chr.10: 89206473-89206473 on Build GRCh38
rs907184023 Chr.10: 89206255-89206255 on Build GRCh38
rs562050713 Chr.10: 89206166-89206166 on Build GRCh38
rs544674398 Chr.10: 89206201-89206201 on Build GRCh38
rs773830629 Chr.10: 89206604-89206604 on Build GRCh38
rs768734911 Chr.10: 89206567-89206567 on Build GRCh38
rs376586376 Chr.10: 89206435-89206435 on Build GRCh38
rs1003911504 Chr.10: 89206263-89206263 on Build GRCh38
rs987888063 Chr.10: 89206437-89206437 on Build GRCh38
rs966419331 Chr.10: 89206365-89206365 on Build GRCh38
rs922152351 Chr.10: 89206386-89206386 on Build GRCh38
rs765812799 Chr.10: 89206489-89206489 on Build GRCh38
rs754551239 Chr.10: 89206435-89206435 on Build GRCh38
rs1028547167 Chr.10: 89206374-89206374 on Build GRCh38
rs780275315 Chr.10: 89206561-89206561 on Build GRCh38
rs747139452 Chr.10: 89206564-89206564 on Build GRCh38
rs769819297 Chr.10: 89206484-89206484 on Build GRCh38
rs748346822 Chr.10: 89206587-89206587 on Build GRCh38
rs766365587 Chr.10: 89206596-89206596 on Build GRCh38
rs768964778 Chr.10: 89206188-89206188 on Build GRCh38
rs989275670 Chr.10: 89206329-89206329 on Build GRCh38
rs931224181 Chr.10: 89206602-89206602 on Build GRCh38
rs776949102 Chr.10: 89206576-89206576 on Build GRCh38
rs530045830 Chr.10: 89206512-89206512 on Build GRCh38
rs780550891 Chr.10: 89206152-89206152 on Build GRCh38
rs376974654 Chr.10: 89206537-89206537 on Build GRCh38
rs561916032 Chr.10: 89206287-89206287 on Build GRCh38
rs141627981 Chr.10: 89206477-89206477 on Build GRCh38
rs772710249 Chr.10: 89206559-89206559 on Build GRCh38
rs371119485 Chr.10: 89206447-89206447 on Build GRCh38
rs529381773 Chr.10: 89206354-89206354 on Build GRCh38
rs1039267635 Chr.10: 89206147-89206147 on Build GRCh38
rs569250905 Chr.10: 89206391-89206391 on Build GRCh38
rs926553993 Chr.10: 89206186-89206186 on Build GRCh38
rs775724371 Chr.10: 89206439-89206439 on Build GRCh38
rs977852838 Chr.10: 89206367-89206367 on Build GRCh38
rs201125779 Chr.10: 89206514-89206514 on Build GRCh38
rs770492300 Chr.10: 89206220-89206220 on Build GRCh38
rs201666773 Chr.10: 89206504-89206504 on Build GRCh38
rs776272600 Chr.10: 89206457-89206457 on Build GRCh38
rs747298540 Chr.10: 89206157-89206157 on Build GRCh38
rs201985510 Chr.10: 89206498-89206498 on Build GRCh38
rs949265504 Chr.10: 89206168-89206168 on Build GRCh38
rs904781476 Chr.10: 89206381-89206381 on Build GRCh38
rs146594256 Chr.10: 89206368-89206368 on Build GRCh38
rs765050529 Chr.10: 89206469-89206469 on Build GRCh38
rs761712979 Chr.10: 89206465-89206465 on Build GRCh38

View Full Product Details