Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 90912746-90913262 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD1
Entrez Gene ID
27063
Gene Name
ankyrin repeat domain 1
Gene Aliases
ALRP, C-193, CARP, CVARP, MCARP, bA320F15.2
UniGene
Hs.448589
Gene Chromosome Location
Chr.10: 90912100-90921275 on Build GRCh38
Transcripts
NM_014391.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
236350 c.944G>A p.R315H Chr.10: 90912882-90912882 on Build GRCh38
6194140 c.862C>T p.P288S Chr.10: 90912964-90912964 on Build GRCh38
1297496 c.858G>T p.K286N Chr.10: 90912968-90912968 on Build GRCh38
5837443 c.914G>A p.S305N Chr.10: 90912912-90912912 on Build GRCh38
7090104 c.943C>A p.R315S Chr.10: 90912883-90912883 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201220507 Chr.10: 90912912-90912912 on Build GRCh38
rs988209463 Chr.10: 90912780-90912780 on Build GRCh38
rs765966193 Chr.10: 90912921-90912921 on Build GRCh38
rs879470350 Chr.10: 90913095-90913097 on Build GRCh38
rs749896803 Chr.10: 90912963-90912963 on Build GRCh38
rs754657888 Chr.10: 90912945-90912945 on Build GRCh38
rs922522089 Chr.10: 90913012-90913012 on Build GRCh38
rs776479970 Chr.10: 90912899-90912899 on Build GRCh38
rs771333829 Chr.10: 90912877-90912877 on Build GRCh38
rs78242909 Chr.10: 90912924-90912924 on Build GRCh38
rs542984358 Chr.10: 90912811-90912811 on Build GRCh38
rs767568556 Chr.10: 90912895-90912897 on Build GRCh38
rs751174664 Chr.10: 90912930-90912930 on Build GRCh38
rs746373922 Chr.10: 90913234-90913234 on Build GRCh38
rs770226995 Chr.10: 90913238-90913238 on Build GRCh38
rs562661245 Chr.10: 90913047-90913047 on Build GRCh38
rs774666943 Chr.10: 90912879-90912879 on Build GRCh38
rs752330851 Chr.10: 90913222-90913222 on Build GRCh38
rs988178315 Chr.10: 90912991-90912991 on Build GRCh38
rs776659156 Chr.10: 90913007-90913007 on Build GRCh38
rs377661125 Chr.10: 90912919-90912919 on Build GRCh38
rs746630281 Chr.10: 90912840-90912840 on Build GRCh38
rs779362034 Chr.10: 90912974-90912974 on Build GRCh38
rs750346292 Chr.10: 90912918-90912918 on Build GRCh38
rs747854507 Chr.10: 90912955-90912955 on Build GRCh38
rs762117265 Chr.10: 90913019-90913019 on Build GRCh38
rs761447714 Chr.10: 90912908-90912908 on Build GRCh38
rs534524638 Chr.10: 90912923-90912923 on Build GRCh38
rs369311387 Chr.10: 90912830-90912830 on Build GRCh38
rs1027015592 Chr.10: 90913130-90913130 on Build GRCh38
rs775584071 Chr.10: 90912883-90912883 on Build GRCh38
rs569974704 Chr.10: 90913201-90913201 on Build GRCh38
rs1023104225 Chr.10: 90912904-90912904 on Build GRCh38
rs981398941 Chr.10: 90912809-90912809 on Build GRCh38
rs781529309 Chr.10: 90913229-90913229 on Build GRCh38
rs533806653 Chr.10: 90912796-90912796 on Build GRCh38
rs971266687 Chr.10: 90913054-90913054 on Build GRCh38
rs956776066 Chr.10: 90912771-90912771 on Build GRCh38
rs779743034 Chr.10: 90912838-90912838 on Build GRCh38
rs184765469 Chr.10: 90913154-90913154 on Build GRCh38
rs746392266 Chr.10: 90912882-90912882 on Build GRCh38
rs3939 Chr.10: 90912807-90912807 on Build GRCh38
rs547856037 Chr.10: 90913110-90913110 on Build GRCh38
rs772613237 Chr.10: 90912984-90912984 on Build GRCh38
rs866363957 Chr.10: 90913236-90913236 on Build GRCh38
rs17102350 Chr.10: 90913090-90913090 on Build GRCh38
rs550067720 Chr.10: 90913191-90913191 on Build GRCh38
rs189379353 Chr.10: 90913059-90913059 on Build GRCh38
rs794728975 Chr.10: 90912966-90912966 on Build GRCh38
rs764279574 Chr.10: 90912893-90912893 on Build GRCh38
rs931562060 Chr.10: 90913122-90913122 on Build GRCh38
rs984409218 Chr.10: 90913140-90913140 on Build GRCh38
rs750377661 Chr.10: 90912828-90912828 on Build GRCh38
rs768785507 Chr.10: 90913004-90913004 on Build GRCh38
rs373605037 Chr.10: 90912844-90912844 on Build GRCh38
rs1028634898 Chr.10: 90912929-90912929 on Build GRCh38
rs139438263 Chr.10: 90912965-90912965 on Build GRCh38
rs753622461 Chr.10: 90912816-90912816 on Build GRCh38
rs765093690 Chr.10: 90912917-90912917 on Build GRCh38
rs370857813 Chr.10: 90912975-90912975 on Build GRCh38
rs749684620 Chr.10: 90912859-90912859 on Build GRCh38
rs150520404 Chr.10: 90912825-90912825 on Build GRCh38
rs760862177 Chr.10: 90912892-90912892 on Build GRCh38
rs778781886 Chr.10: 90912827-90912827 on Build GRCh38
rs747156370 Chr.10: 90913003-90913003 on Build GRCh38
rs553516617 Chr.10: 90912806-90912806 on Build GRCh38
rs182272788 Chr.10: 90913146-90913146 on Build GRCh38
rs1049567215 Chr.10: 90913206-90913206 on Build GRCh38
rs889526713 Chr.10: 90912787-90912787 on Build GRCh38
rs778189431 Chr.10: 90912854-90912854 on Build GRCh38
rs758584429 Chr.10: 90912962-90912962 on Build GRCh38
rs985321183 Chr.10: 90913237-90913237 on Build GRCh38
rs867948483 Chr.10: 90912937-90912937 on Build GRCh38
rs577632256 Chr.10: 90912795-90912795 on Build GRCh38
rs775531200 Chr.10: 90913000-90913000 on Build GRCh38
rs1023156517 Chr.10: 90913155-90913155 on Build GRCh38
rs933863194 Chr.10: 90913013-90913013 on Build GRCh38
rs988823253 Chr.10: 90913203-90913203 on Build GRCh38
rs780645558 Chr.10: 90912952-90912952 on Build GRCh38
rs994170048 Chr.10: 90912849-90912849 on Build GRCh38
rs939747588 Chr.10: 90912887-90912887 on Build GRCh38
rs900938404 Chr.10: 90913194-90913194 on Build GRCh38
rs1049558928 Chr.10: 90913034-90913034 on Build GRCh38
rs557441275 Chr.10: 90912788-90912788 on Build GRCh38
rs970183253 Chr.10: 90913183-90913183 on Build GRCh38
rs778099662 Chr.10: 90913113-90913113 on Build GRCh38

View Full Product Details