Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 93527976-93528483 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP55
Entrez Gene ID
55165
Gene Name
centrosomal protein 55
Gene Aliases
C10orf3, CT111, URCC6
UniGene
Hs.14559
Gene Chromosome Location
Chr.10: 93496612-93529094 on Build GRCh38
Transcripts
NM_001127182.1 NM_018131.4 XM_011539918.1 XM_011539919.1 XM_011539920.2 XM_017016372.1 XM_017016373.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
428153 c.1335_1336insC p.Q446fs*6 Chr.10: 93528093-93528094 on Build GRCh38
4016755 c.1310T>C p.L437P Chr.10: 93528068-93528068 on Build GRCh38
5359883 c.1283G>T p.S428I Chr.10: 93528041-93528041 on Build GRCh38
7188185 c.1278delA p.S428fs*>37 Chr.10: 93528036-93528036 on Build GRCh38
1702770 c.1357C>T p.R453C Chr.10: 93528115-93528115 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1028668062 Chr.10: 93528154-93528154 on Build GRCh38
rs181345919 Chr.10: 93528022-93528022 on Build GRCh38
rs769833873 Chr.10: 93528076-93528076 on Build GRCh38
rs969614202 Chr.10: 93528386-93528386 on Build GRCh38
rs560507013 Chr.10: 93528194-93528194 on Build GRCh38
rs756685363 Chr.10: 93528136-93528136 on Build GRCh38
rs753355777 Chr.10: 93528131-93528131 on Build GRCh38
rs781021700 Chr.10: 93528027-93528027 on Build GRCh38
rs140426998 Chr.10: 93528034-93528034 on Build GRCh38
rs901776694 Chr.10: 93528105-93528105 on Build GRCh38
rs554798269 Chr.10: 93528334-93528334 on Build GRCh38
rs770553739 Chr.10: 93528045-93528045 on Build GRCh38
rs34087318 Chr.10: 93528462-93528463 on Build GRCh38
rs146160605 Chr.10: 93528003-93528003 on Build GRCh38
rs760563203 Chr.10: 93528163-93528166 on Build GRCh38
rs751413806 Chr.10: 93528014-93528014 on Build GRCh38
rs777056315 Chr.10: 93528064-93528064 on Build GRCh38
rs574084114 Chr.10: 93528261-93528261 on Build GRCh38
rs372414634 Chr.10: 93528079-93528079 on Build GRCh38
rs992748106 Chr.10: 93528352-93528352 on Build GRCh38
rs755379142 Chr.10: 93528193-93528193 on Build GRCh38
rs186840462 Chr.10: 93528438-93528438 on Build GRCh38
rs918134214 Chr.10: 93528367-93528367 on Build GRCh38
rs776571066 Chr.10: 93528423-93528423 on Build GRCh38
rs201430235 Chr.10: 93528032-93528032 on Build GRCh38
rs778562855 Chr.10: 93528141-93528141 on Build GRCh38
rs766713236 Chr.10: 93528089-93528089 on Build GRCh38
rs900048835 Chr.10: 93528244-93528244 on Build GRCh38
rs113512774 Chr.10: 93528059-93528059 on Build GRCh38
rs75477224 Chr.10: 93528030-93528030 on Build GRCh38
rs369071979 Chr.10: 93528164-93528164 on Build GRCh38
rs111293847 Chr.10: 93528397-93528397 on Build GRCh38
rs1051511803 Chr.10: 93528358-93528358 on Build GRCh38
rs763070570 Chr.10: 93528086-93528086 on Build GRCh38
rs755422759 Chr.10: 93528129-93528129 on Build GRCh38
rs968202669 Chr.10: 93528227-93528227 on Build GRCh38
rs756072428 Chr.10: 93528127-93528127 on Build GRCh38
rs200014799 Chr.10: 93528035-93528035 on Build GRCh38
rs751792256 Chr.10: 93528112-93528112 on Build GRCh38
rs75712754 Chr.10: 93528445-93528445 on Build GRCh38
rs770861276 Chr.10: 93528364-93528364 on Build GRCh38
rs201119474 Chr.10: 93528115-93528115 on Build GRCh38
rs772620389 Chr.10: 93528396-93528397 on Build GRCh38
rs752614348 Chr.10: 93528125-93528125 on Build GRCh38
rs747409579 Chr.10: 93528051-93528051 on Build GRCh38
rs190350954 Chr.10: 93528012-93528012 on Build GRCh38
rs376713552 Chr.10: 93528162-93528162 on Build GRCh38
rs762354067 Chr.10: 93528067-93528067 on Build GRCh38
rs572620662 Chr.10: 93528047-93528047 on Build GRCh38
rs76392816 Chr.10: 93528087-93528087 on Build GRCh38
rs202234576 Chr.10: 93528028-93528028 on Build GRCh38
rs980735946 Chr.10: 93528415-93528415 on Build GRCh38
rs544562270 Chr.10: 93528231-93528231 on Build GRCh38
rs767321494 Chr.10: 93528116-93528116 on Build GRCh38
rs200547927 Chr.10: 93528056-93528056 on Build GRCh38
rs543160106 Chr.10: 93528424-93528424 on Build GRCh38
rs768963170 Chr.10: 93528057-93528057 on Build GRCh38

View Full Product Details