Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 93681419-93681952 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FRA10AC1
Entrez Gene ID
118924
Gene Name
fragile site, folic acid type, rare, fra(10)(q23.3) or fra(10)(q24.2) candidate 1
Gene Aliases
C10orf4, F26C11.1-like, FRA10A
UniGene
Hs.586650
Gene Chromosome Location
Chr.10: 93667883-93702572 on Build GRCh38
Transcripts
NM_145246.4 XM_005269515.4 XM_005269516.4 XM_011539259.2 XM_011539260.2 XM_017015656.1 XM_017015657.1 XM_017015658.1 XM_017015659.1 XM_017015660.1 XM_017015661.1 XM_017015662.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5999154 c.756T>C p.S252S Chr.10: 93681511-93681511 on Build GRCh38
6274180 c.708C>A p.T236T Chr.10: 93681559-93681559 on Build GRCh38
5961092 c.760G>A p.E254K Chr.10: 93681507-93681507 on Build GRCh38
4333798 c.694delA p.R232fs*>61 Chr.10: 93681573-93681573 on Build GRCh38
5961090 c.760G>A p.E254K Chr.10: 93681507-93681507 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs71480876 Chr.10: 93681869-93681869 on Build GRCh38
rs906618220 Chr.10: 93681841-93681841 on Build GRCh38
rs751715737 Chr.10: 93681477-93681477 on Build GRCh38
rs915465048 Chr.10: 93681776-93681776 on Build GRCh38
rs752795769 Chr.10: 93681630-93681630 on Build GRCh38
rs775948504 Chr.10: 93681573-93681573 on Build GRCh38
rs774931601 Chr.10: 93681560-93681560 on Build GRCh38
rs1057373755 Chr.10: 93681887-93681887 on Build GRCh38
rs746694549 Chr.10: 93681573-93681573 on Build GRCh38
rs992722697 Chr.10: 93681823-93681823 on Build GRCh38
rs375390820 Chr.10: 93681549-93681549 on Build GRCh38
rs527853731 Chr.10: 93681700-93681700 on Build GRCh38
rs917162343 Chr.10: 93681854-93681854 on Build GRCh38
rs765610649 Chr.10: 93681458-93681458 on Build GRCh38
rs771583990 Chr.10: 93681558-93681558 on Build GRCh38
rs11593026 Chr.10: 93681843-93681843 on Build GRCh38
rs777753842 Chr.10: 93681512-93681512 on Build GRCh38
rs200530414 Chr.10: 93681922-93681923 on Build GRCh38
rs370230472 Chr.10: 93681470-93681470 on Build GRCh38
rs963045288 Chr.10: 93681565-93681565 on Build GRCh38
rs767204685 Chr.10: 93681506-93681506 on Build GRCh38
rs770995294 Chr.10: 93681765-93681768 on Build GRCh38
rs760440101 Chr.10: 93681459-93681463 on Build GRCh38
rs760076086 Chr.10: 93681616-93681616 on Build GRCh38
rs773672925 Chr.10: 93681577-93681577 on Build GRCh38
rs78315882 Chr.10: 93681648-93681648 on Build GRCh38
rs967073541 Chr.10: 93681868-93681868 on Build GRCh38
rs7914441 Chr.10: 93681495-93681495 on Build GRCh38
rs774721843 Chr.10: 93681615-93681615 on Build GRCh38
rs142903469 Chr.10: 93681783-93681783 on Build GRCh38
rs532523836 Chr.10: 93681511-93681511 on Build GRCh38
rs371183108 Chr.10: 93681473-93681473 on Build GRCh38
rs377093425 Chr.10: 93681446-93681446 on Build GRCh38
rs764669527 Chr.10: 93681624-93681628 on Build GRCh38
rs746823411 Chr.10: 93681459-93681459 on Build GRCh38
rs988053271 Chr.10: 93681672-93681672 on Build GRCh38
rs11187583 Chr.10: 93681515-93681515 on Build GRCh38
rs954425235 Chr.10: 93681659-93681659 on Build GRCh38
rs752993930 Chr.10: 93681505-93681505 on Build GRCh38
rs971699309 Chr.10: 93681566-93681566 on Build GRCh38
rs910692296 Chr.10: 93681718-93681718 on Build GRCh38
rs548073257 Chr.10: 93681775-93681775 on Build GRCh38
rs771531865 Chr.10: 93681603-93681603 on Build GRCh38
rs543481815 Chr.10: 93681700-93681700 on Build GRCh38
rs147872799 Chr.10: 93681485-93681485 on Build GRCh38
rs567824487 Chr.10: 93681780-93681780 on Build GRCh38
rs770671653 Chr.10: 93681563-93681567 on Build GRCh38
rs745751956 Chr.10: 93681555-93681555 on Build GRCh38
rs768265366 Chr.10: 93681574-93681574 on Build GRCh38
rs193119517 Chr.10: 93681628-93681628 on Build GRCh38
rs763171602 Chr.10: 93681616-93681617 on Build GRCh38
rs545360784 Chr.10: 93681588-93681588 on Build GRCh38
rs900771638 Chr.10: 93681467-93681467 on Build GRCh38
rs943636782 Chr.10: 93681730-93681730 on Build GRCh38
rs568951794 Chr.10: 93681740-93681740 on Build GRCh38
rs760343965 Chr.10: 93681842-93681842 on Build GRCh38
rs760263097 Chr.10: 93681457-93681457 on Build GRCh38

View Full Product Details