Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 97435970-97436514 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXOSC1
Entrez Gene ID
51013
Gene Name
exosome component 1
Gene Aliases
CGI-108, CSL4, Csl4p, SKI4, Ski4p, p13
UniGene
Hs.632089
Gene Chromosome Location
Chr.10: 97435909-97446065 on Build GRCh38
Transcripts
NM_001318362.1 NM_001318363.1 NM_001318364.1 NM_001318365.1 NM_001318366.1 NM_016046.4 NR_134586.1 NR_134587.1 XM_011539847.2 XM_017016304.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4017064 c.560G>A p.R187Q Chr.10: 97436473-97436473 on Build GRCh38
6657266 c.547C>T p.R183W Chr.10: 97436486-97436486 on Build GRCh38
7318622 c.534C>A p.H178Q Chr.10: 97436499-97436499 on Build GRCh38
1661724 c.571G>A p.E191K Chr.10: 97436462-97436462 on Build GRCh38
4017065 c.546C>G p.F182L Chr.10: 97436487-97436487 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372655992 Chr.10: 97436359-97436359 on Build GRCh38
rs926381034 Chr.10: 97436271-97436271 on Build GRCh38
rs750106196 Chr.10: 97436258-97436258 on Build GRCh38
rs939689142 Chr.10: 97436326-97436326 on Build GRCh38
rs375542814 Chr.10: 97436427-97436427 on Build GRCh38
rs544559760 Chr.10: 97436310-97436310 on Build GRCh38
rs796616843 Chr.10: 97436309-97436310 on Build GRCh38
rs112292467 Chr.10: 97436267-97436267 on Build GRCh38
rs769017478 Chr.10: 97436425-97436425 on Build GRCh38
rs139925103 Chr.10: 97436349-97436350 on Build GRCh38
rs1039049622 Chr.10: 97436345-97436345 on Build GRCh38
rs904745680 Chr.10: 97436056-97436056 on Build GRCh38
rs763158354 Chr.10: 97436447-97436447 on Build GRCh38
rs1008232084 Chr.10: 97436024-97436024 on Build GRCh38
rs540572166 Chr.10: 97436066-97436066 on Build GRCh38
rs754986273 Chr.10: 97436463-97436463 on Build GRCh38
rs565141558 Chr.10: 97436215-97436215 on Build GRCh38
rs904795316 Chr.10: 97436474-97436474 on Build GRCh38
rs770567388 Chr.10: 97436401-97436401 on Build GRCh38
rs149684008 Chr.10: 97436466-97436466 on Build GRCh38
rs201390574 Chr.10: 97436351-97436351 on Build GRCh38
rs1043345541 Chr.10: 97436045-97436045 on Build GRCh38
rs555091336 Chr.10: 97436023-97436023 on Build GRCh38
rs200086623 Chr.10: 97436411-97436411 on Build GRCh38
rs368248968 Chr.10: 97436462-97436462 on Build GRCh38
rs866810782 Chr.10: 97436477-97436477 on Build GRCh38
rs777641312 Chr.10: 97436473-97436473 on Build GRCh38
rs773144056 Chr.10: 97436364-97436364 on Build GRCh38
rs765534888 Chr.10: 97436440-97436440 on Build GRCh38
rs772078293 Chr.10: 97436258-97436261 on Build GRCh38
rs563234145 Chr.10: 97436357-97436357 on Build GRCh38
rs766540907 Chr.10: 97436455-97436455 on Build GRCh38
rs750655639 Chr.10: 97436444-97436444 on Build GRCh38
rs773727544 Chr.10: 97436406-97436406 on Build GRCh38
rs773023594 Chr.10: 97436394-97436394 on Build GRCh38
rs980567152 Chr.10: 97436225-97436225 on Build GRCh38
rs749011600 Chr.10: 97436484-97436484 on Build GRCh38
rs145472895 Chr.10: 97436486-97436486 on Build GRCh38
rs1062893 Chr.10: 97436232-97436232 on Build GRCh38
rs200404619 Chr.10: 97436346-97436346 on Build GRCh38
rs142445171 Chr.10: 97436360-97436360 on Build GRCh38
rs760596875 Chr.10: 97436405-97436405 on Build GRCh38
rs1022150932 Chr.10: 97436162-97436162 on Build GRCh38
rs998669468 Chr.10: 97436297-97436297 on Build GRCh38
rs770692182 Chr.10: 97436485-97436485 on Build GRCh38
rs766982184 Chr.10: 97436257-97436257 on Build GRCh38
rs984059312 Chr.10: 97436320-97436320 on Build GRCh38
rs965530646 Chr.10: 97436256-97436256 on Build GRCh38
rs748895019 Chr.10: 97436399-97436399 on Build GRCh38
rs967896078 Chr.10: 97436187-97436187 on Build GRCh38
rs752635286 Chr.10: 97436465-97436465 on Build GRCh38
rs568244423 Chr.10: 97436163-97436165 on Build GRCh38
rs1009930344 Chr.10: 97436134-97436134 on Build GRCh38
rs781025980 Chr.10: 97436464-97436464 on Build GRCh38
rs762197185 Chr.10: 97436437-97436437 on Build GRCh38
rs372289644 Chr.10: 97436038-97436038 on Build GRCh38
rs763623301 Chr.10: 97436404-97436406 on Build GRCh38
rs960404099 Chr.10: 97436305-97436305 on Build GRCh38
rs532803388 Chr.10: 97436309-97436309 on Build GRCh38

View Full Product Details