Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 97443167-97443514 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXOSC1
Entrez Gene ID
51013
Gene Name
exosome component 1
Gene Aliases
CGI-108, CSL4, Csl4p, SKI4, Ski4p, p13
UniGene
Hs.632089
Gene Chromosome Location
Chr.10: 97435909-97446065 on Build GRCh38
Transcripts
NM_001318362.1 NM_001318363.1 NM_001318364.1 NM_001318365.1 NM_001318366.1 NM_016046.4 NR_134586.1 NR_134587.1 XM_011539847.2 XM_017016304.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4963654 c.186G>A p.Q62Q Chr.10: 97443273-97443273 on Build GRCh38
4827158 c.170G>C p.R57T Chr.10: 97443289-97443289 on Build GRCh38
1972216 c.193C>T p.P65S Chr.10: 97443266-97443266 on Build GRCh38
6067077 c.201G>C p.V67V Chr.10: 97443258-97443258 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs937835556 Chr.10: 97443302-97443302 on Build GRCh38
rs779947939 Chr.10: 97443196-97443196 on Build GRCh38
rs111511939 Chr.10: 97443293-97443293 on Build GRCh38
rs550878305 Chr.10: 97443389-97443389 on Build GRCh38
rs754061288 Chr.10: 97443284-97443284 on Build GRCh38
rs974121266 Chr.10: 97443410-97443411 on Build GRCh38
rs150226020 Chr.10: 97443262-97443262 on Build GRCh38
rs750447292 Chr.10: 97443312-97443312 on Build GRCh38
rs760141247 Chr.10: 97443243-97443243 on Build GRCh38
rs528200594 Chr.10: 97443218-97443218 on Build GRCh38
rs10101 Chr.10: 97443290-97443290 on Build GRCh38
rs764382222 Chr.10: 97443273-97443273 on Build GRCh38
rs772838751 Chr.10: 97443238-97443238 on Build GRCh38
rs908662976 Chr.10: 97443397-97443397 on Build GRCh38
rs41290444 Chr.10: 97443408-97443408 on Build GRCh38
rs185973814 Chr.10: 97443321-97443321 on Build GRCh38
rs377082810 Chr.10: 97443233-97443233 on Build GRCh38
rs569214606 Chr.10: 97443400-97443400 on Build GRCh38
rs532171831 Chr.10: 97443382-97443382 on Build GRCh38
rs772430419 Chr.10: 97443250-97443250 on Build GRCh38
rs1000519724 Chr.10: 97443213-97443213 on Build GRCh38
rs375551957 Chr.10: 97443332-97443332 on Build GRCh38
rs937973668 Chr.10: 97443470-97443470 on Build GRCh38
rs762016854 Chr.10: 97443289-97443289 on Build GRCh38
rs758399895 Chr.10: 97443320-97443320 on Build GRCh38
rs896199171 Chr.10: 97443386-97443386 on Build GRCh38
rs61861914 Chr.10: 97443483-97443483 on Build GRCh38
rs376542058 Chr.10: 97443439-97443439 on Build GRCh38
rs747737708 Chr.10: 97443235-97443235 on Build GRCh38
rs746750260 Chr.10: 97443216-97443216 on Build GRCh38
rs758253626 Chr.10: 97443193-97443193 on Build GRCh38
rs368996987 Chr.10: 97443253-97443253 on Build GRCh38
rs751457559 Chr.10: 97443333-97443333 on Build GRCh38
rs754734995 Chr.10: 97443340-97443340 on Build GRCh38
rs566810692 Chr.10: 97443472-97443472 on Build GRCh38
rs190038400 Chr.10: 97443352-97443352 on Build GRCh38
rs769497935 Chr.10: 97443236-97443236 on Build GRCh38
rs997073036 Chr.10: 97443279-97443280 on Build GRCh38

View Full Product Details