Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 125002873-125003378 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTBP2
Entrez Gene ID
1488
Gene Name
C-terminal binding protein 2
Gene Aliases
-
UniGene
Hs.501345
Gene Chromosome Location
Chr.10: 124987849-125161061 on Build GRCh38
Transcripts
NM_001083914.2 NM_001290214.2 NM_001290215.2 NM_001321012.1 NM_001321013.1 NM_001321014.1 NM_001329.3 NM_022802.2 XM_005269561.2 XM_005269564.2 XM_005269567.2 XM_005269568.4 XM_005269569.2 XM_005269571.2 XM_005269572.3 XM_006717642.2 XM_011539349.2 XM_011539351.1 XM_011539353.1 XM_011539354.1 XM_011539355.1 XM_011539357.1 XM_011539358.1 XM_017015756.1 XM_017015757.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2021399 c.191C>T p.S64L Chr.10: 125003360-125003360 on Build GRCh38
6055815 c.1871C>A p.T624N Chr.10: 125003067-125003067 on Build GRCh38
4012470 c.186G>A p.A62A Chr.10: 125003365-125003365 on Build GRCh38
5009185 c.1923C>T p.I641I Chr.10: 125003015-125003015 on Build GRCh38
5763463 c.1806G>T p.A602A Chr.10: 125003365-125003365 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs796213917 Chr.10: 125003048-125003048 on Build GRCh38
rs763088730 Chr.10: 125003356-125003356 on Build GRCh38
rs767975764 Chr.10: 125003108-125003108 on Build GRCh38
rs747478715 Chr.10: 125003320-125003320 on Build GRCh38
rs777026257 Chr.10: 125003130-125003130 on Build GRCh38
rs766930815 Chr.10: 125003252-125003252 on Build GRCh38
rs755930819 Chr.10: 125003155-125003155 on Build GRCh38
rs748896311 Chr.10: 125003089-125003089 on Build GRCh38
rs780995499 Chr.10: 125002952-125002952 on Build GRCh38
rs766694674 Chr.10: 125003029-125003029 on Build GRCh38
rs981877618 Chr.10: 125003264-125003264 on Build GRCh38
rs754593092 Chr.10: 125003312-125003312 on Build GRCh38
rs376730588 Chr.10: 125003154-125003154 on Build GRCh38
rs200069004 Chr.10: 125002939-125002939 on Build GRCh38
rs750232291 Chr.10: 125003081-125003081 on Build GRCh38
rs751476858 Chr.10: 125003061-125003061 on Build GRCh38
rs376425040 Chr.10: 125002918-125002918 on Build GRCh38
rs199688093 Chr.10: 125003313-125003313 on Build GRCh38
rs761727708 Chr.10: 125003146-125003146 on Build GRCh38
rs796348541 Chr.10: 125003066-125003066 on Build GRCh38
rs376708829 Chr.10: 125002929-125002929 on Build GRCh38
rs564040819 Chr.10: 125002914-125002914 on Build GRCh38
rs148802158 Chr.10: 125002970-125002970 on Build GRCh38
rs796470232 Chr.10: 125003011-125003011 on Build GRCh38
rs767260867 Chr.10: 125003072-125003072 on Build GRCh38
rs762085279 Chr.10: 125002927-125002927 on Build GRCh38
rs370542092 Chr.10: 125003330-125003330 on Build GRCh38
rs372182878 Chr.10: 125002923-125002923 on Build GRCh38
rs145557678 Chr.10: 125003015-125003015 on Build GRCh38
rs753535345 Chr.10: 125003297-125003297 on Build GRCh38
rs796400395 Chr.10: 125003040-125003040 on Build GRCh38
rs760489730 Chr.10: 125003010-125003010 on Build GRCh38
rs990260929 Chr.10: 125003234-125003234 on Build GRCh38
rs200605790 Chr.10: 125002943-125002943 on Build GRCh38
rs998701062 Chr.10: 125003205-125003205 on Build GRCh38
rs754890815 Chr.10: 125002919-125002928 on Build GRCh38
rs201673487 Chr.10: 125003316-125003316 on Build GRCh38
rs374514317 Chr.10: 125003143-125003143 on Build GRCh38
rs3198935 Chr.10: 125003091-125003091 on Build GRCh38
rs3198931 Chr.10: 125003359-125003359 on Build GRCh38
rs765114681 Chr.10: 125003149-125003149 on Build GRCh38
rs201760950 Chr.10: 125003036-125003036 on Build GRCh38
rs140067472 Chr.10: 125003083-125003083 on Build GRCh38
rs926218020 Chr.10: 125003166-125003166 on Build GRCh38
rs368944665 Chr.10: 125003084-125003084 on Build GRCh38
rs76079088 Chr.10: 125002969-125002969 on Build GRCh38
rs774780122 Chr.10: 125002926-125002926 on Build GRCh38
rs957165248 Chr.10: 125003233-125003233 on Build GRCh38
rs147937735 Chr.10: 125003045-125003045 on Build GRCh38
rs368312635 Chr.10: 125003328-125003328 on Build GRCh38
rs74161001 Chr.10: 125003158-125003158 on Build GRCh38
rs191421803 Chr.10: 125003096-125003096 on Build GRCh38
rs188280364 Chr.10: 125003014-125003014 on Build GRCh38
rs936227031 Chr.10: 125003223-125003223 on Build GRCh38
rs745561150 Chr.10: 125002960-125002960 on Build GRCh38
rs561234353 Chr.10: 125002951-125002951 on Build GRCh38
rs879071956 Chr.10: 125003105-125003338 on Build GRCh38
rs796604157 Chr.10: 125003059-125003059 on Build GRCh38
rs112919818 Chr.10: 125002930-125002930 on Build GRCh38
rs796602813 Chr.10: 125003026-125003026 on Build GRCh38
rs112239066 Chr.10: 125003006-125003006 on Build GRCh38
rs376628142 Chr.10: 125003129-125003129 on Build GRCh38
rs548115460 Chr.10: 125003174-125003174 on Build GRCh38
rs376221975 Chr.10: 125002944-125002944 on Build GRCh38
rs764999884 Chr.10: 125002966-125002966 on Build GRCh38
rs75794788 Chr.10: 125003065-125003065 on Build GRCh38
rs930382544 Chr.10: 125003134-125003134 on Build GRCh38
rs1031485227 Chr.10: 125003220-125003220 on Build GRCh38
rs144209188 Chr.10: 125002967-125002967 on Build GRCh38
rs797010298 Chr.10: 125003046-125003046 on Build GRCh38
rs750771534 Chr.10: 125003184-125003184 on Build GRCh38
rs776936228 Chr.10: 125003033-125003033 on Build GRCh38
rs913389958 Chr.10: 125003251-125003251 on Build GRCh38
rs768573864 Chr.10: 125003357-125003357 on Build GRCh38
rs940240409 Chr.10: 125003111-125003111 on Build GRCh38
rs775494980 Chr.10: 125003341-125003341 on Build GRCh38
rs757198924 Chr.10: 125003063-125003063 on Build GRCh38
rs796433756 Chr.10: 125002983-125002983 on Build GRCh38
rs3198936 Chr.10: 125003062-125003062 on Build GRCh38
rs753961985 Chr.10: 125002940-125002940 on Build GRCh38
rs111794401 Chr.10: 125003007-125003007 on Build GRCh38
rs375594155 Chr.10: 125003327-125003327 on Build GRCh38
rs796960419 Chr.10: 125003074-125003074 on Build GRCh38
rs148437170 Chr.10: 125003282-125003282 on Build GRCh38
rs142442450 Chr.10: 125003009-125003009 on Build GRCh38
rs754004626 Chr.10: 125003272-125003272 on Build GRCh38
rs545609926 Chr.10: 125003293-125003293 on Build GRCh38
rs201919800 Chr.10: 125003090-125003090 on Build GRCh38
rs1019311624 Chr.10: 125003018-125003018 on Build GRCh38
rs747392692 Chr.10: 125003118-125003118 on Build GRCh38
rs757787645 Chr.10: 125003314-125003314 on Build GRCh38
rs756689890 Chr.10: 125003236-125003236 on Build GRCh38
rs770054294 Chr.10: 125003331-125003331 on Build GRCh38
rs375075079 Chr.10: 125002935-125002935 on Build GRCh38
rs535205127 Chr.10: 125003321-125003321 on Build GRCh38
rs781676037 Chr.10: 125003324-125003324 on Build GRCh38
rs796562265 Chr.10: 125002994-125002994 on Build GRCh38
rs112508291 Chr.10: 125003008-125003008 on Build GRCh38
rs369168989 Chr.10: 125002922-125002922 on Build GRCh38
rs534315612 Chr.10: 125003163-125003163 on Build GRCh38
rs774960418 Chr.10: 125002919-125002919 on Build GRCh38
rs950209915 Chr.10: 125003263-125003263 on Build GRCh38
rs773506784 Chr.10: 125003114-125003114 on Build GRCh38
rs2241856 Chr.10: 125003142-125003142 on Build GRCh38
rs554950642 Chr.10: 125003157-125003157 on Build GRCh38
rs796136029 Chr.10: 125003350-125003350 on Build GRCh38
rs200173220 Chr.10: 125003039-125003039 on Build GRCh38
rs4348846 Chr.10: 125002902-125002902 on Build GRCh38
rs551272100 Chr.10: 125003109-125003109 on Build GRCh38
rs111629462 Chr.10: 125002961-125002961 on Build GRCh38

View Full Product Details