Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 31432659-31433161 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJC24
Entrez Gene ID
120526
Gene Name
DnaJ heat shock protein family (Hsp40) member C24
Gene Aliases
DPH4, JJJ3, ZCSL3
UniGene
Hs.187269
Gene Chromosome Location
Chr.11: 31369829-31432835 on Build GRCh38
Transcripts
NM_181706.4 XM_017017216.1 NM_001304274.1 NM_144981.2 XM_005252812.3 XM_011519942.1 XM_011519943.2 XM_011519944.2 XM_011519945.2 XM_011519946.2 XM_011519947.2 XM_017017305.1 XM_017017306.1

Gene Symbol
IMMP1L
Entrez Gene ID
196294
Gene Name
inner mitochondrial membrane peptidase subunit 1
Gene Aliases
IMMP1, IMP1, IMP1-LIKE
UniGene
Hs.502223
Gene Chromosome Location
Chr.11: 31432399-31509644 on Build GRCh38
Transcripts
NM_181706.4 XM_017017216.1 NM_001304274.1 NM_144981.2 XM_005252812.3 XM_011519942.1 XM_011519943.2 XM_011519944.2 XM_011519945.2 XM_011519946.2 XM_011519947.2 XM_017017305.1 XM_017017306.1

SNP Detail

SNP ID Chromosome Location
rs998715611 Chr.11: 31432774-31432774 on Build GRCh38
rs560637187 Chr.11: 31432887-31432887 on Build GRCh38
rs753924313 Chr.11: 31433030-31433030 on Build GRCh38
rs916068934 Chr.11: 31432963-31432963 on Build GRCh38
rs750903334 Chr.11: 31432780-31432780 on Build GRCh38
rs756584770 Chr.11: 31432927-31432927 on Build GRCh38
rs905979453 Chr.11: 31432957-31432957 on Build GRCh38
rs766645264 Chr.11: 31432899-31432899 on Build GRCh38
rs899625654 Chr.11: 31432710-31432710 on Build GRCh38
rs926157268 Chr.11: 31432693-31432693 on Build GRCh38
rs549346100 Chr.11: 31433052-31433052 on Build GRCh38
rs950610628 Chr.11: 31433104-31433104 on Build GRCh38
rs998014405 Chr.11: 31433078-31433078 on Build GRCh38
rs566046408 Chr.11: 31432879-31432879 on Build GRCh38
rs539864741 Chr.11: 31433125-31433125 on Build GRCh38
rs951387184 Chr.11: 31432778-31432779 on Build GRCh38
rs751554151 Chr.11: 31433032-31433032 on Build GRCh38
rs1008591730 Chr.11: 31433116-31433116 on Build GRCh38
rs144126408 Chr.11: 31433135-31433135 on Build GRCh38
rs899695350 Chr.11: 31433074-31433074 on Build GRCh38
rs950086247 Chr.11: 31432855-31432855 on Build GRCh38
rs191154551 Chr.11: 31433059-31433059 on Build GRCh38
rs914875226 Chr.11: 31433063-31433063 on Build GRCh38
rs1017404077 Chr.11: 31433018-31433018 on Build GRCh38
rs936146564 Chr.11: 31432741-31432741 on Build GRCh38
rs527645853 Chr.11: 31432987-31432987 on Build GRCh38
rs1039621644 Chr.11: 31433072-31433072 on Build GRCh38
rs1001559358 Chr.11: 31432978-31432978 on Build GRCh38
rs927704925 Chr.11: 31433034-31433034 on Build GRCh38
rs533363287 Chr.11: 31432689-31432689 on Build GRCh38
rs1029741509 Chr.11: 31432775-31432775 on Build GRCh38
rs551432389 Chr.11: 31432816-31432816 on Build GRCh38
rs538114585 Chr.11: 31433067-31433067 on Build GRCh38
rs899015603 Chr.11: 31433053-31433053 on Build GRCh38
rs1016920711 Chr.11: 31432870-31432870 on Build GRCh38
rs892006507 Chr.11: 31432821-31432821 on Build GRCh38
rs1051202919 Chr.11: 31433106-31433106 on Build GRCh38
rs752677500 Chr.11: 31432847-31432849 on Build GRCh38

View Full Product Details