Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 34438844-34439340 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAT
Entrez Gene ID
847
Gene Name
catalase
Gene Aliases
-
UniGene
Hs.502302
Gene Chromosome Location
Chr.11: 34438925-34472060 on Build GRCh38
Transcripts
NM_001752.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1604507 c.5C>A p.A2D Chr.11: 34439018-34439018 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs759989720 Chr.11: 34439117-34439117 on Build GRCh38
rs17882039 Chr.11: 34439315-34439315 on Build GRCh38
rs961226473 Chr.11: 34439255-34439255 on Build GRCh38
rs754943583 Chr.11: 34438982-34438982 on Build GRCh38
rs775538627 Chr.11: 34438999-34438999 on Build GRCh38
rs763613655 Chr.11: 34439085-34439085 on Build GRCh38
rs549489322 Chr.11: 34439226-34439226 on Build GRCh38
rs766875436 Chr.11: 34438981-34438981 on Build GRCh38
rs979411943 Chr.11: 34438880-34438880 on Build GRCh38
rs761943573 Chr.11: 34439001-34439002 on Build GRCh38
rs1020793534 Chr.11: 34438872-34438872 on Build GRCh38
rs755612125 Chr.11: 34439082-34439082 on Build GRCh38
rs752119282 Chr.11: 34439073-34439073 on Build GRCh38
rs868807194 Chr.11: 34439048-34439048 on Build GRCh38
rs1008184375 Chr.11: 34439242-34439242 on Build GRCh38
rs759951974 Chr.11: 34439068-34439068 on Build GRCh38
rs755700704 Chr.11: 34438996-34438996 on Build GRCh38
rs780078064 Chr.11: 34439103-34439103 on Build GRCh38
rs541723807 Chr.11: 34439112-34439112 on Build GRCh38
rs369780335 Chr.11: 34439166-34439166 on Build GRCh38
rs565118068 Chr.11: 34439279-34439279 on Build GRCh38
rs776556838 Chr.11: 34439017-34439017 on Build GRCh38
rs898531876 Chr.11: 34438952-34438952 on Build GRCh38
rs919853915 Chr.11: 34439115-34439115 on Build GRCh38
rs1046769903 Chr.11: 34438927-34438927 on Build GRCh38
rs10836235 Chr.11: 34439157-34439157 on Build GRCh38
rs752057739 Chr.11: 34438985-34438985 on Build GRCh38
rs780567690 Chr.11: 34438991-34438991 on Build GRCh38
rs771337950 Chr.11: 34439108-34439108 on Build GRCh38
rs758245446 Chr.11: 34439097-34439097 on Build GRCh38
rs763284357 Chr.11: 34439037-34439037 on Build GRCh38
rs1007998317 Chr.11: 34438948-34438948 on Build GRCh38
rs200040258 Chr.11: 34439022-34439022 on Build GRCh38
rs937249904 Chr.11: 34438900-34438900 on Build GRCh38
rs375278815 Chr.11: 34439014-34439014 on Build GRCh38
rs997097817 Chr.11: 34438984-34438984 on Build GRCh38
rs553504423 Chr.11: 34439136-34439136 on Build GRCh38
rs899959891 Chr.11: 34439187-34439187 on Build GRCh38
rs7943316 Chr.11: 34438925-34438925 on Build GRCh38
rs920039337 Chr.11: 34439306-34439306 on Build GRCh38
rs914001789 Chr.11: 34438870-34438870 on Build GRCh38
rs1000556400 Chr.11: 34439208-34439208 on Build GRCh38
rs534053039 Chr.11: 34439105-34439105 on Build GRCh38
rs748642827 Chr.11: 34439268-34439268 on Build GRCh38
rs752041214 Chr.11: 34439055-34439055 on Build GRCh38
rs938817273 Chr.11: 34439179-34439179 on Build GRCh38
rs750791814 Chr.11: 34438976-34438976 on Build GRCh38
rs967763945 Chr.11: 34439032-34439032 on Build GRCh38
rs773526579 Chr.11: 34439034-34439034 on Build GRCh38
rs1042167749 Chr.11: 34439203-34439203 on Build GRCh38
rs574706277 Chr.11: 34439049-34439049 on Build GRCh38
rs17887248 Chr.11: 34439292-34439292 on Build GRCh38
rs995140605 Chr.11: 34438926-34438926 on Build GRCh38
rs536361162 Chr.11: 34438877-34438877 on Build GRCh38
rs755516735 Chr.11: 34438987-34438987 on Build GRCh38
rs988191013 Chr.11: 34438866-34438866 on Build GRCh38
rs373225524 Chr.11: 34438890-34438891 on Build GRCh38
rs768119395 Chr.11: 34439072-34439072 on Build GRCh38
rs928517721 Chr.11: 34438894-34438894 on Build GRCh38
rs772046658 Chr.11: 34439102-34439110 on Build GRCh38
rs1009852536 Chr.11: 34439254-34439254 on Build GRCh38
rs550205524 Chr.11: 34438886-34438886 on Build GRCh38
rs1009252894 Chr.11: 34439196-34439196 on Build GRCh38
rs765549657 Chr.11: 34438969-34438969 on Build GRCh38
rs12270780 Chr.11: 34439164-34439164 on Build GRCh38
rs769872790 Chr.11: 34439025-34439025 on Build GRCh38
rs953953385 Chr.11: 34439225-34439225 on Build GRCh38
rs1044678175 Chr.11: 34438973-34438973 on Build GRCh38
rs997030843 Chr.11: 34439194-34439194 on Build GRCh38
rs763040216 Chr.11: 34439013-34439013 on Build GRCh38
rs145028042 Chr.11: 34439146-34439146 on Build GRCh38
rs777084804 Chr.11: 34439312-34439312 on Build GRCh38
rs1049982 Chr.11: 34438994-34438994 on Build GRCh38
rs769855947 Chr.11: 34439106-34439106 on Build GRCh38
rs921079483 Chr.11: 34438885-34438886 on Build GRCh38
rs1020187286 Chr.11: 34438974-34438974 on Build GRCh38
rs11604331 Chr.11: 34439222-34439222 on Build GRCh38
rs1045955972 Chr.11: 34438889-34438889 on Build GRCh38
rs986854207 Chr.11: 34438921-34438921 on Build GRCh38
rs560430662 Chr.11: 34439128-34439128 on Build GRCh38
rs753429263 Chr.11: 34439086-34439086 on Build GRCh38
rs912618928 Chr.11: 34439127-34439127 on Build GRCh38
rs746948846 Chr.11: 34439104-34439104 on Build GRCh38
rs759427596 Chr.11: 34438942-34438942 on Build GRCh38
rs968361899 Chr.11: 34438874-34438874 on Build GRCh38
rs368753937 Chr.11: 34439150-34439150 on Build GRCh38
rs772612322 Chr.11: 34439309-34439309 on Build GRCh38
rs773430994 Chr.11: 34439107-34439107 on Build GRCh38
rs571944149 Chr.11: 34439147-34439147 on Build GRCh38
rs756907377 Chr.11: 34439087-34439087 on Build GRCh38
rs894045116 Chr.11: 34439240-34439240 on Build GRCh38
rs972782730 Chr.11: 34439259-34439259 on Build GRCh38
rs1055680029 Chr.11: 34438917-34438917 on Build GRCh38
rs903489456 Chr.11: 34439204-34439204 on Build GRCh38
rs766786703 Chr.11: 34439043-34439043 on Build GRCh38
rs758961898 Chr.11: 34439293-34439293 on Build GRCh38
rs964276907 Chr.11: 34438979-34438979 on Build GRCh38
rs761999073 Chr.11: 34439024-34439024 on Build GRCh38
rs761932291 Chr.11: 34438968-34438968 on Build GRCh38
rs777522545 Chr.11: 34438997-34438997 on Build GRCh38
rs371015721 Chr.11: 34439094-34439094 on Build GRCh38
rs557973663 Chr.11: 34439006-34439006 on Build GRCh38
rs527997748 Chr.11: 34439304-34439304 on Build GRCh38
rs562711090 Chr.11: 34439274-34439274 on Build GRCh38
rs939965572 Chr.11: 34438929-34438929 on Build GRCh38
rs745733562 Chr.11: 34439095-34439095 on Build GRCh38

View Full Product Details