Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 57660742-57661243 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLP1
Entrez Gene ID
10978
Gene Name
cleavage and polyadenylation factor I subunit 1
Gene Aliases
HEAB, hClp1
UniGene
Hs.523687
Gene Chromosome Location
Chr.11: 57657744-57661865 on Build GRCh38
Transcripts
NM_001142597.1 NM_006831.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7122532 c.803G>T p.R268L Chr.11: 57660961-57660961 on Build GRCh38
1637845 c.905G>A p.R302H Chr.11: 57661063-57661063 on Build GRCh38
1628082 c.770T>C p.L257P Chr.11: 57660928-57660928 on Build GRCh38
1604769 c.692G>C p.C231S Chr.11: 57660850-57660850 on Build GRCh38
688379 c.957C>A p.V319V Chr.11: 57661115-57661115 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs138402857 Chr.11: 57660959-57660959 on Build GRCh38
rs777290319 Chr.11: 57660774-57660774 on Build GRCh38
rs1804745 Chr.11: 57661048-57661048 on Build GRCh38
rs760641774 Chr.11: 57660918-57660918 on Build GRCh38
rs964731231 Chr.11: 57660876-57660876 on Build GRCh38
rs764684265 Chr.11: 57661057-57661057 on Build GRCh38
rs746040503 Chr.11: 57661020-57661020 on Build GRCh38
rs775322826 Chr.11: 57661034-57661034 on Build GRCh38
rs767902920 Chr.11: 57660791-57660791 on Build GRCh38
rs111754731 Chr.11: 57661109-57661109 on Build GRCh38
rs892919486 Chr.11: 57660848-57660848 on Build GRCh38
rs746135188 Chr.11: 57661149-57661149 on Build GRCh38
rs777777223 Chr.11: 57660991-57660991 on Build GRCh38
rs1008640713 Chr.11: 57660813-57660813 on Build GRCh38
rs769813236 Chr.11: 57660918-57660920 on Build GRCh38
rs952926202 Chr.11: 57660942-57660942 on Build GRCh38
rs147951568 Chr.11: 57660808-57660808 on Build GRCh38
rs543506228 Chr.11: 57660880-57660880 on Build GRCh38
rs145579673 Chr.11: 57661106-57661106 on Build GRCh38
rs758021571 Chr.11: 57661063-57661063 on Build GRCh38
rs757438894 Chr.11: 57661129-57661129 on Build GRCh38
rs750096202 Chr.11: 57661062-57661062 on Build GRCh38
rs982882973 Chr.11: 57660943-57660943 on Build GRCh38
rs776580990 Chr.11: 57661212-57661212 on Build GRCh38
rs773313264 Chr.11: 57660875-57660875 on Build GRCh38
rs140965892 Chr.11: 57660965-57660965 on Build GRCh38
rs539245351 Chr.11: 57660981-57660981 on Build GRCh38
rs964253837 Chr.11: 57660939-57660939 on Build GRCh38
rs767305155 Chr.11: 57660911-57660911 on Build GRCh38
rs1007335389 Chr.11: 57660917-57660917 on Build GRCh38
rs768946938 Chr.11: 57660823-57660823 on Build GRCh38
rs772331151 Chr.11: 57661156-57661156 on Build GRCh38
rs753383188 Chr.11: 57660949-57660949 on Build GRCh38
rs200588495 Chr.11: 57660969-57660969 on Build GRCh38
rs17152015 Chr.11: 57660956-57660956 on Build GRCh38
rs771813411 Chr.11: 57661021-57661021 on Build GRCh38
rs748646139 Chr.11: 57660836-57660836 on Build GRCh38
rs775202618 Chr.11: 57661179-57661179 on Build GRCh38
rs747360654 Chr.11: 57660821-57660821 on Build GRCh38
rs752599991 Chr.11: 57660914-57660914 on Build GRCh38
rs775609000 Chr.11: 57661034-57661034 on Build GRCh38
rs150146614 Chr.11: 57661056-57661056 on Build GRCh38
rs774661332 Chr.11: 57660894-57660894 on Build GRCh38
rs778348932 Chr.11: 57661111-57661111 on Build GRCh38
rs749776228 Chr.11: 57661128-57661128 on Build GRCh38
rs141596718 Chr.11: 57660893-57660893 on Build GRCh38
rs200917931 Chr.11: 57660990-57660990 on Build GRCh38
rs375721461 Chr.11: 57660825-57660825 on Build GRCh38
rs764078062 Chr.11: 57661035-57661035 on Build GRCh38
rs763551843 Chr.11: 57660947-57660947 on Build GRCh38
rs771068295 Chr.11: 57661011-57661011 on Build GRCh38
rs779116291 Chr.11: 57661130-57661130 on Build GRCh38
rs768269102 Chr.11: 57661192-57661192 on Build GRCh38
rs372647012 Chr.11: 57660814-57660814 on Build GRCh38
rs370085533 Chr.11: 57660802-57660802 on Build GRCh38
rs781569229 Chr.11: 57660971-57660971 on Build GRCh38
rs774746868 Chr.11: 57661019-57661019 on Build GRCh38
rs759409566 Chr.11: 57660898-57660898 on Build GRCh38
rs762862019 Chr.11: 57661048-57661052 on Build GRCh38
rs776492811 Chr.11: 57661045-57661045 on Build GRCh38
rs767839746 Chr.11: 57661077-57661077 on Build GRCh38
rs749396873 Chr.11: 57661010-57661010 on Build GRCh38
rs768698651 Chr.11: 57661192-57661193 on Build GRCh38
rs951442414 Chr.11: 57660968-57660968 on Build GRCh38
rs754578147 Chr.11: 57661083-57661083 on Build GRCh38
rs78489374 Chr.11: 57660838-57660838 on Build GRCh38
rs746830893 Chr.11: 57661190-57661190 on Build GRCh38

View Full Product Details