Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 59148850-59149370 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM111A
Entrez Gene ID
63901
Gene Name
family with sequence similarity 111 member A
Gene Aliases
GCLEB, KCS2
UniGene
Hs.150651
Gene Chromosome Location
Chr.11: 59142746-59155039 on Build GRCh38
Transcripts
NM_001142519.2 NM_001142520.2 NM_001142521.2 NM_001312909.1 NM_001312910.1 NM_001312911.1 NM_022074.3 NM_198847.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5050929 c.19A>G p.R7G Chr.11: 59148891-59148891 on Build GRCh38
1206021 c.30G>T p.K10N Chr.11: 59148902-59148902 on Build GRCh38
6461899 c.77C>T p.S26F Chr.11: 59148949-59148949 on Build GRCh38
928965 c.63C>T p.I21I Chr.11: 59148935-59148935 on Build GRCh38
4682378 c.25C>T p.R9W Chr.11: 59148897-59148897 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777428096 Chr.11: 59148963-59148963 on Build GRCh38
rs778533177 Chr.11: 59148976-59148976 on Build GRCh38
rs777189581 Chr.11: 59148902-59148902 on Build GRCh38
rs751507334 Chr.11: 59149073-59149073 on Build GRCh38
rs1050929157 Chr.11: 59149255-59149255 on Build GRCh38
rs377285829 Chr.11: 59148958-59148958 on Build GRCh38
rs940705195 Chr.11: 59149061-59149061 on Build GRCh38
rs911861251 Chr.11: 59149058-59149059 on Build GRCh38
rs184058547 Chr.11: 59149105-59149105 on Build GRCh38
rs748812251 Chr.11: 59148880-59148880 on Build GRCh38
rs619509 Chr.11: 59149205-59149205 on Build GRCh38
rs956195934 Chr.11: 59149150-59149150 on Build GRCh38
rs553507586 Chr.11: 59148876-59148876 on Build GRCh38
rs1004270053 Chr.11: 59149032-59149032 on Build GRCh38
rs762118689 Chr.11: 59148913-59148913 on Build GRCh38
rs147786828 Chr.11: 59148935-59148935 on Build GRCh38
rs142539088 Chr.11: 59148952-59148952 on Build GRCh38
rs11229847 Chr.11: 59149122-59149122 on Build GRCh38
rs369821333 Chr.11: 59149230-59149230 on Build GRCh38
rs796750211 Chr.11: 59149255-59149255 on Build GRCh38
rs558415069 Chr.11: 59149107-59149107 on Build GRCh38
rs996690957 Chr.11: 59148954-59148954 on Build GRCh38
rs772579967 Chr.11: 59148882-59148882 on Build GRCh38
rs571420431 Chr.11: 59149331-59149331 on Build GRCh38
rs769268368 Chr.11: 59148994-59148994 on Build GRCh38
rs766524323 Chr.11: 59148939-59148939 on Build GRCh38
rs906177013 Chr.11: 59149226-59149226 on Build GRCh38
rs561606923 Chr.11: 59149225-59149225 on Build GRCh38
rs576470299 Chr.11: 59149217-59149217 on Build GRCh38
rs77837887 Chr.11: 59149095-59149095 on Build GRCh38
rs61083600 Chr.11: 59149246-59149246 on Build GRCh38
rs758432983 Chr.11: 59148897-59148897 on Build GRCh38
rs192882280 Chr.11: 59149097-59149097 on Build GRCh38
rs1011967634 Chr.11: 59149221-59149221 on Build GRCh38
rs775891705 Chr.11: 59148894-59148894 on Build GRCh38
rs958555400 Chr.11: 59149271-59149271 on Build GRCh38
rs577003143 Chr.11: 59149112-59149112 on Build GRCh38
rs527651045 Chr.11: 59149320-59149320 on Build GRCh38
rs757684482 Chr.11: 59149241-59149241 on Build GRCh38
rs11229845 Chr.11: 59149021-59149021 on Build GRCh38
rs764343220 Chr.11: 59148898-59148898 on Build GRCh38
rs946409431 Chr.11: 59149188-59149188 on Build GRCh38
rs890992908 Chr.11: 59149281-59149281 on Build GRCh38
rs531657976 Chr.11: 59149264-59149264 on Build GRCh38
rs780840101 Chr.11: 59149275-59149275 on Build GRCh38
rs550142140 Chr.11: 59149307-59149307 on Build GRCh38
rs747457670 Chr.11: 59148984-59148984 on Build GRCh38
rs748789460 Chr.11: 59148973-59148973 on Build GRCh38
rs747211416 Chr.11: 59148953-59148953 on Build GRCh38
rs762977097 Chr.11: 59148936-59148936 on Build GRCh38
rs913650218 Chr.11: 59149183-59149183 on Build GRCh38
rs752494544 Chr.11: 59148955-59148955 on Build GRCh38
rs565541701 Chr.11: 59148957-59148957 on Build GRCh38
rs574529384 Chr.11: 59149046-59149046 on Build GRCh38
rs988321519 Chr.11: 59149172-59149172 on Build GRCh38
rs928780848 Chr.11: 59149139-59149139 on Build GRCh38
rs79009474 Chr.11: 59149222-59149222 on Build GRCh38
rs751433614 Chr.11: 59148951-59148951 on Build GRCh38
rs149259997 Chr.11: 59149093-59149093 on Build GRCh38

View Full Product Details