Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 61007578-61008081 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD6
Entrez Gene ID
923
Gene Name
CD6 molecule
Gene Aliases
TP120
UniGene
Hs.744366
Gene Chromosome Location
Chr.11: 60971641-61020377 on Build GRCh38
Transcripts
NM_001254750.1 NM_001254751.1 NM_006725.4 NR_045638.1 XM_006718738.1 XM_006718739.1 XM_006718740.1 XM_006718741.1 XM_011545360.1 XM_011545362.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1742961 c.259G>A p.G87S Chr.11: 61007700-61007700 on Build GRCh38
5033929 c.301A>C p.T101P Chr.11: 61007742-61007742 on Build GRCh38
6501032 c.268G>A p.G90R Chr.11: 61007709-61007709 on Build GRCh38
4802177 c.171G>C p.T57T Chr.11: 61007612-61007612 on Build GRCh38
4993594 c.310_311ins15 p.L104>HAAP* Chr.11: 61007751-61007752 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs922676046 Chr.11: 61007741-61007741 on Build GRCh38
rs908359920 Chr.11: 61007969-61007969 on Build GRCh38
rs536627807 Chr.11: 61007751-61007751 on Build GRCh38
rs1047209045 Chr.11: 61007942-61007942 on Build GRCh38
rs971132060 Chr.11: 61007805-61007805 on Build GRCh38
rs1011394472 Chr.11: 61007630-61007630 on Build GRCh38
rs1038626418 Chr.11: 61008038-61008038 on Build GRCh38
rs924114067 Chr.11: 61007632-61007632 on Build GRCh38
rs989718758 Chr.11: 61007645-61007645 on Build GRCh38
rs757974950 Chr.11: 61007614-61007614 on Build GRCh38
rs772074450 Chr.11: 61007771-61007771 on Build GRCh38
rs1011744720 Chr.11: 61007977-61007977 on Build GRCh38
rs942692563 Chr.11: 61007671-61007671 on Build GRCh38
rs986688315 Chr.11: 61007660-61007660 on Build GRCh38
rs887931049 Chr.11: 61007742-61007742 on Build GRCh38
rs998556221 Chr.11: 61007603-61007603 on Build GRCh38
rs972734137 Chr.11: 61007699-61007699 on Build GRCh38
rs902807224 Chr.11: 61007829-61007829 on Build GRCh38
rs956892375 Chr.11: 61007824-61007824 on Build GRCh38
rs1012557482 Chr.11: 61007790-61007790 on Build GRCh38
rs896768411 Chr.11: 61007628-61007628 on Build GRCh38
rs760617231 Chr.11: 61007831-61007831 on Build GRCh38
rs752014931 Chr.11: 61007881-61007881 on Build GRCh38
rs576602370 Chr.11: 61007757-61007757 on Build GRCh38
rs750025413 Chr.11: 61007889-61007889 on Build GRCh38
rs750009088 Chr.11: 61007613-61007613 on Build GRCh38
rs778388668 Chr.11: 61007742-61007743 on Build GRCh38
rs1047913038 Chr.11: 61007738-61007738 on Build GRCh38
rs572519877 Chr.11: 61007920-61007920 on Build GRCh38
rs1017283902 Chr.11: 61007921-61007921 on Build GRCh38
rs1038320908 Chr.11: 61007764-61007764 on Build GRCh38
rs139918339 Chr.11: 61007755-61007755 on Build GRCh38
rs1005461801 Chr.11: 61007762-61007762 on Build GRCh38
rs541743404 Chr.11: 61007940-61007940 on Build GRCh38
rs781764855 Chr.11: 61007624-61007624 on Build GRCh38
rs771235467 Chr.11: 61007708-61007708 on Build GRCh38
rs1031422326 Chr.11: 61007604-61007604 on Build GRCh38
rs975398455 Chr.11: 61007915-61007915 on Build GRCh38
rs748604659 Chr.11: 61007651-61007651 on Build GRCh38
rs920011640 Chr.11: 61007740-61007740 on Build GRCh38
rs775422862 Chr.11: 61007816-61007816 on Build GRCh38
rs915466416 Chr.11: 61007882-61007882 on Build GRCh38
rs575606847 Chr.11: 61007681-61007681 on Build GRCh38
rs141701259 Chr.11: 61007774-61007774 on Build GRCh38
rs929546008 Chr.11: 61007705-61007705 on Build GRCh38
rs879906657 Chr.11: 61007709-61007709 on Build GRCh38
rs1024399865 Chr.11: 61007802-61007802 on Build GRCh38
rs761496891 Chr.11: 61007873-61007873 on Build GRCh38
rs941151250 Chr.11: 61008022-61008022 on Build GRCh38
rs146243965 Chr.11: 61007957-61007957 on Build GRCh38
rs931401022 Chr.11: 61007743-61007743 on Build GRCh38
rs1028124038 Chr.11: 61007984-61007984 on Build GRCh38
rs763829539 Chr.11: 61007844-61007844 on Build GRCh38
rs776532953 Chr.11: 61007852-61007852 on Build GRCh38
rs998601429 Chr.11: 61007807-61007807 on Build GRCh38
rs536765881 Chr.11: 61007886-61007886 on Build GRCh38
rs934070374 Chr.11: 61007750-61007750 on Build GRCh38
rs956944329 Chr.11: 61007640-61007640 on Build GRCh38
rs756455957 Chr.11: 61007670-61007670 on Build GRCh38
rs749584244 Chr.11: 61007690-61007690 on Build GRCh38
rs989625260 Chr.11: 61007834-61007834 on Build GRCh38
rs934053429 Chr.11: 61007935-61007935 on Build GRCh38

View Full Product Details