Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 61010930-61011456 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD6
Entrez Gene ID
923
Gene Name
CD6 molecule
Gene Aliases
TP120
UniGene
Hs.744366
Gene Chromosome Location
Chr.11: 60971641-61020377 on Build GRCh38
Transcripts
NM_001254750.1 NM_001254751.1 NM_006725.4 NR_045638.1 XM_006718738.1 XM_006718739.1 XM_006718740.1 XM_006718741.1 XM_011545360.1 XM_011545362.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
929351 c.1099C>T p.H367Y Chr.11: 61011084-61011084 on Build GRCh38
294223 c.1150G>T p.E384* Chr.11: 61011135-61011135 on Build GRCh38
1237195 c.1147A>G p.I383V Chr.11: 61011132-61011132 on Build GRCh38
2036882 c.1125T>C p.P375P Chr.11: 61011110-61011110 on Build GRCh38
6292868 c.1109C>A p.S370Y Chr.11: 61011094-61011094 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs944492588 Chr.11: 61010978-61010978 on Build GRCh38
rs377541031 Chr.11: 61011055-61011055 on Build GRCh38
rs1033648324 Chr.11: 61011341-61011341 on Build GRCh38
rs776735105 Chr.11: 61011033-61011033 on Build GRCh38
rs748002885 Chr.11: 61011177-61011178 on Build GRCh38
rs539122937 Chr.11: 61011284-61011284 on Build GRCh38
rs570515725 Chr.11: 61011282-61011282 on Build GRCh38
rs774259594 Chr.11: 61011157-61011170 on Build GRCh38
rs150601725 Chr.11: 61011412-61011412 on Build GRCh38
rs796569535 Chr.11: 61011008-61011008 on Build GRCh38
rs972173466 Chr.11: 61011427-61011427 on Build GRCh38
rs1014342980 Chr.11: 61011092-61011092 on Build GRCh38
rs575200956 Chr.11: 61011160-61011160 on Build GRCh38
rs1034181176 Chr.11: 61011228-61011228 on Build GRCh38
rs557000218 Chr.11: 61011029-61011029 on Build GRCh38
rs780714674 Chr.11: 61011102-61011102 on Build GRCh38
rs766369636 Chr.11: 61011059-61011059 on Build GRCh38
rs971890276 Chr.11: 61011230-61011230 on Build GRCh38
rs770940045 Chr.11: 61011149-61011149 on Build GRCh38
rs185793883 Chr.11: 61011388-61011388 on Build GRCh38
rs550448976 Chr.11: 61011281-61011281 on Build GRCh38
rs752049647 Chr.11: 61011179-61011180 on Build GRCh38
rs759429483 Chr.11: 61011155-61011155 on Build GRCh38
rs866822757 Chr.11: 61011190-61011191 on Build GRCh38
rs986089293 Chr.11: 61011387-61011387 on Build GRCh38
rs535377510 Chr.11: 61011337-61011337 on Build GRCh38
rs536760442 Chr.11: 61011010-61011010 on Build GRCh38
rs763058341 Chr.11: 61011207-61011207 on Build GRCh38
rs552754660 Chr.11: 61011295-61011295 on Build GRCh38
rs375096109 Chr.11: 61011258-61011258 on Build GRCh38
rs781647415 Chr.11: 61011115-61011115 on Build GRCh38
rs897708309 Chr.11: 61010982-61010982 on Build GRCh38
rs986236329 Chr.11: 61010951-61010951 on Build GRCh38
rs771906206 Chr.11: 61011076-61011076 on Build GRCh38
rs568094929 Chr.11: 61011255-61011255 on Build GRCh38
rs138880843 Chr.11: 61011178-61011178 on Build GRCh38
rs536114927 Chr.11: 61010955-61010955 on Build GRCh38
rs945017940 Chr.11: 61011250-61011250 on Build GRCh38
rs900338210 Chr.11: 61011298-61011298 on Build GRCh38
rs374328736 Chr.11: 61011131-61011131 on Build GRCh38
rs749372795 Chr.11: 61011133-61011133 on Build GRCh38
rs752057899 Chr.11: 61011105-61011105 on Build GRCh38
rs184272477 Chr.11: 61011215-61011215 on Build GRCh38
rs913524276 Chr.11: 61011247-61011247 on Build GRCh38
rs150035626 Chr.11: 61011178-61011179 on Build GRCh38
rs533408206 Chr.11: 61011186-61011186 on Build GRCh38
rs10528068 Chr.11: 61011204-61011205 on Build GRCh38
rs370567112 Chr.11: 61011027-61011027 on Build GRCh38
rs930565578 Chr.11: 61011236-61011236 on Build GRCh38
rs190800807 Chr.11: 61011003-61011003 on Build GRCh38
rs868736558 Chr.11: 61011203-61011204 on Build GRCh38
rs966111878 Chr.11: 61011402-61011402 on Build GRCh38
rs1038862940 Chr.11: 61011021-61011021 on Build GRCh38
rs536857558 Chr.11: 61011409-61011409 on Build GRCh38
rs771872898 Chr.11: 61011157-61011157 on Build GRCh38
rs757711560 Chr.11: 61011181-61011182 on Build GRCh38
rs1020448452 Chr.11: 61011119-61011119 on Build GRCh38
rs753448654 Chr.11: 61011096-61011096 on Build GRCh38
rs755831022 Chr.11: 61011087-61011087 on Build GRCh38
rs768629911 Chr.11: 61011143-61011143 on Build GRCh38
rs937650810 Chr.11: 61011259-61011259 on Build GRCh38
rs555227028 Chr.11: 61011374-61011374 on Build GRCh38
rs192304573 Chr.11: 61011101-61011101 on Build GRCh38
rs936427834 Chr.11: 61011291-61011291 on Build GRCh38
rs774292537 Chr.11: 61011153-61011153 on Build GRCh38
rs371627356 Chr.11: 61011159-61011159 on Build GRCh38
rs1034708421 Chr.11: 61011070-61011070 on Build GRCh38
rs1018997285 Chr.11: 61011399-61011399 on Build GRCh38
rs112163504 Chr.11: 61011192-61011193 on Build GRCh38
rs919081123 Chr.11: 61011434-61011434 on Build GRCh38
rs761426204 Chr.11: 61011166-61011166 on Build GRCh38
rs3220042 Chr.11: 61011180-61011181 on Build GRCh38
rs763634669 Chr.11: 61011091-61011091 on Build GRCh38
rs776455161 Chr.11: 61011433-61011433 on Build GRCh38
rs774206247 Chr.11: 61011065-61011065 on Build GRCh38
rs74860834 Chr.11: 61011041-61011041 on Build GRCh38
rs564584694 Chr.11: 61011179-61011179 on Build GRCh38
rs145393446 Chr.11: 61011075-61011075 on Build GRCh38
rs749358989 Chr.11: 61011120-61011123 on Build GRCh38
rs1036203128 Chr.11: 61010981-61010981 on Build GRCh38
rs748436641 Chr.11: 61011122-61011122 on Build GRCh38
rs368349001 Chr.11: 61011242-61011242 on Build GRCh38
rs112209800 Chr.11: 61011232-61011232 on Build GRCh38
rs73478336 Chr.11: 61011246-61011246 on Build GRCh38
rs111475000 Chr.11: 61010977-61010977 on Build GRCh38
rs749917202 Chr.11: 61011175-61011175 on Build GRCh38
rs113542558 Chr.11: 61011072-61011305 on Build GRCh38
rs117785308 Chr.11: 61011158-61011158 on Build GRCh38
rs541677968 Chr.11: 61011123-61011123 on Build GRCh38
rs188333072 Chr.11: 61011331-61011331 on Build GRCh38
rs755421143 Chr.11: 61011112-61011112 on Build GRCh38
rs748358126 Chr.11: 61011035-61011035 on Build GRCh38
rs903631493 Chr.11: 61010998-61010998 on Build GRCh38
rs539615034 Chr.11: 61011054-61011054 on Build GRCh38
rs181148801 Chr.11: 61011383-61011383 on Build GRCh38
rs980241559 Chr.11: 61011229-61011229 on Build GRCh38
rs1048717392 Chr.11: 61011038-61011038 on Build GRCh38
rs752324073 Chr.11: 61011077-61011077 on Build GRCh38

View Full Product Details