Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 64859982-64860426 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EHD1
Entrez Gene ID
10938
Gene Name
EH domain containing 1
Gene Aliases
H-PAST, HPAST1, PAST, PAST1
UniGene
Hs.523774
Gene Chromosome Location
Chr.11: 64852727-64879713 on Build GRCh38
Transcripts
NM_001282444.1 NM_001282445.1 NM_006795.3 XM_011544739.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
429569 c.588C>T p.I196I Chr.11: 64860251-64860251 on Build GRCh38
5530410 c.549C>T p.D183D Chr.11: 64860290-64860290 on Build GRCh38
6014488 c.643C>T p.R215C Chr.11: 64860196-64860196 on Build GRCh38
4035278 c.602C>T p.S201L Chr.11: 64860237-64860237 on Build GRCh38
1355834 c.724A>G p.K242E Chr.11: 64860115-64860115 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1056427086 Chr.11: 64860386-64860386 on Build GRCh38
rs749849055 Chr.11: 64860339-64860340 on Build GRCh38
rs1017807009 Chr.11: 64860316-64860316 on Build GRCh38
rs138422263 Chr.11: 64860245-64860245 on Build GRCh38
rs375175532 Chr.11: 64860339-64860339 on Build GRCh38
rs1016799349 Chr.11: 64860263-64860263 on Build GRCh38
rs761790920 Chr.11: 64860056-64860056 on Build GRCh38
rs766624042 Chr.11: 64860380-64860380 on Build GRCh38
rs938959498 Chr.11: 64860377-64860377 on Build GRCh38
rs772990090 Chr.11: 64860058-64860058 on Build GRCh38
rs142323286 Chr.11: 64860145-64860145 on Build GRCh38
rs758729682 Chr.11: 64860036-64860036 on Build GRCh38
rs370296006 Chr.11: 64860233-64860233 on Build GRCh38
rs374651114 Chr.11: 64860166-64860166 on Build GRCh38
rs199636551 Chr.11: 64860160-64860160 on Build GRCh38
rs61745217 Chr.11: 64860005-64860005 on Build GRCh38
rs374259413 Chr.11: 64860098-64860098 on Build GRCh38
rs746917690 Chr.11: 64860244-64860244 on Build GRCh38
rs766122136 Chr.11: 64860148-64860148 on Build GRCh38
rs144914499 Chr.11: 64860320-64860320 on Build GRCh38
rs936411919 Chr.11: 64860118-64860118 on Build GRCh38
rs773078280 Chr.11: 64860140-64860140 on Build GRCh38
rs765330673 Chr.11: 64860076-64860076 on Build GRCh38
rs758930905 Chr.11: 64860078-64860078 on Build GRCh38
rs376041030 Chr.11: 64860046-64860046 on Build GRCh38
rs748144371 Chr.11: 64860029-64860029 on Build GRCh38
rs77426065 Chr.11: 64860280-64860280 on Build GRCh38
rs767059988 Chr.11: 64860338-64860339 on Build GRCh38
rs71581707 Chr.11: 64860340-64860340 on Build GRCh38
rs150536999 Chr.11: 64860201-64860201 on Build GRCh38
rs754658374 Chr.11: 64860229-64860229 on Build GRCh38
rs145274478 Chr.11: 64860037-64860037 on Build GRCh38
rs372642668 Chr.11: 64860116-64860116 on Build GRCh38
rs757723688 Chr.11: 64860209-64860209 on Build GRCh38
rs772683924 Chr.11: 64860162-64860162 on Build GRCh38
rs941880771 Chr.11: 64860397-64860397 on Build GRCh38
rs987867025 Chr.11: 64860345-64860345 on Build GRCh38
rs748105429 Chr.11: 64860117-64860117 on Build GRCh38
rs748757800 Chr.11: 64860047-64860047 on Build GRCh38
rs184690603 Chr.11: 64860197-64860197 on Build GRCh38
rs753192607 Chr.11: 64860077-64860077 on Build GRCh38
rs576058223 Chr.11: 64860297-64860297 on Build GRCh38
rs61738612 Chr.11: 64860020-64860020 on Build GRCh38
rs759743110 Chr.11: 64860251-64860251 on Build GRCh38
rs541250479 Chr.11: 64860394-64860394 on Build GRCh38
rs374683879 Chr.11: 64860248-64860248 on Build GRCh38
rs778347400 Chr.11: 64860092-64860092 on Build GRCh38
rs536317553 Chr.11: 64860257-64860257 on Build GRCh38
rs989198708 Chr.11: 64860127-64860127 on Build GRCh38
rs897861323 Chr.11: 64860295-64860295 on Build GRCh38
rs534199009 Chr.11: 64860368-64860368 on Build GRCh38
rs137874978 Chr.11: 64860057-64860057 on Build GRCh38
rs764527030 Chr.11: 64860195-64860195 on Build GRCh38
rs752061690 Chr.11: 64860205-64860205 on Build GRCh38
rs913639525 Chr.11: 64860333-64860333 on Build GRCh38
rs764227474 Chr.11: 64860272-64860272 on Build GRCh38
rs775515682 Chr.11: 64860269-64860269 on Build GRCh38
rs927587406 Chr.11: 64860358-64860358 on Build GRCh38
rs910736633 Chr.11: 64860043-64860043 on Build GRCh38
rs571846024 Chr.11: 64860044-64860044 on Build GRCh38
rs368130910 Chr.11: 64860196-64860196 on Build GRCh38
rs375914175 Chr.11: 64860314-64860314 on Build GRCh38
rs763144350 Chr.11: 64860271-64860271 on Build GRCh38
rs755044438 Chr.11: 64860074-64860074 on Build GRCh38
rs777243803 Chr.11: 64860319-64860319 on Build GRCh38
rs772003242 Chr.11: 64860139-64860139 on Build GRCh38
rs753495293 Chr.11: 64860223-64860223 on Build GRCh38
rs774181653 Chr.11: 64860055-64860055 on Build GRCh38
rs775809648 Chr.11: 64860341-64860341 on Build GRCh38
rs774571412 Chr.11: 64860357-64860357 on Build GRCh38
rs372156615 Chr.11: 64860367-64860367 on Build GRCh38
rs763231410 Chr.11: 64860347-64860347 on Build GRCh38
rs938894652 Chr.11: 64860169-64860169 on Build GRCh38
rs776759353 Chr.11: 64860161-64860161 on Build GRCh38
rs774171970 Chr.11: 64860098-64860098 on Build GRCh38
rs71581721 Chr.11: 64860023-64860023 on Build GRCh38
rs781659919 Chr.11: 64860214-64860214 on Build GRCh38
rs367741962 Chr.11: 64860302-64860302 on Build GRCh38
rs771522896 Chr.11: 64860242-64860242 on Build GRCh38
rs747628133 Chr.11: 64860288-64860288 on Build GRCh38
rs368895974 Chr.11: 64860107-64860107 on Build GRCh38
rs748926280 Chr.11: 64860102-64860102 on Build GRCh38
rs745572506 Chr.11: 64860039-64860039 on Build GRCh38
rs144337563 Chr.11: 64860289-64860289 on Build GRCh38
rs766603148 Chr.11: 64860070-64860070 on Build GRCh38
rs761920706 Chr.11: 64860366-64860366 on Build GRCh38
rs761134372 Chr.11: 64860369-64860369 on Build GRCh38
rs761517796 Chr.11: 64860304-64860310 on Build GRCh38
rs547652824 Chr.11: 64860101-64860101 on Build GRCh38
rs768903050 Chr.11: 64860356-64860356 on Build GRCh38
rs377293606 Chr.11: 64860167-64860167 on Build GRCh38
rs754012326 Chr.11: 64860073-64860073 on Build GRCh38
rs760398339 Chr.11: 64860065-64860065 on Build GRCh38
rs148805183 Chr.11: 64860305-64860305 on Build GRCh38
rs372656658 Chr.11: 64860304-64860304 on Build GRCh38
rs11545093 Chr.11: 64860041-64860041 on Build GRCh38
rs747711359 Chr.11: 64860236-64860236 on Build GRCh38
rs764708094 Chr.11: 64860204-64860204 on Build GRCh38
rs370682065 Chr.11: 64860336-64860336 on Build GRCh38
rs768160178 Chr.11: 64860053-64860053 on Build GRCh38
rs147528048 Chr.11: 64860329-64860329 on Build GRCh38

View Full Product Details