Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 55349620-55350183 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COX6B2
Entrez Gene ID
125965
Gene Name
cytochrome c oxidase subunit 6B2
Gene Aliases
COXVIB2, CT59
UniGene
Hs.550544
Gene Chromosome Location
Chr.19: 55349702-55354814 on Build GRCh38
Transcripts
NM_144613.4

SNP Detail

SNP ID Chromosome Location
rs765913278 Chr.19: 55350017-55350017 on Build GRCh38
rs913466433 Chr.19: 55349662-55349662 on Build GRCh38
rs931814425 Chr.19: 55350030-55350030 on Build GRCh38
rs935031900 Chr.19: 55350147-55350147 on Build GRCh38
rs142583946 Chr.19: 55350130-55350130 on Build GRCh38
rs919952939 Chr.19: 55349787-55349787 on Build GRCh38
rs928728511 Chr.19: 55349760-55349760 on Build GRCh38
rs1004726306 Chr.19: 55349834-55349834 on Build GRCh38
rs894892000 Chr.19: 55349738-55349738 on Build GRCh38
rs1040192375 Chr.19: 55349843-55349843 on Build GRCh38
rs957283111 Chr.19: 55349887-55349887 on Build GRCh38
rs199726415 Chr.19: 55350136-55350136 on Build GRCh38
rs928780853 Chr.19: 55349955-55349955 on Build GRCh38
rs867066810 Chr.19: 55350161-55350161 on Build GRCh38
rs1055814273 Chr.19: 55349778-55349778 on Build GRCh38
rs976019960 Chr.19: 55350154-55350154 on Build GRCh38
rs967555989 Chr.19: 55349664-55349664 on Build GRCh38
rs35958433 Chr.19: 55350032-55350033 on Build GRCh38
rs981565320 Chr.19: 55349676-55349676 on Build GRCh38
rs755070147 Chr.19: 55349795-55349797 on Build GRCh38
rs1043458179 Chr.19: 55349983-55349988 on Build GRCh38
rs928185544 Chr.19: 55349943-55349943 on Build GRCh38
rs958890389 Chr.19: 55349956-55349956 on Build GRCh38
rs983451182 Chr.19: 55350105-55350105 on Build GRCh38
rs944005318 Chr.19: 55350142-55350142 on Build GRCh38
rs1022385389 Chr.19: 55349772-55349772 on Build GRCh38
rs572818822 Chr.19: 55349954-55349954 on Build GRCh38
rs962552970 Chr.19: 55350049-55350049 on Build GRCh38
rs999867241 Chr.19: 55349805-55349805 on Build GRCh38
rs1018066585 Chr.19: 55350038-55350038 on Build GRCh38
rs187057939 Chr.19: 55349911-55349911 on Build GRCh38
rs1040243402 Chr.19: 55350015-55350015 on Build GRCh38
rs183306970 Chr.19: 55349891-55349891 on Build GRCh38
rs996299741 Chr.19: 55349865-55349865 on Build GRCh38
rs554721425 Chr.19: 55349806-55349806 on Build GRCh38
rs371298511 Chr.19: 55349986-55349991 on Build GRCh38
rs892876070 Chr.19: 55350048-55350048 on Build GRCh38
rs1029557801 Chr.19: 55350002-55350002 on Build GRCh38
rs990834503 Chr.19: 55349640-55349640 on Build GRCh38
rs982798957 Chr.19: 55350133-55350133 on Build GRCh38
rs983735099 Chr.19: 55349936-55349936 on Build GRCh38
rs916752522 Chr.19: 55350130-55350131 on Build GRCh38
rs931312454 Chr.19: 55349797-55349797 on Build GRCh38
rs902189856 Chr.19: 55349995-55349995 on Build GRCh38
rs80037506 Chr.19: 55350128-55350128 on Build GRCh38
rs56986472 Chr.19: 55350136-55350136 on Build GRCh38
rs937401786 Chr.19: 55349770-55349770 on Build GRCh38
rs552747755 Chr.19: 55349945-55349945 on Build GRCh38
rs867965191 Chr.19: 55350051-55350051 on Build GRCh38
rs1047986883 Chr.19: 55349646-55349646 on Build GRCh38
rs538080455 Chr.19: 55349791-55349791 on Build GRCh38
rs368147583 Chr.19: 55350139-55350141 on Build GRCh38
rs536398146 Chr.19: 55350019-55350019 on Build GRCh38
rs201225343 Chr.19: 55350127-55350128 on Build GRCh38
rs1055289709 Chr.19: 55349950-55349950 on Build GRCh38
rs903460685 Chr.19: 55350071-55350071 on Build GRCh38
rs886348180 Chr.19: 55349813-55349813 on Build GRCh38
rs371848368 Chr.19: 55350052-55350052 on Build GRCh38
rs761307209 Chr.19: 55349704-55349704 on Build GRCh38
rs187577008 Chr.19: 55349997-55349997 on Build GRCh38
rs1028727962 Chr.19: 55349866-55349866 on Build GRCh38
rs754281348 Chr.19: 55349849-55349849 on Build GRCh38
rs544986783 Chr.19: 55349973-55349973 on Build GRCh38
rs965166351 Chr.19: 55349877-55349877 on Build GRCh38
rs534007840 Chr.19: 55350137-55350139 on Build GRCh38
rs967567206 Chr.19: 55349946-55349946 on Build GRCh38
rs529811887 Chr.19: 55350082-55350082 on Build GRCh38
rs991666173 Chr.19: 55350114-55350114 on Build GRCh38
rs923208816 Chr.19: 55350157-55350157 on Build GRCh38
rs1049048085 Chr.19: 55349812-55349812 on Build GRCh38
rs928232977 Chr.19: 55350110-55350110 on Build GRCh38
rs574837115 Chr.19: 55349960-55349960 on Build GRCh38
rs751028048 Chr.19: 55350123-55350123 on Build GRCh38
rs1050637402 Chr.19: 55350042-55350042 on Build GRCh38
rs768372124 Chr.19: 55349966-55349966 on Build GRCh38
rs764660833 Chr.19: 55349821-55349821 on Build GRCh38
rs931340639 Chr.19: 55349987-55349987 on Build GRCh38
rs959169158 Chr.19: 55350100-55350100 on Build GRCh38
rs1054907303 Chr.19: 55349731-55349731 on Build GRCh38
rs569171583 Chr.19: 55349641-55349641 on Build GRCh38
rs769542725 Chr.19: 55350043-55350043 on Build GRCh38
rs184097500 Chr.19: 55349991-55349991 on Build GRCh38
rs929297032 Chr.19: 55349926-55349926 on Build GRCh38
rs147585942 Chr.19: 55350056-55350056 on Build GRCh38
rs561129563 Chr.19: 55350081-55350081 on Build GRCh38
rs762168050 Chr.19: 55349858-55349858 on Build GRCh38
rs191810490 Chr.19: 55349988-55349988 on Build GRCh38

View Full Product Details