Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 31373416-31373827 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DEFB118
Entrez Gene ID
117285
Gene Name
defensin beta 118
Gene Aliases
C20orf63, DEFB-18, ESC42, ESP13.6
UniGene
Hs.274124
Gene Chromosome Location
Chr.20: 31368618-31373923 on Build GRCh38
Transcripts
NM_054112.2

SNP Detail

SNP ID Chromosome Location
rs1045305517 Chr.20: 31373773-31373773 on Build GRCh38
rs45452494 Chr.20: 31373784-31373784 on Build GRCh38
rs6119976 Chr.20: 31373672-31373672 on Build GRCh38
rs1013254112 Chr.20: 31373554-31373554 on Build GRCh38
rs931078264 Chr.20: 31373592-31373592 on Build GRCh38
rs965685591 Chr.20: 31373801-31373801 on Build GRCh38
rs979617313 Chr.20: 31373583-31373583 on Build GRCh38
rs905189894 Chr.20: 31373745-31373745 on Build GRCh38
rs920909569 Chr.20: 31373550-31373550 on Build GRCh38
rs142733756 Chr.20: 31373590-31373590 on Build GRCh38
rs956555041 Chr.20: 31373610-31373610 on Build GRCh38
rs538948407 Chr.20: 31373746-31373746 on Build GRCh38
rs745501948 Chr.20: 31373499-31373499 on Build GRCh38
rs577247336 Chr.20: 31373557-31373557 on Build GRCh38
rs956754058 Chr.20: 31373783-31373783 on Build GRCh38
rs1038446504 Chr.20: 31373469-31373469 on Build GRCh38
rs988052354 Chr.20: 31373482-31373482 on Build GRCh38
rs1025921359 Chr.20: 31373481-31373481 on Build GRCh38
rs758742517 Chr.20: 31373477-31373477 on Build GRCh38
rs943929457 Chr.20: 31373543-31373543 on Build GRCh38
rs887987581 Chr.20: 31373659-31373659 on Build GRCh38
rs987905023 Chr.20: 31373637-31373637 on Build GRCh38
rs969073625 Chr.20: 31373582-31373583 on Build GRCh38
rs896735138 Chr.20: 31373703-31373703 on Build GRCh38
rs986065495 Chr.20: 31373669-31373669 on Build GRCh38
rs1000512055 Chr.20: 31373794-31373794 on Build GRCh38
rs1032380353 Chr.20: 31373761-31373761 on Build GRCh38
rs979458473 Chr.20: 31373440-31373440 on Build GRCh38
rs911249375 Chr.20: 31373677-31373677 on Build GRCh38
rs748561694 Chr.20: 31373788-31373788 on Build GRCh38
rs1039794373 Chr.20: 31373548-31373548 on Build GRCh38
rs778149663 Chr.20: 31373498-31373498 on Build GRCh38
rs1013815182 Chr.20: 31373717-31373717 on Build GRCh38
rs1019419720 Chr.20: 31373800-31373800 on Build GRCh38
rs1048152088 Chr.20: 31373593-31373593 on Build GRCh38
rs896704456 Chr.20: 31373514-31373514 on Build GRCh38
rs748677603 Chr.20: 31373456-31373456 on Build GRCh38
rs1036500016 Chr.20: 31373700-31373700 on Build GRCh38
rs898694293 Chr.20: 31373472-31373472 on Build GRCh38
rs930135444 Chr.20: 31373725-31373725 on Build GRCh38
rs942704129 Chr.20: 31373682-31373682 on Build GRCh38
rs1024071040 Chr.20: 31373722-31373722 on Build GRCh38
rs912477010 Chr.20: 31373504-31373504 on Build GRCh38
rs1005048606 Chr.20: 31373683-31373683 on Build GRCh38

View Full Product Details