Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 38045272-38045696 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DLEC1
Entrez Gene ID
9940
Gene Name
deleted in lung and esophageal cancer 1
Gene Aliases
CFAP81, DLC-1, DLC1, F56
UniGene
Hs.714499
Gene Chromosome Location
Chr.3: 38038595-38122741 on Build GRCh38
Transcripts
NM_001321153.1 NM_007335.3 NM_007337.3 XM_006713438.3 XM_006713439.3 XM_006713440.3 XM_011534317.2 XM_011534318.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3373162 c.487G>C p.A163P Chr.3: 38045618-38045618 on Build GRCh38
4548512 c.448G>C p.E150Q Chr.3: 38045579-38045579 on Build GRCh38
2985767 c.485G>A p.R162Q Chr.3: 38045616-38045616 on Build GRCh38
3683111 c.520G>A p.A174T Chr.3: 38045651-38045651 on Build GRCh38
1044092 c.450A>C p.E150D Chr.3: 38045581-38045581 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs529903759 Chr.3: 38045427-38045427 on Build GRCh38
rs748274741 Chr.3: 38045510-38045510 on Build GRCh38
rs751864396 Chr.3: 38045434-38045434 on Build GRCh38
rs746671970 Chr.3: 38045625-38045625 on Build GRCh38
rs368394132 Chr.3: 38045649-38045649 on Build GRCh38
rs767914514 Chr.3: 38045654-38045654 on Build GRCh38
rs752945398 Chr.3: 38045637-38045637 on Build GRCh38
rs750548075 Chr.3: 38045441-38045441 on Build GRCh38
rs917102248 Chr.3: 38045569-38045569 on Build GRCh38
rs186625169 Chr.3: 38045546-38045546 on Build GRCh38
rs372850249 Chr.3: 38045522-38045522 on Build GRCh38
rs1005289043 Chr.3: 38045402-38045402 on Build GRCh38
rs752932471 Chr.3: 38045655-38045655 on Build GRCh38
rs7630340 Chr.3: 38045632-38045632 on Build GRCh38
rs778895093 Chr.3: 38045513-38045513 on Build GRCh38
rs117463277 Chr.3: 38045651-38045651 on Build GRCh38
rs895697043 Chr.3: 38045334-38045334 on Build GRCh38
rs543923488 Chr.3: 38045359-38045359 on Build GRCh38
rs1053442587 Chr.3: 38045326-38045326 on Build GRCh38
rs988628704 Chr.3: 38045560-38045560 on Build GRCh38
rs781635890 Chr.3: 38045486-38045486 on Build GRCh38
rs1044057584 Chr.3: 38045378-38045378 on Build GRCh38
rs373832144 Chr.3: 38045586-38045586 on Build GRCh38
rs760716991 Chr.3: 38045662-38045662 on Build GRCh38
rs748532240 Chr.3: 38045525-38045525 on Build GRCh38
rs754101978 Chr.3: 38045553-38045553 on Build GRCh38
rs778755536 Chr.3: 38045606-38045606 on Build GRCh38
rs190145969 Chr.3: 38045436-38045436 on Build GRCh38
rs369333925 Chr.3: 38045493-38045493 on Build GRCh38
rs756254536 Chr.3: 38045595-38045595 on Build GRCh38
rs36046028 Chr.3: 38045533-38045533 on Build GRCh38
rs560884619 Chr.3: 38045424-38045424 on Build GRCh38
rs370196668 Chr.3: 38045547-38045547 on Build GRCh38
rs878930280 Chr.3: 38045588-38045588 on Build GRCh38
rs746612922 Chr.3: 38045541-38045541 on Build GRCh38
rs769748995 Chr.3: 38045410-38045410 on Build GRCh38
rs750351143 Chr.3: 38045613-38045613 on Build GRCh38
rs763471746 Chr.3: 38045648-38045648 on Build GRCh38
rs375143949 Chr.3: 38045636-38045636 on Build GRCh38
rs1016715549 Chr.3: 38045415-38045415 on Build GRCh38
rs771233103 Chr.3: 38045650-38045650 on Build GRCh38
rs375033485 Chr.3: 38045393-38045393 on Build GRCh38
rs139958483 Chr.3: 38045674-38045674 on Build GRCh38
rs956394926 Chr.3: 38045473-38045473 on Build GRCh38
rs61672339 Chr.3: 38045428-38045428 on Build GRCh38
rs934540766 Chr.3: 38045299-38045299 on Build GRCh38
rs367791302 Chr.3: 38045455-38045455 on Build GRCh38
rs768241128 Chr.3: 38045626-38045626 on Build GRCh38
rs780810186 Chr.3: 38045630-38045630 on Build GRCh38
rs919217880 Chr.3: 38045409-38045409 on Build GRCh38
rs181422045 Chr.3: 38045464-38045464 on Build GRCh38
rs146035754 Chr.3: 38045668-38045668 on Build GRCh38
rs751855448 Chr.3: 38045581-38045581 on Build GRCh38
rs749240828 Chr.3: 38045503-38045503 on Build GRCh38
rs377465375 Chr.3: 38045615-38045615 on Build GRCh38
rs1014188794 Chr.3: 38045374-38045374 on Build GRCh38
rs1037682566 Chr.3: 38045349-38045349 on Build GRCh38
rs758286831 Chr.3: 38045669-38045669 on Build GRCh38
rs905547269 Chr.3: 38045400-38045400 on Build GRCh38
rs766745203 Chr.3: 38045577-38045577 on Build GRCh38
rs201980264 Chr.3: 38045592-38045592 on Build GRCh38
rs1017534729 Chr.3: 38045469-38045469 on Build GRCh38
rs201197582 Chr.3: 38045585-38045585 on Build GRCh38
rs762324276 Chr.3: 38045564-38045564 on Build GRCh38
rs779857875 Chr.3: 38045621-38045621 on Build GRCh38
rs371878341 Chr.3: 38045653-38045653 on Build GRCh38
rs569356843 Chr.3: 38045510-38045510 on Build GRCh38
rs568511981 Chr.3: 38045584-38045584 on Build GRCh38
rs763267155 Chr.3: 38045571-38045571 on Build GRCh38
rs773769116 Chr.3: 38045566-38045566 on Build GRCh38
rs755186945 Chr.3: 38045565-38045565 on Build GRCh38
rs775050978 Chr.3: 38045535-38045535 on Build GRCh38

View Full Product Details